Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205918795-205919040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC26A9
Entrez Gene ID
115019
Gene Name
solute carrier family 26 member 9
Gene Aliases
-
UniGene
Hs.164073
Gene Chromosome Location
Chr.1: 205913048-205943472 on Build GRCh38
Transcripts
NM_052934.3 NM_134325.2 XM_011509121.2 XM_011509122.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887181 c.2113C>A p.Q705K Chr.1: 205918983-205918983 on Build GRCh38
6781090 c.2230G>A p.V744M Chr.1: 205918866-205918866 on Build GRCh38
4771603 c.2237C>G p.P746R Chr.1: 205918859-205918859 on Build GRCh38
6781113 c.2191C>T p.H731Y Chr.1: 205918905-205918905 on Build GRCh38
2215572 c.2225G>A p.R742K Chr.1: 205918871-205918871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752033414 Chr.1: 205918908-205918908 on Build GRCh38
rs754475427 Chr.1: 205918945-205918945 on Build GRCh38
rs16856462 Chr.1: 205918853-205918853 on Build GRCh38
rs371716247 Chr.1: 205918937-205918937 on Build GRCh38
rs953779807 Chr.1: 205918994-205918994 on Build GRCh38
rs764624823 Chr.1: 205918907-205918907 on Build GRCh38
rs535977682 Chr.1: 205918879-205918879 on Build GRCh38
rs779732490 Chr.1: 205918929-205918929 on Build GRCh38
rs1027973210 Chr.1: 205918989-205918989 on Build GRCh38
rs746855221 Chr.1: 205918997-205918997 on Build GRCh38
rs767873155 Chr.1: 205918915-205918915 on Build GRCh38
rs1002366495 Chr.1: 205918974-205918974 on Build GRCh38
rs905648898 Chr.1: 205918970-205918970 on Build GRCh38
rs139528306 Chr.1: 205918920-205918920 on Build GRCh38
rs376675948 Chr.1: 205918828-205918828 on Build GRCh38
rs146098587 Chr.1: 205918854-205918854 on Build GRCh38
rs367640041 Chr.1: 205918900-205918900 on Build GRCh38
rs748808643 Chr.1: 205918849-205918849 on Build GRCh38
rs771922619 Chr.1: 205918961-205918961 on Build GRCh38
rs757563806 Chr.1: 205918911-205918911 on Build GRCh38
rs769228669 Chr.1: 205918833-205918833 on Build GRCh38
rs907398450 Chr.1: 205918871-205918871 on Build GRCh38
rs3811428 Chr.1: 205918866-205918866 on Build GRCh38
rs773133674 Chr.1: 205918962-205918962 on Build GRCh38
rs199967063 Chr.1: 205918837-205918837 on Build GRCh38
rs11240593 Chr.1: 205919015-205919015 on Build GRCh38
rs775623589 Chr.1: 205918899-205918899 on Build GRCh38
rs748048445 Chr.1: 205918954-205918954 on Build GRCh38
rs753937198 Chr.1: 205918888-205918889 on Build GRCh38
rs755752528 Chr.1: 205918921-205918921 on Build GRCh38
rs575418036 Chr.1: 205919019-205919019 on Build GRCh38
rs772014773 Chr.1: 205918867-205918867 on Build GRCh38
rs868242345 Chr.1: 205919003-205919003 on Build GRCh38
rs778860166 Chr.1: 205918953-205918953 on Build GRCh38
rs1004393006 Chr.1: 205918888-205918888 on Build GRCh38
rs759839133 Chr.1: 205918873-205918873 on Build GRCh38
rs1055331302 Chr.1: 205918986-205918986 on Build GRCh38

View Full Product Details