Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113338661-113338912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC12
Entrez Gene ID
54970
Gene Name
tetratricopeptide repeat domain 12
Gene Aliases
TPARM
UniGene
Hs.288772
Gene Chromosome Location
Chr.11: 113314529-113385952 on Build GRCh38
Transcripts
NM_001318533.1 NM_017868.3 XM_005271604.3 XM_011542890.2 XM_017017943.1 XM_017017944.1 XM_017017945.1 XM_017017946.1 XM_017017947.1 XM_017017948.1 XM_017017949.1 XM_017017950.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
923168 c.626C>T p.T209I Chr.11: 113338823-113338823 on Build GRCh38
5729031 c.637+1G>A p.? Chr.11: 113338835-113338835 on Build GRCh38
4395533 c.628C>T p.Q210* Chr.11: 113338825-113338825 on Build GRCh38
6810876 c.588T>G p.C196W Chr.11: 113338785-113338785 on Build GRCh38
4184437 c.604G>T p.E202* Chr.11: 113338801-113338801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782006805 Chr.11: 113338853-113338853 on Build GRCh38
rs782271400 Chr.11: 113338883-113338884 on Build GRCh38
rs782036468 Chr.11: 113338741-113338741 on Build GRCh38
rs782217398 Chr.11: 113338815-113338815 on Build GRCh38
rs782490661 Chr.11: 113338862-113338862 on Build GRCh38
rs782312903 Chr.11: 113338736-113338736 on Build GRCh38
rs891179405 Chr.11: 113338711-113338711 on Build GRCh38
rs369367049 Chr.11: 113338887-113338887 on Build GRCh38
rs370192944 Chr.11: 113338866-113338866 on Build GRCh38
rs782248659 Chr.11: 113338855-113338855 on Build GRCh38
rs781997349 Chr.11: 113338702-113338702 on Build GRCh38
rs781943440 Chr.11: 113338874-113338874 on Build GRCh38
rs782060952 Chr.11: 113338875-113338875 on Build GRCh38
rs955956796 Chr.11: 113338888-113338888 on Build GRCh38
rs781864891 Chr.11: 113338802-113338802 on Build GRCh38
rs782738243 Chr.11: 113338843-113338843 on Build GRCh38
rs782722946 Chr.11: 113338768-113338768 on Build GRCh38
rs1004264736 Chr.11: 113338723-113338723 on Build GRCh38
rs564888768 Chr.11: 113338692-113338692 on Build GRCh38
rs527591656 Chr.11: 113338693-113338693 on Build GRCh38
rs781866119 Chr.11: 113338760-113338760 on Build GRCh38
rs782631852 Chr.11: 113338811-113338811 on Build GRCh38
rs782520830 Chr.11: 113338807-113338807 on Build GRCh38
rs781881632 Chr.11: 113338800-113338800 on Build GRCh38
rs782788067 Chr.11: 113338759-113338759 on Build GRCh38
rs781798879 Chr.11: 113338771-113338771 on Build GRCh38
rs782149418 Chr.11: 113338746-113338746 on Build GRCh38
rs182513253 Chr.11: 113338775-113338775 on Build GRCh38
rs782094314 Chr.11: 113338763-113338763 on Build GRCh38
rs782066644 Chr.11: 113338728-113338728 on Build GRCh38
rs782688793 Chr.11: 113338784-113338784 on Build GRCh38
rs367770762 Chr.11: 113338766-113338766 on Build GRCh38
rs1015514735 Chr.11: 113338725-113338725 on Build GRCh38
rs782563892 Chr.11: 113338827-113338827 on Build GRCh38
rs567492345 Chr.11: 113338709-113338709 on Build GRCh38
rs200116175 Chr.11: 113338842-113338842 on Build GRCh38
rs10891536 Chr.11: 113338706-113338706 on Build GRCh38
rs782686940 Chr.11: 113338884-113338884 on Build GRCh38
rs782329289 Chr.11: 113338823-113338823 on Build GRCh38

View Full Product Details