Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23694577-23694850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL11
Entrez Gene ID
6135
Gene Name
ribosomal protein L11
Gene Aliases
DBA7, GIG34, L11
UniGene
Hs.719951
Gene Chromosome Location
Chr.1: 23691779-23696425 on Build GRCh38
Transcripts
NM_000975.3 NM_001199802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6274398 c.340G>T p.D114Y Chr.1: 23694735-23694735 on Build GRCh38
6214572 c.357T>G p.Y119* Chr.1: 23694752-23694752 on Build GRCh38
2049293 c.380G>A p.G127D Chr.1: 23694775-23694775 on Build GRCh38
6238116 c.291C>T p.N97N Chr.1: 23694686-23694686 on Build GRCh38
6062572 c.299C>T p.S100L Chr.1: 23694694-23694694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755457813 Chr.1: 23694808-23694809 on Build GRCh38
rs190679931 Chr.1: 23694606-23694606 on Build GRCh38
rs773506948 Chr.1: 23694748-23694748 on Build GRCh38
rs751955522 Chr.1: 23694619-23694621 on Build GRCh38
rs765671467 Chr.1: 23694624-23694624 on Build GRCh38
rs201640468 Chr.1: 23694762-23694762 on Build GRCh38
rs759798958 Chr.1: 23694644-23694644 on Build GRCh38
rs779597775 Chr.1: 23694697-23694697 on Build GRCh38
rs773770841 Chr.1: 23694626-23694626 on Build GRCh38
rs780742884 Chr.1: 23694822-23694822 on Build GRCh38
rs780832428 Chr.1: 23694720-23694720 on Build GRCh38
rs879004992 Chr.1: 23694655-23694655 on Build GRCh38
rs369675968 Chr.1: 23694813-23694813 on Build GRCh38
rs369949969 Chr.1: 23694682-23694682 on Build GRCh38
rs879071077 Chr.1: 23694657-23694657 on Build GRCh38
rs746549825 Chr.1: 23694698-23694698 on Build GRCh38
rs774450430 Chr.1: 23694773-23694773 on Build GRCh38
rs1017655517 Chr.1: 23694777-23694777 on Build GRCh38
rs771380294 Chr.1: 23694767-23694767 on Build GRCh38
rs759713829 Chr.1: 23694790-23694790 on Build GRCh38
rs1014769825 Chr.1: 23694612-23694612 on Build GRCh38
rs879073715 Chr.1: 23694785-23694785 on Build GRCh38
rs1804298 Chr.1: 23694727-23694727 on Build GRCh38
rs8880 Chr.1: 23694734-23694734 on Build GRCh38
rs371930769 Chr.1: 23694633-23694633 on Build GRCh38
rs768246944 Chr.1: 23694710-23694710 on Build GRCh38
rs1804299 Chr.1: 23694713-23694713 on Build GRCh38
rs938474010 Chr.1: 23694623-23694623 on Build GRCh38
rs751010573 Chr.1: 23694818-23694818 on Build GRCh38
rs11556036 Chr.1: 23694694-23694694 on Build GRCh38
rs755213760 Chr.1: 23694706-23694706 on Build GRCh38
rs113058387 Chr.1: 23694794-23694794 on Build GRCh38
rs201786327 Chr.1: 23694800-23694800 on Build GRCh38
rs1059937 Chr.1: 23694692-23694692 on Build GRCh38
rs535398404 Chr.1: 23694820-23694820 on Build GRCh38
rs1804296 Chr.1: 23694751-23694751 on Build GRCh38
rs1804297 Chr.1: 23694690-23694690 on Build GRCh38
rs767791246 Chr.1: 23694796-23694796 on Build GRCh38
rs199673309 Chr.1: 23694686-23694686 on Build GRCh38
rs749471113 Chr.1: 23694619-23694619 on Build GRCh38
rs757131066 Chr.1: 23694664-23694664 on Build GRCh38
rs1804300 Chr.1: 23694743-23694743 on Build GRCh38
rs373100789 Chr.1: 23694814-23694814 on Build GRCh38
rs765071374 Chr.1: 23694812-23694812 on Build GRCh38
rs951529962 Chr.1: 23694622-23694622 on Build GRCh38
rs770189198 Chr.1: 23694738-23694738 on Build GRCh38
rs778653081 Chr.1: 23694671-23694671 on Build GRCh38
rs369294702 Chr.1: 23694824-23694824 on Build GRCh38
rs753639398 Chr.1: 23694663-23694663 on Build GRCh38
rs763435650 Chr.1: 23694628-23694628 on Build GRCh38
rs761047900 Chr.1: 23694811-23694811 on Build GRCh38
rs886046309 Chr.1: 23694701-23694701 on Build GRCh38
rs374769720 Chr.1: 23694645-23694645 on Build GRCh38
rs372622822 Chr.1: 23694695-23694695 on Build GRCh38
rs886046308 Chr.1: 23694665-23694665 on Build GRCh38
rs779874957 Chr.1: 23694815-23694815 on Build GRCh38
rs751638388 Chr.1: 23694639-23694639 on Build GRCh38

View Full Product Details