Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 6923596-6923861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D14
Entrez Gene ID
57533
Gene Name
TBC1 domain family member 14
Gene Aliases
-
UniGene
Hs.518611
Gene Chromosome Location
Chr.4: 6909444-7033118 on Build GRCh38
Transcripts
NM_001113361.1 NM_020773.2 XM_005247986.2 XM_006713895.2 XM_011513507.2 XM_011513508.1 XM_017008477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797172 c.174C>T p.I58I Chr.4: 6923608-6923608 on Build GRCh38
1149925 c.428C>A p.P143Q Chr.4: 6923817-6923817 on Build GRCh38
5641667 c.299C>G p.P100R Chr.4: 6923733-6923733 on Build GRCh38
5641666 c.344C>G p.P115R Chr.4: 6923733-6923733 on Build GRCh38
359885 c.379A>T p.S127C Chr.4: 6923813-6923813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543358259 Chr.4: 6923762-6923762 on Build GRCh38
rs147348518 Chr.4: 6923758-6923758 on Build GRCh38
rs777084442 Chr.4: 6923668-6923668 on Build GRCh38
rs376311994 Chr.4: 6923782-6923782 on Build GRCh38
rs752220496 Chr.4: 6923616-6923616 on Build GRCh38
rs776959745 Chr.4: 6923728-6923728 on Build GRCh38
rs762393997 Chr.4: 6923671-6923671 on Build GRCh38
rs747713715 Chr.4: 6923817-6923817 on Build GRCh38
rs753567001 Chr.4: 6923752-6923752 on Build GRCh38
rs759191151 Chr.4: 6923827-6923827 on Build GRCh38
rs756133689 Chr.4: 6923637-6923637 on Build GRCh38
rs772814515 Chr.4: 6923820-6923820 on Build GRCh38
rs760378685 Chr.4: 6923748-6923748 on Build GRCh38
rs148550060 Chr.4: 6923815-6923815 on Build GRCh38
rs780068290 Chr.4: 6923638-6923638 on Build GRCh38
rs150525115 Chr.4: 6923705-6923705 on Build GRCh38
rs142720842 Chr.4: 6923743-6923743 on Build GRCh38
rs903958844 Chr.4: 6923628-6923628 on Build GRCh38
rs751635325 Chr.4: 6923841-6923841 on Build GRCh38
rs536762985 Chr.4: 6923680-6923680 on Build GRCh38
rs373638955 Chr.4: 6923776-6923776 on Build GRCh38
rs141049958 Chr.4: 6923763-6923763 on Build GRCh38
rs766440023 Chr.4: 6923700-6923700 on Build GRCh38
rs760466866 Chr.4: 6923823-6923823 on Build GRCh38
rs571166476 Chr.4: 6923669-6923669 on Build GRCh38
rs371738551 Chr.4: 6923699-6923699 on Build GRCh38
rs954232247 Chr.4: 6923717-6923717 on Build GRCh38
rs751829808 Chr.4: 6923783-6923783 on Build GRCh38
rs530223501 Chr.4: 6923636-6923636 on Build GRCh38
rs199606822 Chr.4: 6923642-6923642 on Build GRCh38
rs200567309 Chr.4: 6923837-6923837 on Build GRCh38
rs554489915 Chr.4: 6923732-6923732 on Build GRCh38
rs1029200917 Chr.4: 6923713-6923713 on Build GRCh38
rs764200583 Chr.4: 6923838-6923838 on Build GRCh38
rs771238502 Chr.4: 6923725-6923725 on Build GRCh38
rs758663978 Chr.4: 6923707-6923707 on Build GRCh38
rs751175855 Chr.4: 6923621-6923621 on Build GRCh38
rs553894612 Chr.4: 6923701-6923701 on Build GRCh38
rs144795768 Chr.4: 6923814-6923814 on Build GRCh38
rs761584212 Chr.4: 6923688-6923688 on Build GRCh38
rs746160356 Chr.4: 6923800-6923800 on Build GRCh38
rs117097571 Chr.4: 6923662-6923662 on Build GRCh38
rs375225757 Chr.4: 6923842-6923842 on Build GRCh38
rs757440529 Chr.4: 6923788-6923788 on Build GRCh38
rs141690496 Chr.4: 6923694-6923694 on Build GRCh38
rs34768850 Chr.4: 6923644-6923644 on Build GRCh38
rs145484437 Chr.4: 6923624-6923624 on Build GRCh38
rs763687892 Chr.4: 6923620-6923620 on Build GRCh38
rs935404049 Chr.4: 6923634-6923634 on Build GRCh38
rs115334884 Chr.4: 6923617-6923617 on Build GRCh38
rs368426774 Chr.4: 6923801-6923801 on Build GRCh38
rs770198522 Chr.4: 6923734-6923734 on Build GRCh38
rs773703122 Chr.4: 6923683-6923683 on Build GRCh38
rs565234468 Chr.4: 6923799-6923799 on Build GRCh38
rs577074529 Chr.4: 6923753-6923753 on Build GRCh38
rs773924583 Chr.4: 6923737-6923737 on Build GRCh38
rs142845338 Chr.4: 6923635-6923635 on Build GRCh38
rs772949671 Chr.4: 6923747-6923747 on Build GRCh38
rs766313548 Chr.4: 6923751-6923751 on Build GRCh38
rs572204539 Chr.4: 6923704-6923704 on Build GRCh38
rs772197062 Chr.4: 6923704-6923705 on Build GRCh38
rs772183488 Chr.4: 6923649-6923649 on Build GRCh38
rs151075047 Chr.4: 6923824-6923824 on Build GRCh38
rs765253296 Chr.4: 6923828-6923828 on Build GRCh38
rs370873478 Chr.4: 6923698-6923698 on Build GRCh38
rs748167132 Chr.4: 6923706-6923706 on Build GRCh38
rs141292073 Chr.4: 6923826-6923826 on Build GRCh38
rs781745652 Chr.4: 6923721-6923721 on Build GRCh38
rs747407757 Chr.4: 6923724-6923724 on Build GRCh38
rs375266892 Chr.4: 6923641-6923641 on Build GRCh38
rs142871950 Chr.4: 6923818-6923818 on Build GRCh38
rs570309392 Chr.4: 6923645-6923645 on Build GRCh38

View Full Product Details