Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19918027-19918300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005742.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3592007 c.1121G>A p.G374E Chr.3: 19918288-19918288 on Build GRCh38
7094602 c.1146C>T p.T382T Chr.3: 19918263-19918263 on Build GRCh38
5101953 c.1177+5delT p.? Chr.3: 19918227-19918227 on Build GRCh38
1617428 c.1114G>A p.A372T Chr.3: 19918295-19918295 on Build GRCh38
4680323 c.1166C>T p.A389V Chr.3: 19918243-19918243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141808957 Chr.3: 19918091-19918091 on Build GRCh38
rs776763743 Chr.3: 19918210-19918210 on Build GRCh38
rs964652814 Chr.3: 19918100-19918100 on Build GRCh38
rs2929366 Chr.3: 19918264-19918264 on Build GRCh38
rs768686614 Chr.3: 19918199-19918199 on Build GRCh38
rs1003081434 Chr.3: 19918077-19918077 on Build GRCh38
rs77241805 Chr.3: 19918084-19918084 on Build GRCh38
rs923341392 Chr.3: 19918127-19918127 on Build GRCh38
rs907365235 Chr.3: 19918062-19918062 on Build GRCh38
rs976981750 Chr.3: 19918220-19918220 on Build GRCh38
rs372861700 Chr.3: 19918190-19918190 on Build GRCh38
rs753163048 Chr.3: 19918261-19918261 on Build GRCh38
rs759940580 Chr.3: 19918256-19918256 on Build GRCh38
rs140954715 Chr.3: 19918270-19918270 on Build GRCh38
rs747963168 Chr.3: 19918215-19918216 on Build GRCh38
rs990342490 Chr.3: 19918093-19918093 on Build GRCh38
rs189219316 Chr.3: 19918061-19918061 on Build GRCh38
rs986273703 Chr.3: 19918156-19918156 on Build GRCh38
rs1014110873 Chr.3: 19918071-19918071 on Build GRCh38
rs764178909 Chr.3: 19918265-19918265 on Build GRCh38
rs772607418 Chr.3: 19918215-19918215 on Build GRCh38
rs111403113 Chr.3: 19918184-19918184 on Build GRCh38
rs979303561 Chr.3: 19918109-19918109 on Build GRCh38
rs1021598256 Chr.3: 19918098-19918098 on Build GRCh38
rs145670243 Chr.3: 19918258-19918258 on Build GRCh38
rs80119282 Chr.3: 19918227-19918227 on Build GRCh38
rs764937014 Chr.3: 19918272-19918272 on Build GRCh38
rs773308200 Chr.3: 19918216-19918217 on Build GRCh38
rs527790508 Chr.3: 19918235-19918235 on Build GRCh38
rs534260126 Chr.3: 19918216-19918216 on Build GRCh38
rs566831665 Chr.3: 19918051-19918051 on Build GRCh38
rs933400032 Chr.3: 19918132-19918132 on Build GRCh38
rs986685734 Chr.3: 19918178-19918178 on Build GRCh38
rs769278210 Chr.3: 19918211-19918211 on Build GRCh38
rs76211675 Chr.3: 19918226-19918226 on Build GRCh38
rs944201528 Chr.3: 19918204-19918204 on Build GRCh38
rs774665883 Chr.3: 19918251-19918251 on Build GRCh38
rs1027297252 Chr.3: 19918122-19918122 on Build GRCh38
rs961968029 Chr.3: 19918089-19918089 on Build GRCh38
rs953105610 Chr.3: 19918135-19918135 on Build GRCh38
rs143695761 Chr.3: 19918268-19918268 on Build GRCh38

View Full Product Details