Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48481293-48481522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTSJ1
Entrez Gene ID
24140
Gene Name
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases
CDLIV, JM23, MRX44, MRX9, SPB1, TRMT7
UniGene
Hs.23170
Gene Chromosome Location
Chr.X: 48476021-48486364 on Build GRCh38
Transcripts
NM_001282157.1 NM_012280.3 NM_177439.2 XM_005272595.1 XM_011543893.2 XM_011543894.2 XM_011543895.1 XM_017029382.1

SNP Detail

SNP ID Chromosome Location
rs374928864 Chr.X: 48481467-48481467 on Build GRCh38
rs782178418 Chr.X: 48481380-48481380 on Build GRCh38
rs797044780 Chr.X: 48481342-48481342 on Build GRCh38
rs782593946 Chr.X: 48481399-48481399 on Build GRCh38
rs781814995 Chr.X: 48481450-48481450 on Build GRCh38
rs782260696 Chr.X: 48481400-48481400 on Build GRCh38
rs377377919 Chr.X: 48481389-48481389 on Build GRCh38
rs782590411 Chr.X: 48481379-48481379 on Build GRCh38
rs781911440 Chr.X: 48481339-48481339 on Build GRCh38
rs201340333 Chr.X: 48481397-48481397 on Build GRCh38
rs34407506 Chr.X: 48481452-48481452 on Build GRCh38
rs781961176 Chr.X: 48481419-48481419 on Build GRCh38
rs782711243 Chr.X: 48481453-48481453 on Build GRCh38
rs200606065 Chr.X: 48481403-48481403 on Build GRCh38
rs1028523356 Chr.X: 48481393-48481393 on Build GRCh38
rs373151125 Chr.X: 48481463-48481463 on Build GRCh38
rs781813037 Chr.X: 48481318-48481318 on Build GRCh38
rs376189373 Chr.X: 48481411-48481411 on Build GRCh38
rs369250616 Chr.X: 48481436-48481436 on Build GRCh38
rs782785682 Chr.X: 48481438-48481438 on Build GRCh38
rs368351966 Chr.X: 48481333-48481333 on Build GRCh38
rs781977293 Chr.X: 48481440-48481440 on Build GRCh38
rs782308437 Chr.X: 48481418-48481418 on Build GRCh38
rs782644044 Chr.X: 48481483-48481483 on Build GRCh38
rs76994219 Chr.X: 48481486-48481486 on Build GRCh38
rs782541514 Chr.X: 48481367-48481367 on Build GRCh38
rs782064899 Chr.X: 48481416-48481416 on Build GRCh38
rs782167486 Chr.X: 48481446-48481446 on Build GRCh38
rs782075530 Chr.X: 48481386-48481386 on Build GRCh38
rs782805894 Chr.X: 48481448-48481448 on Build GRCh38
rs150788310 Chr.X: 48481335-48481335 on Build GRCh38

View Full Product Details