Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180950048-180950273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_001159710.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534651.2 XM_011534652.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4159824 c.1109C>A p.T370N Chr.5: 180950150-180950150 on Build GRCh38
1436742 c.1180C>T p.R394C Chr.5: 180950221-180950221 on Build GRCh38
4880961 c.1075C>T p.R359W Chr.5: 180950116-180950116 on Build GRCh38
482665 c.1076G>A p.R359Q Chr.5: 180950117-180950117 on Build GRCh38
6613603 c.1027T>C p.W343R Chr.5: 180950068-180950068 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752895838 Chr.5: 180950080-180950080 on Build GRCh38
rs377161453 Chr.5: 180950145-180950145 on Build GRCh38
rs544101054 Chr.5: 180950101-180950101 on Build GRCh38
rs778289052 Chr.5: 180950117-180950117 on Build GRCh38
rs868531369 Chr.5: 180950192-180950192 on Build GRCh38
rs547122671 Chr.5: 180950167-180950167 on Build GRCh38
rs761458116 Chr.5: 180950166-180950166 on Build GRCh38
rs757779355 Chr.5: 180950103-180950103 on Build GRCh38
rs997854641 Chr.5: 180950100-180950100 on Build GRCh38
rs369461521 Chr.5: 180950124-180950124 on Build GRCh38
rs373083701 Chr.5: 180950161-180950161 on Build GRCh38
rs775692069 Chr.5: 180950146-180950146 on Build GRCh38
rs778088974 Chr.5: 180950232-180950232 on Build GRCh38
rs199751211 Chr.5: 180950132-180950132 on Build GRCh38
rs758031578 Chr.5: 180950187-180950187 on Build GRCh38
rs938837722 Chr.5: 180950197-180950197 on Build GRCh38
rs746102922 Chr.5: 180950134-180950134 on Build GRCh38
rs938756261 Chr.5: 180950170-180950170 on Build GRCh38
rs764178692 Chr.5: 180950084-180950084 on Build GRCh38
rs927434600 Chr.5: 180950164-180950164 on Build GRCh38
rs370942451 Chr.5: 180950234-180950234 on Build GRCh38
rs370827491 Chr.5: 180950160-180950160 on Build GRCh38
rs141157357 Chr.5: 180950102-180950102 on Build GRCh38
rs770261754 Chr.5: 180950116-180950116 on Build GRCh38
rs369872041 Chr.5: 180950079-180950079 on Build GRCh38
rs759481966 Chr.5: 180950071-180950071 on Build GRCh38
rs753548010 Chr.5: 180950088-180950088 on Build GRCh38
rs750251421 Chr.5: 180950169-180950169 on Build GRCh38
rs757775137 Chr.5: 180950247-180950247 on Build GRCh38
rs374848638 Chr.5: 180950104-180950104 on Build GRCh38
rs772485260 Chr.5: 180950135-180950135 on Build GRCh38
rs754512426 Chr.5: 180950222-180950222 on Build GRCh38
rs570090091 Chr.5: 180950220-180950220 on Build GRCh38
rs760679093 Chr.5: 180950083-180950083 on Build GRCh38
rs200221003 Chr.5: 180950110-180950110 on Build GRCh38
rs376465401 Chr.5: 180950189-180950189 on Build GRCh38

View Full Product Details