Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 99903599-99903844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJB14
Entrez Gene ID
79982
Gene Name
DnaJ heat shock protein family (Hsp40) member B14
Gene Aliases
EGNR9427, PRO34683
UniGene
Hs.577426
Gene Chromosome Location
Chr.4: 99896248-99946726 on Build GRCh38
Transcripts
NM_001031723.3 NM_001278310.1 XM_011532262.2 XM_011532263.2 XM_011532265.2 XM_017008628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594157 c.985C>T p.R329* Chr.4: 99903756-99903756 on Build GRCh38
4120970 c.978T>C p.T326T Chr.4: 99903763-99903763 on Build GRCh38
4120974 c.922T>A p.Y308N Chr.4: 99903819-99903819 on Build GRCh38
1137116 c.1010A>G p.Q337R Chr.4: 99903731-99903731 on Build GRCh38
4120972 c.959T>C p.V320A Chr.4: 99903782-99903782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749827989 Chr.4: 99903719-99903719 on Build GRCh38
rs368485449 Chr.4: 99903679-99903679 on Build GRCh38
rs911506565 Chr.4: 99903626-99903626 on Build GRCh38
rs771075864 Chr.4: 99903811-99903811 on Build GRCh38
rs576417920 Chr.4: 99903727-99903727 on Build GRCh38
rs186252908 Chr.4: 99903653-99903653 on Build GRCh38
rs765991734 Chr.4: 99903722-99903722 on Build GRCh38
rs752480776 Chr.4: 99903686-99903686 on Build GRCh38
rs756764955 Chr.4: 99903705-99903705 on Build GRCh38
rs779256174 Chr.4: 99903755-99903755 on Build GRCh38
rs374557022 Chr.4: 99903706-99903706 on Build GRCh38
rs112067684 Chr.4: 99903688-99903688 on Build GRCh38
rs1047502999 Chr.4: 99903704-99903704 on Build GRCh38
rs1031197645 Chr.4: 99903803-99903803 on Build GRCh38
rs545420966 Chr.4: 99903674-99903674 on Build GRCh38
rs747099781 Chr.4: 99903787-99903787 on Build GRCh38
rs757385758 Chr.4: 99903768-99903768 on Build GRCh38
rs889662531 Chr.4: 99903666-99903666 on Build GRCh38
rs748510027 Chr.4: 99903764-99903764 on Build GRCh38
rs758126461 Chr.4: 99903687-99903687 on Build GRCh38
rs751086167 Chr.4: 99903690-99903690 on Build GRCh38
rs35086003 Chr.4: 99903639-99903640 on Build GRCh38
rs149086010 Chr.4: 99903784-99903784 on Build GRCh38
rs1044330763 Chr.4: 99903804-99903804 on Build GRCh38

View Full Product Details