Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44722003-44722264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPDA2
Entrez Gene ID
132789
Gene Name
glucosamine-6-phosphate deaminase 2
Gene Aliases
GNP2, SB52
UniGene
Hs.21398
Gene Chromosome Location
Chr.4: 44701795-44726634 on Build GRCh38
Transcripts
NM_001270880.1 NM_001270881.1 NM_138335.2 NR_073094.1 NR_073095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158912 c.60C>T p.I20I Chr.4: 44722148-44722148 on Build GRCh38
5736961 c.40G>A p.E14K Chr.4: 44722168-44722168 on Build GRCh38
3825895 c.16C>T p.L6F Chr.4: 44722192-44722192 on Build GRCh38
6672435 c.50C>T p.A17V Chr.4: 44722158-44722158 on Build GRCh38
4808495 c.21T>A p.D7E Chr.4: 44722187-44722187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145396953 Chr.4: 44722141-44722141 on Build GRCh38
rs751819125 Chr.4: 44722181-44722181 on Build GRCh38
rs760020359 Chr.4: 44722147-44722147 on Build GRCh38
rs770410949 Chr.4: 44722140-44722140 on Build GRCh38
rs964923132 Chr.4: 44722059-44722059 on Build GRCh38
rs771947021 Chr.4: 44722120-44722120 on Build GRCh38
rs369601120 Chr.4: 44722058-44722058 on Build GRCh38
rs911343338 Chr.4: 44722065-44722065 on Build GRCh38
rs754899233 Chr.4: 44722211-44722211 on Build GRCh38
rs942713857 Chr.4: 44722088-44722088 on Build GRCh38
rs1018211324 Chr.4: 44722051-44722051 on Build GRCh38
rs1027416172 Chr.4: 44722089-44722089 on Build GRCh38
rs758167070 Chr.4: 44722037-44722037 on Build GRCh38
rs767640240 Chr.4: 44722206-44722206 on Build GRCh38
rs757568346 Chr.4: 44722188-44722188 on Build GRCh38
rs373637467 Chr.4: 44722216-44722216 on Build GRCh38
rs757410390 Chr.4: 44722072-44722072 on Build GRCh38
rs200496880 Chr.4: 44722114-44722114 on Build GRCh38
rs772815722 Chr.4: 44722121-44722121 on Build GRCh38
rs775874824 Chr.4: 44722154-44722154 on Build GRCh38
rs765693828 Chr.4: 44722150-44722150 on Build GRCh38
rs763197890 Chr.4: 44722155-44722155 on Build GRCh38
rs781481085 Chr.4: 44722073-44722073 on Build GRCh38
rs764620194 Chr.4: 44722175-44722175 on Build GRCh38
rs746727657 Chr.4: 44722127-44722127 on Build GRCh38
rs368788777 Chr.4: 44722145-44722145 on Build GRCh38
rs751871507 Chr.4: 44722069-44722069 on Build GRCh38
rs750502303 Chr.4: 44722210-44722210 on Build GRCh38
rs760778655 Chr.4: 44722217-44722217 on Build GRCh38
rs370534654 Chr.4: 44722043-44722043 on Build GRCh38
rs950039042 Chr.4: 44722201-44722201 on Build GRCh38
rs749181971 Chr.4: 44722096-44722096 on Build GRCh38
rs779062583 Chr.4: 44722053-44722053 on Build GRCh38
rs754878043 Chr.4: 44722109-44722109 on Build GRCh38
rs764201855 Chr.4: 44722066-44722066 on Build GRCh38
rs141733877 Chr.4: 44722112-44722112 on Build GRCh38

View Full Product Details