Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 113844532-113844785 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAMD1C
Entrez Gene ID
54762
Gene Name
GRAM domain containing 1C
Gene Aliases
-
UniGene
Hs.24583
Gene Chromosome Location
Chr.3: 113828182-113947174 on Build GRCh38
Transcripts
NM_017577.4 XM_005247546.2 XM_005247547.1 XM_011512930.1 XM_017006646.1 XM_017006647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5486306 c.136C>T p.P46S Chr.3: 113844611-113844611 on Build GRCh38
1036566 c.132G>A p.Q44Q Chr.3: 113844607-113844607 on Build GRCh38
1036565 c.61G>A p.D21N Chr.3: 113844536-113844536 on Build GRCh38
4642710 c.74A>G p.D25G Chr.3: 113844549-113844549 on Build GRCh38
6162636 c.133G>T p.G45W Chr.3: 113844608-113844608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778649929 Chr.3: 113844700-113844700 on Build GRCh38
rs571871598 Chr.3: 113844598-113844599 on Build GRCh38
rs756557871 Chr.3: 113844675-113844675 on Build GRCh38
rs767889803 Chr.3: 113844695-113844696 on Build GRCh38
rs759361836 Chr.3: 113844648-113844648 on Build GRCh38
rs941692973 Chr.3: 113844585-113844585 on Build GRCh38
rs766282265 Chr.3: 113844606-113844607 on Build GRCh38
rs2033380 Chr.3: 113844667-113844667 on Build GRCh38
rs957359779 Chr.3: 113844726-113844726 on Build GRCh38
rs754137408 Chr.3: 113844722-113844722 on Build GRCh38
rs894738820 Chr.3: 113844649-113844649 on Build GRCh38
rs781684434 Chr.3: 113844557-113844557 on Build GRCh38
rs761336771 Chr.3: 113844603-113844603 on Build GRCh38
rs747973964 Chr.3: 113844693-113844694 on Build GRCh38
rs887481849 Chr.3: 113844580-113844580 on Build GRCh38
rs780745325 Chr.3: 113844681-113844681 on Build GRCh38
rs773132771 Chr.3: 113844612-113844612 on Build GRCh38
rs769162987 Chr.3: 113844697-113844697 on Build GRCh38
rs990567876 Chr.3: 113844739-113844739 on Build GRCh38
rs760520551 Chr.3: 113844617-113844617 on Build GRCh38
rs369374562 Chr.3: 113844572-113844572 on Build GRCh38
rs775687623 Chr.3: 113844700-113844701 on Build GRCh38
rs746367958 Chr.3: 113844564-113844564 on Build GRCh38
rs774176282 Chr.3: 113844600-113844600 on Build GRCh38
rs749630667 Chr.3: 113844694-113844694 on Build GRCh38
rs759633360 Chr.3: 113844694-113844695 on Build GRCh38
rs202089640 Chr.3: 113844569-113844569 on Build GRCh38
rs190950009 Chr.3: 113844666-113844666 on Build GRCh38
rs764253873 Chr.3: 113844650-113844650 on Build GRCh38
rs143046502 Chr.3: 113844590-113844590 on Build GRCh38
rs1017496878 Chr.3: 113844745-113844745 on Build GRCh38
rs751796938 Chr.3: 113844665-113844665 on Build GRCh38
rs771492632 Chr.3: 113844610-113844610 on Build GRCh38
rs998980239 Chr.3: 113844721-113844721 on Build GRCh38
rs145377695 Chr.3: 113844701-113844701 on Build GRCh38
rs753603270 Chr.3: 113844643-113844643 on Build GRCh38
rs552290581 Chr.3: 113844621-113844621 on Build GRCh38

View Full Product Details