Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12696084-12696321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW9
Entrez Gene ID
84261
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Fbw9
UniGene
Hs.515154
Gene Chromosome Location
Chr.19: 12688916-12696900 on Build GRCh38
Transcripts
NM_032301.2 XM_005260096.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4140271 c.339C>G p.L113L Chr.19: 12696243-12696243 on Build GRCh38
4140270 c.339C>G p.L113L Chr.19: 12696243-12696243 on Build GRCh38
3756335 c.330C>G p.L110L Chr.19: 12696252-12696252 on Build GRCh38
3756336 c.330C>G p.L110L Chr.19: 12696252-12696252 on Build GRCh38
5643128 c.341T>C p.V114A Chr.19: 12696241-12696241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1016013746 Chr.19: 12696105-12696105 on Build GRCh38
rs868516820 Chr.19: 12696292-12696292 on Build GRCh38
rs868025901 Chr.19: 12696248-12696248 on Build GRCh38
rs774288718 Chr.19: 12696235-12696235 on Build GRCh38
rs769950410 Chr.19: 12696287-12696287 on Build GRCh38
rs763650958 Chr.19: 12696153-12696153 on Build GRCh38
rs868168653 Chr.19: 12696257-12696257 on Build GRCh38
rs928234753 Chr.19: 12696139-12696139 on Build GRCh38
rs553892898 Chr.19: 12696136-12696149 on Build GRCh38
rs368929567 Chr.19: 12696194-12696194 on Build GRCh38
rs374984061 Chr.19: 12696211-12696211 on Build GRCh38
rs763810884 Chr.19: 12696256-12696256 on Build GRCh38
rs758872905 Chr.19: 12696193-12696193 on Build GRCh38
rs1045139100 Chr.19: 12696134-12696154 on Build GRCh38
rs747343374 Chr.19: 12696206-12696206 on Build GRCh38
rs769980960 Chr.19: 12696221-12696221 on Build GRCh38
rs935160765 Chr.19: 12696143-12696143 on Build GRCh38
rs755719320 Chr.19: 12696132-12696132 on Build GRCh38
rs775307628 Chr.19: 12696250-12696250 on Build GRCh38
rs139535728 Chr.19: 12696156-12696156 on Build GRCh38
rs752062927 Chr.19: 12696282-12696282 on Build GRCh38
rs973418747 Chr.19: 12696106-12696106 on Build GRCh38
rs548297496 Chr.19: 12696296-12696296 on Build GRCh38
rs929558780 Chr.19: 12696133-12696133 on Build GRCh38
rs984087728 Chr.19: 12696134-12696134 on Build GRCh38
rs753453083 Chr.19: 12696271-12696271 on Build GRCh38
rs376213981 Chr.19: 12696291-12696291 on Build GRCh38
rs568421602 Chr.19: 12696212-12696212 on Build GRCh38
rs749383428 Chr.19: 12696230-12696230 on Build GRCh38
rs759256928 Chr.19: 12696242-12696242 on Build GRCh38
rs756730283 Chr.19: 12696275-12696275 on Build GRCh38
rs767091197 Chr.19: 12696127-12696127 on Build GRCh38
rs775938029 Chr.19: 12696149-12696150 on Build GRCh38
rs369109827 Chr.19: 12696231-12696231 on Build GRCh38
rs752333341 Chr.19: 12696129-12696129 on Build GRCh38
rs867996943 Chr.19: 12696266-12696266 on Build GRCh38
rs894897654 Chr.19: 12696192-12696192 on Build GRCh38
rs371715059 Chr.19: 12696207-12696207 on Build GRCh38
rs781202836 Chr.19: 12696134-12696140 on Build GRCh38
rs6511832 Chr.19: 12696252-12696252 on Build GRCh38
rs766797950 Chr.19: 12696279-12696279 on Build GRCh38
rs530002089 Chr.19: 12696295-12696295 on Build GRCh38
rs769844221 Chr.19: 12696150-12696156 on Build GRCh38
rs773320018 Chr.19: 12696225-12696225 on Build GRCh38
rs755318458 Chr.19: 12696288-12696288 on Build GRCh38
rs988897546 Chr.19: 12696147-12696147 on Build GRCh38

View Full Product Details