Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148404748-148405017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO38
Entrez Gene ID
81545
Gene Name
F-box protein 38
Gene Aliases
Fbx38, HMN2D, MOKA, SP329
UniGene
Hs.483772
Gene Chromosome Location
Chr.5: 148383935-148442836 on Build GRCh38
Transcripts
NM_001271723.1 NM_030793.4 XM_005268513.1 XM_006714797.1 XM_011537684.2 XM_017009899.1 XM_017009900.1 XM_017009901.1 XM_017009902.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389102 c.728C>T p.A243V Chr.5: 148404820-148404820 on Build GRCh38
1064031 c.692G>A p.C231Y Chr.5: 148404784-148404784 on Build GRCh38
4762431 c.717G>C p.M239I Chr.5: 148404809-148404809 on Build GRCh38
4506131 c.711C>T p.F237F Chr.5: 148404803-148404803 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs939407907 Chr.5: 148404955-148404955 on Build GRCh38
rs192946868 Chr.5: 148404930-148404930 on Build GRCh38
rs927937435 Chr.5: 148404936-148404936 on Build GRCh38
rs374815096 Chr.5: 148404936-148404937 on Build GRCh38
rs763481232 Chr.5: 148404780-148404780 on Build GRCh38
rs905002871 Chr.5: 148404865-148404866 on Build GRCh38
rs758427115 Chr.5: 148404827-148404827 on Build GRCh38
rs200459971 Chr.5: 148404965-148404966 on Build GRCh38
rs774425297 Chr.5: 148404788-148404788 on Build GRCh38
rs368329665 Chr.5: 148404826-148404826 on Build GRCh38
rs754487582 Chr.5: 148404841-148404841 on Build GRCh38
rs1034888665 Chr.5: 148404925-148404925 on Build GRCh38
rs144011381 Chr.5: 148404794-148404794 on Build GRCh38
rs769447903 Chr.5: 148404864-148404864 on Build GRCh38
rs771350091 Chr.5: 148404893-148404893 on Build GRCh38
rs780731913 Chr.5: 148404848-148404848 on Build GRCh38
rs144890577 Chr.5: 148404804-148404804 on Build GRCh38
rs116116831 Chr.5: 148404887-148404887 on Build GRCh38
rs370799682 Chr.5: 148404799-148404799 on Build GRCh38
rs945595652 Chr.5: 148404921-148404921 on Build GRCh38
rs915412031 Chr.5: 148404911-148404911 on Build GRCh38
rs951324789 Chr.5: 148404926-148404926 on Build GRCh38
rs780200903 Chr.5: 148404835-148404835 on Build GRCh38
rs989572991 Chr.5: 148404900-148404900 on Build GRCh38
rs532564746 Chr.5: 148404933-148404933 on Build GRCh38
rs751540446 Chr.5: 148404836-148404836 on Build GRCh38
rs547297676 Chr.5: 148404854-148404854 on Build GRCh38
rs973471162 Chr.5: 148404986-148404986 on Build GRCh38
rs199969692 Chr.5: 148404803-148404803 on Build GRCh38

View Full Product Details