Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 117508995-117509224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFTR
Entrez Gene ID
1080
Gene Name
cystic fibrosis transmembrane conductance regulator
Gene Aliases
ABC35, ABCC7, CF, CFTR/MRP, MRP7, TNR-CFTR, dJ760C5.1
UniGene
Hs.489786
Gene Chromosome Location
Chr.7: 117478367-117668665 on Build GRCh38
Transcripts
NM_000492.3 XM_011515751.2 XM_011515753.1 XM_011515754.2 XM_017011699.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454698 c.205C>T p.L69F Chr.7: 117509074-117509074 on Build GRCh38
5043957 c.249C>G p.F83L Chr.7: 117509118-117509118 on Build GRCh38
6175952 c.210T>A p.I70I Chr.7: 117509079-117509079 on Build GRCh38
1248176 c.227_228insT p.W79fs*32 Chr.7: 117509096-117509097 on Build GRCh38
7361064 c.178G>C p.E60Q Chr.7: 117509047-117509047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs397508412 Chr.7: 117509132-117509132 on Build GRCh38
rs397508367 Chr.7: 117509102-117509102 on Build GRCh38
rs569426839 Chr.7: 117509049-117509049 on Build GRCh38
rs372421038 Chr.7: 117509074-117509074 on Build GRCh38
rs780975618 Chr.7: 117509118-117509118 on Build GRCh38
rs749432776 Chr.7: 117509125-117509125 on Build GRCh38
rs762298973 Chr.7: 117509086-117509086 on Build GRCh38
rs397508249 Chr.7: 117509033-117509033 on Build GRCh38
rs77284892 Chr.7: 117509047-117509047 on Build GRCh38
rs397508348 Chr.7: 117509086-117509086 on Build GRCh38
rs397508360 Chr.7: 117509096-117509097 on Build GRCh38
rs750256100 Chr.7: 117509167-117509167 on Build GRCh38
rs397508347 Chr.7: 117509084-117509084 on Build GRCh38
rs1003610229 Chr.7: 117509081-117509081 on Build GRCh38
rs397508332 Chr.7: 117509071-117509071 on Build GRCh38
rs121908791 Chr.7: 117509143-117509143 on Build GRCh38
rs751305135 Chr.7: 117509102-117509102 on Build GRCh38
rs397508371 Chr.7: 117509105-117509105 on Build GRCh38
rs397508418 Chr.7: 117509135-117509135 on Build GRCh38
rs397508295 Chr.7: 117509043-117509046 on Build GRCh38
rs397508250 Chr.7: 117509032-117509032 on Build GRCh38
rs1053857312 Chr.7: 117509045-117509045 on Build GRCh38
rs397508362 Chr.7: 117509097-117509098 on Build GRCh38
rs763513110 Chr.7: 117509158-117509158 on Build GRCh38
rs1800076 Chr.7: 117509093-117509093 on Build GRCh38
rs115545701 Chr.7: 117509089-117509089 on Build GRCh38
rs121909025 Chr.7: 117509040-117509040 on Build GRCh38
rs368128542 Chr.7: 117509165-117509165 on Build GRCh38
rs149662778 Chr.7: 117509133-117509133 on Build GRCh38
rs1033366496 Chr.7: 117509085-117509085 on Build GRCh38
rs772459228 Chr.7: 117509190-117509190 on Build GRCh38
rs780026510 Chr.7: 117509174-117509174 on Build GRCh38
rs121908769 Chr.7: 117509131-117509132 on Build GRCh38
rs387906374 Chr.7: 117509146-117509146 on Build GRCh38
rs893977457 Chr.7: 117509046-117509046 on Build GRCh38
rs397508285 Chr.7: 117509041-117509041 on Build GRCh38
rs397508403 Chr.7: 117509128-117509128 on Build GRCh38
rs142540482 Chr.7: 117509090-117509090 on Build GRCh38
rs773739166 Chr.7: 117509142-117509142 on Build GRCh38
rs756022772 Chr.7: 117509173-117509173 on Build GRCh38
rs777536750 Chr.7: 117509097-117509097 on Build GRCh38
rs371983339 Chr.7: 117509177-117509177 on Build GRCh38
rs397508366 Chr.7: 117509102-117509103 on Build GRCh38
rs757959325 Chr.7: 117509095-117509095 on Build GRCh38
rs780924647 Chr.7: 117509166-117509166 on Build GRCh38
rs397508256 Chr.7: 117509035-117509035 on Build GRCh38
rs558515047 Chr.7: 117509155-117509155 on Build GRCh38
rs397508248 Chr.7: 117509024-117509024 on Build GRCh38
rs397508269 Chr.7: 117509037-117509037 on Build GRCh38
rs374992300 Chr.7: 117509127-117509127 on Build GRCh38
rs769754499 Chr.7: 117509120-117509120 on Build GRCh38
rs397508272 Chr.7: 117509038-117509038 on Build GRCh38
rs774558645 Chr.7: 117509083-117509083 on Build GRCh38
rs769807498 Chr.7: 117509063-117509063 on Build GRCh38
rs121908749 Chr.7: 117509092-117509092 on Build GRCh38
rs754147777 Chr.7: 117509128-117509129 on Build GRCh38
rs111155389 Chr.7: 117509142-117509143 on Build GRCh38
rs397508421 Chr.7: 117509138-117509138 on Build GRCh38
rs375214586 Chr.7: 117509179-117509179 on Build GRCh38
rs397508279 Chr.7: 117509039-117509039 on Build GRCh38
rs397508294 Chr.7: 117509043-117509044 on Build GRCh38
rs121908750 Chr.7: 117509140-117509140 on Build GRCh38
rs397508335 Chr.7: 117509073-117509073 on Build GRCh38
rs369028441 Chr.7: 117509182-117509182 on Build GRCh38
rs75961395 Chr.7: 117509123-117509123 on Build GRCh38
rs745756794 Chr.7: 117509119-117509119 on Build GRCh38
rs397508370 Chr.7: 117509104-117509104 on Build GRCh38
rs397508291 Chr.7: 117509042-117509042 on Build GRCh38
rs748356123 Chr.7: 117509184-117509184 on Build GRCh38
rs1800075 Chr.7: 117509082-117509082 on Build GRCh38
rs751161544 Chr.7: 117509162-117509162 on Build GRCh38
rs74467662 Chr.7: 117509145-117509145 on Build GRCh38
rs200337193 Chr.7: 117509031-117509031 on Build GRCh38
rs747749819 Chr.7: 117509025-117509025 on Build GRCh38
rs752414386 Chr.7: 117509148-117509148 on Build GRCh38
rs368505753 Chr.7: 117509069-117509069 on Build GRCh38

View Full Product Details