Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83643584-83643813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDNK
Entrez Gene ID
414328
Gene Name
IDNK, gluconokinase
Gene Aliases
C9orf103, bA522I20.2, hGntK
UniGene
Hs.530261
Gene Chromosome Location
Chr.9: 83622324-83644130 on Build GRCh38
Transcripts
NM_001001551.3 NM_001256915.1 NR_046421.1 NR_046422.1 XM_006717110.3 XM_006717111.3 XM_017014719.1 XM_017014724.1 XM_017014725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5616157 c.253A>T p.I85F Chr.9: 83643607-83643607 on Build GRCh38
7282526 c.530G>A p.R177H Chr.9: 83643617-83643617 on Build GRCh38
5616156 c.391A>T p.I131F Chr.9: 83643607-83643607 on Build GRCh38
6900478 c.344C>T p.P115L Chr.9: 83643698-83643698 on Build GRCh38
7255932 c.507G>C p.G169G Chr.9: 83643594-83643594 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759610956 Chr.9: 83643720-83643720 on Build GRCh38
rs144600259 Chr.9: 83643765-83643765 on Build GRCh38
rs149306982 Chr.9: 83643742-83643742 on Build GRCh38
rs943380361 Chr.9: 83643785-83643785 on Build GRCh38
rs763164622 Chr.9: 83643708-83643708 on Build GRCh38
rs756205307 Chr.9: 83643779-83643779 on Build GRCh38
rs150432950 Chr.9: 83643786-83643786 on Build GRCh38
rs781341586 Chr.9: 83643676-83643676 on Build GRCh38
rs1034341649 Chr.9: 83643625-83643626 on Build GRCh38
rs754650787 Chr.9: 83643773-83643773 on Build GRCh38
rs752249360 Chr.9: 83643614-83643614 on Build GRCh38
rs770481579 Chr.9: 83643689-83643689 on Build GRCh38
rs781517585 Chr.9: 83643639-83643639 on Build GRCh38
rs145886629 Chr.9: 83643605-83643605 on Build GRCh38
rs749987799 Chr.9: 83643617-83643617 on Build GRCh38
rs1010586329 Chr.9: 83643638-83643638 on Build GRCh38
rs201732497 Chr.9: 83643649-83643649 on Build GRCh38
rs549933426 Chr.9: 83643638-83643639 on Build GRCh38
rs143478756 Chr.9: 83643603-83643603 on Build GRCh38
rs753634538 Chr.9: 83643724-83643724 on Build GRCh38
rs569552366 Chr.9: 83643715-83643715 on Build GRCh38
rs780883418 Chr.9: 83643774-83643774 on Build GRCh38
rs138577393 Chr.9: 83643658-83643658 on Build GRCh38
rs540611979 Chr.9: 83643738-83643739 on Build GRCh38
rs757876395 Chr.9: 83643622-83643622 on Build GRCh38
rs767775643 Chr.9: 83643722-83643722 on Build GRCh38
rs1052690 Chr.9: 83643770-83643770 on Build GRCh38
rs757927745 Chr.9: 83643758-83643758 on Build GRCh38
rs778813883 Chr.9: 83643616-83643616 on Build GRCh38
rs748537708 Chr.9: 83643681-83643681 on Build GRCh38
rs750323425 Chr.9: 83643752-83643752 on Build GRCh38
rs773831604 Chr.9: 83643697-83643697 on Build GRCh38
rs774648107 Chr.9: 83643719-83643719 on Build GRCh38
rs769478824 Chr.9: 83643781-83643801 on Build GRCh38
rs761651473 Chr.9: 83643729-83643729 on Build GRCh38
rs551624003 Chr.9: 83643629-83643629 on Build GRCh38

View Full Product Details