Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134913466-134913731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCN1
Entrez Gene ID
2219
Gene Name
ficolin 1
Gene Aliases
FCNM
UniGene
Hs.440898
Gene Chromosome Location
Chr.9: 134909585-134917960 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
487205 c.332G>C p.C111S Chr.9: 134913589-134913589 on Build GRCh38
3214476 c.323C>T p.S108F Chr.9: 134913598-134913598 on Build GRCh38
5957797 c.336G>A p.A112A Chr.9: 134913585-134913585 on Build GRCh38
297989 c.322T>A p.S108T Chr.9: 134913599-134913599 on Build GRCh38
3655507 c.308G>A p.G103E Chr.9: 134913613-134913613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749646335 Chr.9: 134913586-134913586 on Build GRCh38
rs993958826 Chr.9: 134913553-134913553 on Build GRCh38
rs148406811 Chr.9: 134913591-134913591 on Build GRCh38
rs1014350897 Chr.9: 134913539-134913539 on Build GRCh38
rs762622237 Chr.9: 134913547-134913547 on Build GRCh38
rs932309073 Chr.9: 134913492-134913492 on Build GRCh38
rs1043397045 Chr.9: 134913505-134913505 on Build GRCh38
rs144427314 Chr.9: 134913585-134913585 on Build GRCh38
rs775284331 Chr.9: 134913620-134913620 on Build GRCh38
rs374659908 Chr.9: 134913671-134913671 on Build GRCh38
rs376398394 Chr.9: 134913557-134913557 on Build GRCh38
rs767065050 Chr.9: 134913654-134913654 on Build GRCh38
rs759403505 Chr.9: 134913538-134913538 on Build GRCh38
rs200685680 Chr.9: 134913558-134913558 on Build GRCh38
rs977481331 Chr.9: 134913643-134913643 on Build GRCh38
rs200053918 Chr.9: 134913592-134913592 on Build GRCh38
rs144127766 Chr.9: 134913609-134913609 on Build GRCh38
rs149699377 Chr.9: 134913581-134913581 on Build GRCh38
rs55862654 Chr.9: 134913516-134913516 on Build GRCh38
rs372066467 Chr.9: 134913540-134913540 on Build GRCh38
rs773940213 Chr.9: 134913647-134913647 on Build GRCh38
rs770939464 Chr.9: 134913530-134913530 on Build GRCh38
rs535207786 Chr.9: 134913510-134913510 on Build GRCh38
rs942655237 Chr.9: 134913677-134913677 on Build GRCh38
rs756573092 Chr.9: 134913659-134913659 on Build GRCh38
rs368051850 Chr.9: 134913665-134913665 on Build GRCh38
rs776770144 Chr.9: 134913535-134913535 on Build GRCh38
rs762049552 Chr.9: 134913608-134913608 on Build GRCh38
rs761353382 Chr.9: 134913651-134913651 on Build GRCh38
rs1041095980 Chr.9: 134913683-134913683 on Build GRCh38
rs371593172 Chr.9: 134913669-134913669 on Build GRCh38
rs145530502 Chr.9: 134913662-134913662 on Build GRCh38
rs749960808 Chr.9: 134913655-134913655 on Build GRCh38
rs922261251 Chr.9: 134913705-134913705 on Build GRCh38
rs911150046 Chr.9: 134913486-134913486 on Build GRCh38
rs756775998 Chr.9: 134913565-134913565 on Build GRCh38
rs895618308 Chr.9: 134913531-134913531 on Build GRCh38
rs904422757 Chr.9: 134913515-134913515 on Build GRCh38
rs752560461 Chr.9: 134913546-134913546 on Build GRCh38
rs55694974 Chr.9: 134913632-134913632 on Build GRCh38
rs376671452 Chr.9: 134913638-134913638 on Build GRCh38
rs369810978 Chr.9: 134913570-134913570 on Build GRCh38
rs376494851 Chr.9: 134913533-134913533 on Build GRCh38

View Full Product Details