Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154678119-154678360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB3
Entrez Gene ID
139716
Gene Name
GRB2 associated binding protein 3
Gene Aliases
-
UniGene
Hs.496982
Gene Chromosome Location
Chr.X: 154675249-154751583 on Build GRCh38
Transcripts
NM_001081573.2 NM_001282283.1 NM_080612.3 NR_104114.1 XM_005274648.1 XM_006724804.1 XM_011531103.1 XM_011531105.1 XM_011531106.1 XM_017029276.1

SNP Detail

SNP ID Chromosome Location
rs781924983 Chr.X: 154678145-154678145 on Build GRCh38
rs782473857 Chr.X: 154678306-154678306 on Build GRCh38
rs782188313 Chr.X: 154678320-154678320 on Build GRCh38
rs782741950 Chr.X: 154678181-154678181 on Build GRCh38
rs782453483 Chr.X: 154678232-154678232 on Build GRCh38
rs145503993 Chr.X: 154678246-154678246 on Build GRCh38
rs952103073 Chr.X: 154678158-154678158 on Build GRCh38
rs782169188 Chr.X: 154678163-154678163 on Build GRCh38
rs782589033 Chr.X: 154678315-154678315 on Build GRCh38
rs782544364 Chr.X: 154678252-154678252 on Build GRCh38
rs782605955 Chr.X: 154678144-154678144 on Build GRCh38
rs782545203 Chr.X: 154678310-154678310 on Build GRCh38
rs781900195 Chr.X: 154678250-154678250 on Build GRCh38
rs369433029 Chr.X: 154678170-154678170 on Build GRCh38
rs782110186 Chr.X: 154678171-154678171 on Build GRCh38
rs200068719 Chr.X: 154678199-154678199 on Build GRCh38
rs782227100 Chr.X: 154678276-154678276 on Build GRCh38
rs782792915 Chr.X: 154678168-154678168 on Build GRCh38

View Full Product Details