Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138221863-138222088 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_014154.4 NM_015396.5 NM_213654.3 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 XM_017006100.1 XM_017006101.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1038997 c.82G>T p.D28Y Chr.3: 138221927-138221927 on Build GRCh38
6605003 c.116A>C p.Q39P Chr.3: 138221961-138221961 on Build GRCh38
1038998 c.148C>T p.P50S Chr.3: 138221993-138221993 on Build GRCh38
1038996 c.82G>T p.D28Y Chr.3: 138221927-138221927 on Build GRCh38
6605004 c.116A>C p.Q39P Chr.3: 138221961-138221961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768548541 Chr.3: 138221914-138221914 on Build GRCh38
rs764550018 Chr.3: 138221903-138221904 on Build GRCh38
rs146409458 Chr.3: 138221982-138221982 on Build GRCh38
rs368427196 Chr.3: 138221896-138221896 on Build GRCh38
rs563254250 Chr.3: 138222039-138222039 on Build GRCh38
rs746700300 Chr.3: 138222044-138222044 on Build GRCh38
rs768014498 Chr.3: 138222029-138222029 on Build GRCh38
rs751848770 Chr.3: 138221972-138221972 on Build GRCh38
rs202180644 Chr.3: 138222011-138222011 on Build GRCh38
rs770508053 Chr.3: 138221970-138221970 on Build GRCh38
rs756692084 Chr.3: 138222035-138222035 on Build GRCh38
rs755132196 Chr.3: 138222015-138222015 on Build GRCh38
rs1045125757 Chr.3: 138222056-138222056 on Build GRCh38
rs768156677 Chr.3: 138222046-138222046 on Build GRCh38
rs773294683 Chr.3: 138221936-138221936 on Build GRCh38
rs746634038 Chr.3: 138221913-138221913 on Build GRCh38
rs543444800 Chr.3: 138222037-138222037 on Build GRCh38
rs977682024 Chr.3: 138221996-138221996 on Build GRCh38
rs948094774 Chr.3: 138222027-138222027 on Build GRCh38
rs779397638 Chr.3: 138222043-138222043 on Build GRCh38
rs761773944 Chr.3: 138221930-138221930 on Build GRCh38
rs757990488 Chr.3: 138222040-138222040 on Build GRCh38
rs955210509 Chr.3: 138221923-138221923 on Build GRCh38
rs771633462 Chr.3: 138221903-138221903 on Build GRCh38
rs753071280 Chr.3: 138222031-138222031 on Build GRCh38
rs745309026 Chr.3: 138221902-138221902 on Build GRCh38
rs772304358 Chr.3: 138221916-138221916 on Build GRCh38
rs763151430 Chr.3: 138221956-138221956 on Build GRCh38
rs371410195 Chr.3: 138221910-138221910 on Build GRCh38

View Full Product Details