Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37947004-37947261 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
UniGene
Hs.475963
Gene Chromosome Location
Chr.3: 37862152-37984469 on Build GRCh38
Transcripts
NM_001008392.1 NM_005808.2 XM_017005519.1 XM_017005520.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1044053 c.122G>T p.S41I Chr.3: 37947099-37947099 on Build GRCh38
4909488 c.177A>C p.P59P Chr.3: 37947154-37947154 on Build GRCh38
1044052 c.117C>T p.S39S Chr.3: 37947094-37947094 on Build GRCh38
5350974 c.234+1G>T p.? Chr.3: 37947212-37947212 on Build GRCh38
1044055 c.152G>A p.R51H Chr.3: 37947129-37947129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373415583 Chr.3: 37947054-37947054 on Build GRCh38
rs758006690 Chr.3: 37947198-37947198 on Build GRCh38
rs751176551 Chr.3: 37947212-37947212 on Build GRCh38
rs747525595 Chr.3: 37947129-37947129 on Build GRCh38
rs769160691 Chr.3: 37947031-37947031 on Build GRCh38
rs536573249 Chr.3: 37947096-37947096 on Build GRCh38
rs201081551 Chr.3: 37947181-37947181 on Build GRCh38
rs781774053 Chr.3: 37947236-37947236 on Build GRCh38
rs750356778 Chr.3: 37947237-37947237 on Build GRCh38
rs773334934 Chr.3: 37947161-37947161 on Build GRCh38
rs759607059 Chr.3: 37947177-37947177 on Build GRCh38
rs977570413 Chr.3: 37947061-37947061 on Build GRCh38
rs1803396 Chr.3: 37947124-37947124 on Build GRCh38
rs201170983 Chr.3: 37947074-37947074 on Build GRCh38
rs567415488 Chr.3: 37947071-37947071 on Build GRCh38
rs139111417 Chr.3: 37947175-37947175 on Build GRCh38
rs748570018 Chr.3: 37947139-37947139 on Build GRCh38
rs750061522 Chr.3: 37947188-37947188 on Build GRCh38
rs546609810 Chr.3: 37947145-37947145 on Build GRCh38
rs754512063 Chr.3: 37947234-37947234 on Build GRCh38
rs764569886 Chr.3: 37947112-37947114 on Build GRCh38
rs773458482 Chr.3: 37947057-37947057 on Build GRCh38
rs765034463 Chr.3: 37947187-37947187 on Build GRCh38
rs115062878 Chr.3: 37947201-37947201 on Build GRCh38
rs920949423 Chr.3: 37947194-37947194 on Build GRCh38
rs764069281 Chr.3: 37947178-37947178 on Build GRCh38
rs368806516 Chr.3: 37947038-37947038 on Build GRCh38
rs566856780 Chr.3: 37947163-37947163 on Build GRCh38
rs778471490 Chr.3: 37947117-37947117 on Build GRCh38
rs769357033 Chr.3: 37947135-37947135 on Build GRCh38
rs760547549 Chr.3: 37947086-37947086 on Build GRCh38
rs762267748 Chr.3: 37947033-37947033 on Build GRCh38
rs369543864 Chr.3: 37947182-37947182 on Build GRCh38
rs761930337 Chr.3: 37947164-37947164 on Build GRCh38
rs757873092 Chr.3: 37947126-37947126 on Build GRCh38
rs767617779 Chr.3: 37947095-37947095 on Build GRCh38
rs781760470 Chr.3: 37947138-37947138 on Build GRCh38
rs770930217 Chr.3: 37947168-37947168 on Build GRCh38
rs116703408 Chr.3: 37947070-37947070 on Build GRCh38
rs753698433 Chr.3: 37947094-37947094 on Build GRCh38
rs748621371 Chr.3: 37947238-37947238 on Build GRCh38
rs779563242 Chr.3: 37947128-37947128 on Build GRCh38
rs891013000 Chr.3: 37947162-37947162 on Build GRCh38
rs777071206 Chr.3: 37947032-37947032 on Build GRCh38
rs941395927 Chr.3: 37947030-37947030 on Build GRCh38
rs139535157 Chr.3: 37947064-37947064 on Build GRCh38

View Full Product Details