Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18236359-18236592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEK
Entrez Gene ID
7913
Gene Name
DEK proto-oncogene
Gene Aliases
D6S231E
UniGene
Hs.484813
Gene Chromosome Location
Chr.6: 18224169-18264568 on Build GRCh38
Transcripts
NM_001134709.1 NM_003472.3 XM_017011273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1076392 c.987A>G p.I329M Chr.6: 18236512-18236512 on Build GRCh38
2856288 c.950A>G p.K317R Chr.6: 18236549-18236549 on Build GRCh38
6470979 c.938T>A p.I313N Chr.6: 18236561-18236561 on Build GRCh38
3928246 c.926A>C p.D309A Chr.6: 18236573-18236573 on Build GRCh38
6470980 c.937A>T p.I313F Chr.6: 18236562-18236562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747706115 Chr.6: 18236466-18236466 on Build GRCh38
rs777578246 Chr.6: 18236524-18236524 on Build GRCh38
rs764323078 Chr.6: 18236456-18236456 on Build GRCh38
rs201860723 Chr.6: 18236429-18236429 on Build GRCh38
rs746658896 Chr.6: 18236544-18236544 on Build GRCh38
rs201058972 Chr.6: 18236515-18236515 on Build GRCh38
rs760566685 Chr.6: 18236422-18236422 on Build GRCh38
rs752794463 Chr.6: 18236442-18236442 on Build GRCh38
rs75250173 Chr.6: 18236423-18236423 on Build GRCh38
rs772955873 Chr.6: 18236416-18236416 on Build GRCh38
rs371327855 Chr.6: 18236396-18236396 on Build GRCh38
rs985995668 Chr.6: 18236491-18236491 on Build GRCh38
rs778323884 Chr.6: 18236512-18236512 on Build GRCh38
rs781531063 Chr.6: 18236483-18236483 on Build GRCh38
rs372362254 Chr.6: 18236405-18236405 on Build GRCh38
rs200180483 Chr.6: 18236549-18236549 on Build GRCh38
rs771816265 Chr.6: 18236407-18236407 on Build GRCh38
rs140999137 Chr.6: 18236472-18236472 on Build GRCh38
rs750949391 Chr.6: 18236506-18236506 on Build GRCh38
rs372069467 Chr.6: 18236428-18236428 on Build GRCh38
rs757621134 Chr.6: 18236476-18236476 on Build GRCh38
rs1015824443 Chr.6: 18236391-18236391 on Build GRCh38
rs965864533 Chr.6: 18236538-18236538 on Build GRCh38
rs747721590 Chr.6: 18236514-18236514 on Build GRCh38
rs750511005 Chr.6: 18236510-18236513 on Build GRCh38
rs756621020 Chr.6: 18236507-18236507 on Build GRCh38
rs879681955 Chr.6: 18236508-18236508 on Build GRCh38
rs778775189 Chr.6: 18236432-18236432 on Build GRCh38

View Full Product Details