Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mx1
Entrez Gene ID
17857
Gene Name
MX dynamin-like GTPase 1
Gene Aliases
AI893580, Mx, Mx-1
Chromosome Location
Chr.16: 97447035 - 97462906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010846.1 NP_034976.1 7 - 8 1038 93
GenBank mRNA AK076430.1 - 10 - 11 1214 93
BC011113.1 - 6 - 7 1023 93
CT010406.1 - - 825 93
M12279.1 - 7 - 8 1038 93
M21038.1 - 8 - 9 1110 93
M21039.1 - 7 - 8 1038 93

View Full Product Details