Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 89178381-89178894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYR
Entrez Gene ID
7299
Gene Name
tyrosinase
Gene Aliases
ATN, CMM8, OCA1, OCA1A, OCAIA, SHEP3
UniGene
Hs.503555
Gene Chromosome Location
Chr.11: 89177565-89295759 on Build GRCh38
Transcripts
NM_000372.4 XM_011542970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3454188 c.713G>A p.W238* Chr.11: 89178666-89178666 on Build GRCh38
6552472 c.428A>T p.H143L Chr.11: 89178381-89178381 on Build GRCh38
6014611 c.577G>A p.E193K Chr.11: 89178530-89178530 on Build GRCh38
340041 c.715C>T p.R239W Chr.11: 89178668-89178668 on Build GRCh38
7080351 c.675G>A p.L225L Chr.11: 89178628-89178628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs62645902 Chr.11: 89178769-89178769 on Build GRCh38
rs773366897 Chr.11: 89178672-89178672 on Build GRCh38
rs36006590 Chr.11: 89178669-89178669 on Build GRCh38
rs879988591 Chr.11: 89178683-89178684 on Build GRCh38
rs61754361 Chr.11: 89178521-89178521 on Build GRCh38
rs778827696 Chr.11: 89178604-89178604 on Build GRCh38
rs201920649 Chr.11: 89178707-89178707 on Build GRCh38
rs750553908 Chr.11: 89178546-89178546 on Build GRCh38
rs61754365 Chr.11: 89178603-89178603 on Build GRCh38
rs377209424 Chr.11: 89178588-89178588 on Build GRCh38
rs199500520 Chr.11: 89178713-89178713 on Build GRCh38
rs606231140 Chr.11: 89178685-89178686 on Build GRCh38
rs144018635 Chr.11: 89178621-89178621 on Build GRCh38
rs371734545 Chr.11: 89178406-89178406 on Build GRCh38
rs34878847 Chr.11: 89178618-89178618 on Build GRCh38
rs797046132 Chr.11: 89178533-89178533 on Build GRCh38
rs747995722 Chr.11: 89178608-89178608 on Build GRCh38
rs1029554094 Chr.11: 89178834-89178834 on Build GRCh38
rs774670098 Chr.11: 89178668-89178668 on Build GRCh38
rs61754369 Chr.11: 89178710-89178710 on Build GRCh38
rs112267653 Chr.11: 89178517-89178517 on Build GRCh38
rs748669377 Chr.11: 89178772-89178772 on Build GRCh38
rs531962127 Chr.11: 89178441-89178441 on Build GRCh38
rs200471520 Chr.11: 89178414-89178414 on Build GRCh38
rs538081629 Chr.11: 89178674-89178674 on Build GRCh38
rs751422747 Chr.11: 89178813-89178813 on Build GRCh38
rs147574809 Chr.11: 89178538-89178538 on Build GRCh38
rs770344763 Chr.11: 89178761-89178762 on Build GRCh38
rs61753257 Chr.11: 89178409-89178409 on Build GRCh38
rs1012308221 Chr.11: 89178649-89178649 on Build GRCh38
rs371810580 Chr.11: 89178653-89178653 on Build GRCh38
rs571810545 Chr.11: 89178483-89178483 on Build GRCh38
rs760149792 Chr.11: 89178446-89178446 on Build GRCh38
rs754661359 Chr.11: 89178559-89178559 on Build GRCh38
rs138817690 Chr.11: 89178658-89178658 on Build GRCh38
rs762249667 Chr.11: 89178687-89178688 on Build GRCh38
rs770276552 Chr.11: 89178633-89178633 on Build GRCh38
rs759359525 Chr.11: 89178684-89178684 on Build GRCh38
rs751621384 Chr.11: 89178478-89178478 on Build GRCh38
rs138487695 Chr.11: 89178482-89178482 on Build GRCh38
rs767644694 Chr.11: 89178635-89178635 on Build GRCh38
rs771898244 Chr.11: 89178568-89178568 on Build GRCh38
rs984031305 Chr.11: 89178437-89178437 on Build GRCh38
rs753099090 Chr.11: 89178522-89178522 on Build GRCh38
rs751717415 Chr.11: 89178531-89178531 on Build GRCh38
rs61754363 Chr.11: 89178599-89178599 on Build GRCh38
rs759716361 Chr.11: 89178798-89178798 on Build GRCh38
rs61753258 Chr.11: 89178411-89178412 on Build GRCh38
rs368210899 Chr.11: 89178762-89178762 on Build GRCh38
rs753998592 Chr.11: 89178682-89178682 on Build GRCh38
rs756049855 Chr.11: 89178549-89178549 on Build GRCh38
rs779999546 Chr.11: 89178680-89178681 on Build GRCh38
rs753663519 Chr.11: 89178508-89178508 on Build GRCh38
rs769148847 Chr.11: 89178765-89178765 on Build GRCh38
rs794726885 Chr.11: 89178458-89178460 on Build GRCh38
rs764330630 Chr.11: 89178585-89178585 on Build GRCh38
rs779454147 Chr.11: 89178768-89178768 on Build GRCh38
rs192709164 Chr.11: 89178870-89178870 on Build GRCh38
rs148813091 Chr.11: 89178457-89178457 on Build GRCh38
rs61754370 Chr.11: 89178719-89178719 on Build GRCh38
rs281865525 Chr.11: 89178525-89178525 on Build GRCh38
rs61754364 Chr.11: 89178602-89178602 on Build GRCh38
rs773765028 Chr.11: 89178783-89178791 on Build GRCh38
rs367543066 Chr.11: 89178504-89178504 on Build GRCh38
rs201078577 Chr.11: 89178496-89178496 on Build GRCh38
rs577370259 Chr.11: 89178779-89178779 on Build GRCh38
rs758612817 Chr.11: 89178473-89178473 on Build GRCh38
rs61754360 Chr.11: 89178486-89178486 on Build GRCh38
rs770317561 Chr.11: 89178580-89178580 on Build GRCh38
rs367543068 Chr.11: 89178692-89178692 on Build GRCh38
rs61759522 Chr.11: 89178740-89178740 on Build GRCh38
rs766319114 Chr.11: 89178679-89178679 on Build GRCh38
rs373550309 Chr.11: 89178555-89178555 on Build GRCh38
rs797046083 Chr.11: 89178611-89178611 on Build GRCh38
rs141930049 Chr.11: 89178500-89178500 on Build GRCh38
rs778881311 Chr.11: 89178560-89178560 on Build GRCh38
rs28940880 Chr.11: 89178569-89178569 on Build GRCh38
rs765329261 Chr.11: 89178706-89178706 on Build GRCh38
rs553415682 Chr.11: 89178802-89178802 on Build GRCh38
rs772869664 Chr.11: 89178514-89178514 on Build GRCh38
rs61754362 Chr.11: 89178566-89178566 on Build GRCh38
rs746581409 Chr.11: 89178579-89178579 on Build GRCh38
rs745847253 Chr.11: 89178646-89178646 on Build GRCh38
rs779878377 Chr.11: 89178491-89178491 on Build GRCh38
rs749096874 Chr.11: 89178557-89178557 on Build GRCh38
rs367719053 Chr.11: 89178792-89178792 on Build GRCh38
rs61754366 Chr.11: 89178631-89178633 on Build GRCh38
rs767768322 Chr.11: 89178598-89178598 on Build GRCh38
rs372123800 Chr.11: 89178526-89178526 on Build GRCh38
rs764002598 Chr.11: 89178722-89178722 on Build GRCh38
rs763935436 Chr.11: 89178809-89178809 on Build GRCh38
rs775182073 Chr.11: 89178418-89178418 on Build GRCh38
rs567675753 Chr.11: 89178511-89178511 on Build GRCh38
rs775799670 Chr.11: 89178583-89178583 on Build GRCh38
rs539948496 Chr.11: 89178800-89178800 on Build GRCh38
rs747095957 Chr.11: 89178405-89178405 on Build GRCh38
rs781160747 Chr.11: 89178411-89178411 on Build GRCh38
rs879992392 Chr.11: 89178685-89178686 on Build GRCh38
rs201700612 Chr.11: 89178425-89178425 on Build GRCh38
rs922459500 Chr.11: 89178839-89178839 on Build GRCh38
rs34297847 Chr.11: 89178749-89178749 on Build GRCh38
rs746685788 Chr.11: 89178631-89178631 on Build GRCh38
rs1042602 Chr.11: 89178528-89178528 on Build GRCh38
rs761482859 Chr.11: 89178436-89178436 on Build GRCh38
rs773646545 Chr.11: 89178776-89178776 on Build GRCh38
rs11826502 Chr.11: 89178732-89178732 on Build GRCh38
rs920624553 Chr.11: 89178417-89178417 on Build GRCh38
rs528817446 Chr.11: 89178412-89178412 on Build GRCh38
rs763013244 Chr.11: 89178805-89178805 on Build GRCh38
rs762002935 Chr.11: 89178593-89178593 on Build GRCh38
rs200936835 Chr.11: 89178712-89178712 on Build GRCh38
rs61754368 Chr.11: 89178683-89178684 on Build GRCh38
rs61754367 Chr.11: 89178660-89178660 on Build GRCh38
rs371040611 Chr.11: 89178780-89178780 on Build GRCh38
rs777492860 Chr.11: 89178481-89178484 on Build GRCh38
rs372458096 Chr.11: 89178423-89178423 on Build GRCh38
rs985142970 Chr.11: 89178872-89178872 on Build GRCh38
rs150207593 Chr.11: 89178643-89178643 on Build GRCh38
rs769645029 Chr.11: 89178489-89178489 on Build GRCh38
rs61753259 Chr.11: 89178479-89178479 on Build GRCh38
rs773881379 Chr.11: 89178427-89178427 on Build GRCh38
rs753049427 Chr.11: 89178458-89178458 on Build GRCh38
rs768350532 Chr.11: 89178498-89178498 on Build GRCh38
rs772358456 Chr.11: 89178773-89178773 on Build GRCh38
rs769204360 Chr.11: 89178661-89178661 on Build GRCh38
rs761771576 Chr.11: 89178793-89178793 on Build GRCh38
rs758115945 Chr.11: 89178614-89178614 on Build GRCh38
rs777920667 Chr.11: 89178475-89178475 on Build GRCh38
rs751135308 Chr.11: 89178736-89178736 on Build GRCh38
rs145513733 Chr.11: 89178407-89178407 on Build GRCh38
rs1040900593 Chr.11: 89178515-89178515 on Build GRCh38
rs547058090 Chr.11: 89178471-89178471 on Build GRCh38
rs772184078 Chr.11: 89178677-89178678 on Build GRCh38
rs767539316 Chr.11: 89178544-89178544 on Build GRCh38
rs63159160 Chr.11: 89178602-89178602 on Build GRCh38

View Full Product Details