Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 89227478-89227983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYR
Entrez Gene ID
7299
Gene Name
tyrosinase
Gene Aliases
ATN, CMM8, OCA1, OCA1A, OCAIA, SHEP3
UniGene
Hs.503555
Gene Chromosome Location
Chr.11: 89177565-89295759 on Build GRCh38
Transcripts
NM_000372.4 XM_011542970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4037344 c.1162C>A p.L388I Chr.11: 89227948-89227948 on Build GRCh38
3454190 c.1049C>T p.P350L Chr.11: 89227835-89227835 on Build GRCh38
121053 c.1068T>A p.D356E Chr.11: 89227854-89227854 on Build GRCh38
4037343 c.1086G>A p.M362I Chr.11: 89227872-89227872 on Build GRCh38
3375980 c.1166A>G p.H389R Chr.11: 89227952-89227952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770218017 Chr.11: 89227831-89227832 on Build GRCh38
rs779692959 Chr.11: 89227854-89227854 on Build GRCh38
rs755161413 Chr.11: 89227805-89227805 on Build GRCh38
rs754675944 Chr.11: 89227872-89227872 on Build GRCh38
rs61754388 Chr.11: 89227904-89227904 on Build GRCh38
rs779929859 Chr.11: 89227820-89227820 on Build GRCh38
rs886044493 Chr.11: 89227922-89227922 on Build GRCh38
rs1036750194 Chr.11: 89227740-89227740 on Build GRCh38
rs75375648 Chr.11: 89227648-89227648 on Build GRCh38
rs750566431 Chr.11: 89227929-89227929 on Build GRCh38
rs527352749 Chr.11: 89227804-89227804 on Build GRCh38
rs922867463 Chr.11: 89227549-89227549 on Build GRCh38
rs61754382 Chr.11: 89227822-89227822 on Build GRCh38
rs139091458 Chr.11: 89227887-89227887 on Build GRCh38
rs896863972 Chr.11: 89227516-89227516 on Build GRCh38
rs773970123 Chr.11: 89227823-89227823 on Build GRCh38
rs776790750 Chr.11: 89227691-89227691 on Build GRCh38
rs763127567 Chr.11: 89227847-89227847 on Build GRCh38
rs61754381 Chr.11: 89227816-89227816 on Build GRCh38
rs149912348 Chr.11: 89227892-89227892 on Build GRCh38
rs986067423 Chr.11: 89227577-89227577 on Build GRCh38
rs771720319 Chr.11: 89227830-89227830 on Build GRCh38
rs997774435 Chr.11: 89227518-89227518 on Build GRCh38
rs781554851 Chr.11: 89227805-89227806 on Build GRCh38
rs781052288 Chr.11: 89227805-89227805 on Build GRCh38
rs748901196 Chr.11: 89227821-89227821 on Build GRCh38
rs192156062 Chr.11: 89227579-89227579 on Build GRCh38
rs1010038586 Chr.11: 89227832-89227832 on Build GRCh38
rs946763563 Chr.11: 89227582-89227582 on Build GRCh38
rs765467173 Chr.11: 89227842-89227842 on Build GRCh38
rs568415107 Chr.11: 89227661-89227661 on Build GRCh38
rs61754385 Chr.11: 89227895-89227895 on Build GRCh38
rs747032303 Chr.11: 89227804-89227805 on Build GRCh38
rs927586533 Chr.11: 89227643-89227643 on Build GRCh38
rs971447227 Chr.11: 89227848-89227848 on Build GRCh38
rs1800421 Chr.11: 89227851-89227851 on Build GRCh38
rs932832442 Chr.11: 89227562-89227562 on Build GRCh38
rs770370708 Chr.11: 89227899-89227899 on Build GRCh38
rs61754386 Chr.11: 89227897-89227897 on Build GRCh38
rs61754390 Chr.11: 89227918-89227918 on Build GRCh38
rs13312743 Chr.11: 89227953-89227953 on Build GRCh38
rs767446956 Chr.11: 89227927-89227927 on Build GRCh38
rs184901887 Chr.11: 89227671-89227671 on Build GRCh38
rs576929827 Chr.11: 89227721-89227721 on Build GRCh38
rs281865521 Chr.11: 89227947-89227947 on Build GRCh38
rs1038366981 Chr.11: 89227870-89227871 on Build GRCh38
rs1057031234 Chr.11: 89227624-89227624 on Build GRCh38
rs1005274377 Chr.11: 89227730-89227730 on Build GRCh38
rs61754391 Chr.11: 89227924-89227924 on Build GRCh38
rs775711602 Chr.11: 89227901-89227901 on Build GRCh38
rs758278271 Chr.11: 89227795-89227795 on Build GRCh38
rs180801021 Chr.11: 89227853-89227853 on Build GRCh38
rs12804012 Chr.11: 89227622-89227622 on Build GRCh38
rs775968923 Chr.11: 89227845-89227845 on Build GRCh38
rs776935572 Chr.11: 89227838-89227838 on Build GRCh38
rs961043889 Chr.11: 89227521-89227521 on Build GRCh38
rs61754387 Chr.11: 89227898-89227898 on Build GRCh38
rs1015380269 Chr.11: 89227731-89227731 on Build GRCh38
rs535724702 Chr.11: 89227642-89227642 on Build GRCh38
rs1020373770 Chr.11: 89227583-89227583 on Build GRCh38
rs61754384 Chr.11: 89227886-89227886 on Build GRCh38
rs556548056 Chr.11: 89227615-89227615 on Build GRCh38
rs748447644 Chr.11: 89227813-89227814 on Build GRCh38
rs751239011 Chr.11: 89227743-89227743 on Build GRCh38
rs757185332 Chr.11: 89227803-89227803 on Build GRCh38
rs151206295 Chr.11: 89227850-89227850 on Build GRCh38
rs3834913 Chr.11: 89227795-89227796 on Build GRCh38
rs749137245 Chr.11: 89227855-89227855 on Build GRCh38
rs762057851 Chr.11: 89227926-89227926 on Build GRCh38
rs981400831 Chr.11: 89227639-89227639 on Build GRCh38
rs377467376 Chr.11: 89227563-89227563 on Build GRCh38
rs61754383 Chr.11: 89227867-89227867 on Build GRCh38
rs939365679 Chr.11: 89227917-89227917 on Build GRCh38
rs750304137 Chr.11: 89227793-89227794 on Build GRCh38
rs141967840 Chr.11: 89227947-89227947 on Build GRCh38
rs113438665 Chr.11: 89227746-89227746 on Build GRCh38
rs1042470519 Chr.11: 89227593-89227593 on Build GRCh38
rs61754389 Chr.11: 89227912-89227912 on Build GRCh38
rs104894315 Chr.11: 89227932-89227932 on Build GRCh38
rs62645914 Chr.11: 89227954-89227954 on Build GRCh38
rs769024531 Chr.11: 89227907-89227907 on Build GRCh38
rs1006740678 Chr.11: 89227556-89227556 on Build GRCh38
rs985732783 Chr.11: 89227833-89227833 on Build GRCh38
rs776054795 Chr.11: 89227885-89227885 on Build GRCh38
rs62645908 Chr.11: 89227849-89227849 on Build GRCh38
rs556997125 Chr.11: 89227690-89227690 on Build GRCh38
rs62645909 Chr.11: 89227861-89227861 on Build GRCh38
rs57056852 Chr.11: 89227548-89227548 on Build GRCh38
rs281865522 Chr.11: 89227950-89227950 on Build GRCh38
rs121908011 Chr.11: 89227933-89227933 on Build GRCh38

View Full Product Details