Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124383044-124383585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR8B2
Entrez Gene ID
26595
Gene Name
olfactory receptor family 8 subfamily B member 2
Gene Aliases
OR11-309, OR11-310
UniGene
Hs.576944
Gene Chromosome Location
Chr.11: 124382402-124397319 on Build GRCh38
Transcripts
NM_001005468.1 XM_017017535.1 XM_017017536.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
347792 c.160C>T p.H54Y Chr.11: 124383184-124383184 on Build GRCh38
118461 c.111C>A p.V37V Chr.11: 124383233-124383233 on Build GRCh38
3731103 c.58C>T p.H20Y Chr.11: 124383286-124383286 on Build GRCh38
3738917 c.151C>A p.L51I Chr.11: 124383193-124383193 on Build GRCh38
6215906 c.115A>G p.M39V Chr.11: 124383229-124383229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764254911 Chr.11: 124383206-124383206 on Build GRCh38
rs753383960 Chr.11: 124383392-124383392 on Build GRCh38
rs71491825 Chr.11: 124383086-124383086 on Build GRCh38
rs551581078 Chr.11: 124383317-124383317 on Build GRCh38
rs765617279 Chr.11: 124383223-124383223 on Build GRCh38
rs199876895 Chr.11: 124383196-124383196 on Build GRCh38
rs757643074 Chr.11: 124383092-124383092 on Build GRCh38
rs771319646 Chr.11: 124383345-124383345 on Build GRCh38
rs564384576 Chr.11: 124383262-124383262 on Build GRCh38
rs779246524 Chr.11: 124383093-124383093 on Build GRCh38
rs754119236 Chr.11: 124383153-124383153 on Build GRCh38
rs748175456 Chr.11: 124383329-124383329 on Build GRCh38
rs780733725 Chr.11: 124383231-124383231 on Build GRCh38
rs760128633 Chr.11: 124383375-124383375 on Build GRCh38
rs551718145 Chr.11: 124383482-124383482 on Build GRCh38
rs530765 Chr.11: 124383274-124383274 on Build GRCh38
rs61911135 Chr.11: 124383244-124383244 on Build GRCh38
rs773869538 Chr.11: 124383325-124383326 on Build GRCh38
rs645986 Chr.11: 124383437-124383437 on Build GRCh38
rs754399313 Chr.11: 124383230-124383230 on Build GRCh38
rs760822583 Chr.11: 124383151-124383151 on Build GRCh38
rs777362418 Chr.11: 124383287-124383287 on Build GRCh38
rs778665416 Chr.11: 124383246-124383246 on Build GRCh38
rs757512857 Chr.11: 124383160-124383160 on Build GRCh38
rs779866636 Chr.11: 124383323-124383323 on Build GRCh38
rs769133409 Chr.11: 124383134-124383134 on Build GRCh38
rs904560050 Chr.11: 124383169-124383169 on Build GRCh38
rs779675843 Chr.11: 124383119-124383119 on Build GRCh38
rs754523383 Chr.11: 124383172-124383172 on Build GRCh38
rs946163306 Chr.11: 124383148-124383148 on Build GRCh38
rs745712551 Chr.11: 124383309-124383309 on Build GRCh38
rs202110730 Chr.11: 124383227-124383227 on Build GRCh38
rs762854770 Chr.11: 124383149-124383149 on Build GRCh38
rs778114101 Chr.11: 124383075-124383075 on Build GRCh38
rs780767848 Chr.11: 124383168-124383168 on Build GRCh38
rs746450138 Chr.11: 124383171-124383171 on Build GRCh38
rs73025504 Chr.11: 124383067-124383067 on Build GRCh38
rs773522741 Chr.11: 124383341-124383341 on Build GRCh38
rs775533084 Chr.11: 124383150-124383150 on Build GRCh38
rs142528588 Chr.11: 124383312-124383312 on Build GRCh38
rs373992300 Chr.11: 124383126-124383126 on Build GRCh38
rs777415859 Chr.11: 124383237-124383237 on Build GRCh38
rs569901512 Chr.11: 124383362-124383362 on Build GRCh38
rs746587520 Chr.11: 124383120-124383120 on Build GRCh38
rs71491824 Chr.11: 124383064-124383064 on Build GRCh38
rs150895565 Chr.11: 124383228-124383228 on Build GRCh38
rs776620465 Chr.11: 124383212-124383212 on Build GRCh38
rs547486532 Chr.11: 124383284-124383284 on Build GRCh38
rs748742226 Chr.11: 124383237-124383238 on Build GRCh38
rs763245724 Chr.11: 124383343-124383343 on Build GRCh38
rs772959850 Chr.11: 124383195-124383195 on Build GRCh38
rs530740 Chr.11: 124383265-124383265 on Build GRCh38
rs546367511 Chr.11: 124383249-124383249 on Build GRCh38
rs772030593 Chr.11: 124383313-124383313 on Build GRCh38
rs368014167 Chr.11: 124383140-124383140 on Build GRCh38
rs759876662 Chr.11: 124383361-124383361 on Build GRCh38
rs530704 Chr.11: 124383251-124383251 on Build GRCh38
rs558027045 Chr.11: 124383216-124383216 on Build GRCh38
rs576204895 Chr.11: 124383218-124383218 on Build GRCh38
rs796898996 Chr.11: 124383265-124383266 on Build GRCh38
rs774776498 Chr.11: 124383354-124383354 on Build GRCh38
rs755624011 Chr.11: 124383286-124383286 on Build GRCh38
rs751919102 Chr.11: 124383281-124383281 on Build GRCh38
rs1055074590 Chr.11: 124383485-124383485 on Build GRCh38
rs535367679 Chr.11: 124383107-124383107 on Build GRCh38
rs765672192 Chr.11: 124383161-124383161 on Build GRCh38
rs561859017 Chr.11: 124383267-124383267 on Build GRCh38
rs770155935 Chr.11: 124383313-124383314 on Build GRCh38
rs753174133 Chr.11: 124383365-124383365 on Build GRCh38
rs537258871 Chr.11: 124383463-124383463 on Build GRCh38
rs539582158 Chr.11: 124383177-124383177 on Build GRCh38
rs747341344 Chr.11: 124383130-124383131 on Build GRCh38
rs142056460 Chr.11: 124383561-124383561 on Build GRCh38
rs181240879 Chr.11: 124383225-124383225 on Build GRCh38
rs142893914 Chr.11: 124383146-124383146 on Build GRCh38
rs772752968 Chr.11: 124383115-124383115 on Build GRCh38
rs565957437 Chr.11: 124383291-124383291 on Build GRCh38
rs769383192 Chr.11: 124383194-124383194 on Build GRCh38
rs61911136 Chr.11: 124383457-124383457 on Build GRCh38
rs780672088 Chr.11: 124383176-124383176 on Build GRCh38
rs201727613 Chr.11: 124383164-124383164 on Build GRCh38
rs770796971 Chr.11: 124383197-124383197 on Build GRCh38
rs543542412 Chr.11: 124383266-124383266 on Build GRCh38
rs146094090 Chr.11: 124383152-124383152 on Build GRCh38
rs537171546 Chr.11: 124383221-124383221 on Build GRCh38
rs555473980 Chr.11: 124383481-124383481 on Build GRCh38
rs780366967 Chr.11: 124383078-124383078 on Build GRCh38
rs534569901 Chr.11: 124383502-124383502 on Build GRCh38
rs776766294 Chr.11: 124383256-124383256 on Build GRCh38
rs758985992 Chr.11: 124383343-124383343 on Build GRCh38
rs572657870 Chr.11: 124383245-124383245 on Build GRCh38
rs778735187 Chr.11: 124383295-124383295 on Build GRCh38
rs776850052 Chr.11: 124383136-124383136 on Build GRCh38
rs773086403 Chr.11: 124383147-124383147 on Build GRCh38
rs753510837 Chr.11: 124383288-124383288 on Build GRCh38
rs540691702 Chr.11: 124383235-124383235 on Build GRCh38
rs1001921269 Chr.11: 124383213-124383213 on Build GRCh38
rs71491826 Chr.11: 124383204-124383204 on Build GRCh38
rs10750267 Chr.11: 124383477-124383477 on Build GRCh38
rs770029129 Chr.11: 124383338-124383338 on Build GRCh38
rs763572804 Chr.11: 124383384-124383384 on Build GRCh38
rs776362719 Chr.11: 124383138-124383138 on Build GRCh38
rs1013485127 Chr.11: 124383518-124383518 on Build GRCh38
rs529243218 Chr.11: 124383273-124383273 on Build GRCh38
rs28373946 Chr.11: 124383285-124383285 on Build GRCh38

View Full Product Details