Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4760767-4761270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT8
Entrez Gene ID
26290
Gene Name
polypeptide N-acetylgalactosaminyltransferase 8
Gene Aliases
GALNAC-T8
UniGene
Hs.511985
Gene Chromosome Location
Chr.12: 4720586-4772726 on Build GRCh38
Transcripts
NM_017417.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873873 c.1245G>T p.K415N Chr.12: 4761029-4761029 on Build GRCh38
3812119 c.1267G>A p.A423T Chr.12: 4761051-4761051 on Build GRCh38
459489 c.1197C>T p.V399V Chr.12: 4760981-4760981 on Build GRCh38
1606180 c.1335C>T p.Y445Y Chr.12: 4761119-4761119 on Build GRCh38
1606179 c.1230A>G p.L410L Chr.12: 4761014-4761014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973225701 Chr.12: 4760896-4760896 on Build GRCh38
rs2191157 Chr.12: 4761197-4761197 on Build GRCh38
rs778628161 Chr.12: 4761185-4761185 on Build GRCh38
rs369241514 Chr.12: 4760987-4760987 on Build GRCh38
rs531035776 Chr.12: 4760812-4760812 on Build GRCh38
rs143807268 Chr.12: 4761067-4761067 on Build GRCh38
rs1029461791 Chr.12: 4761093-4761093 on Build GRCh38
rs922441609 Chr.12: 4760920-4760920 on Build GRCh38
rs750358554 Chr.12: 4761000-4761000 on Build GRCh38
rs770609421 Chr.12: 4761110-4761110 on Build GRCh38
rs1001920643 Chr.12: 4760830-4760830 on Build GRCh38
rs750919954 Chr.12: 4760998-4760998 on Build GRCh38
rs998110366 Chr.12: 4761092-4761092 on Build GRCh38
rs373711782 Chr.12: 4761001-4761001 on Build GRCh38
rs559663039 Chr.12: 4761097-4761097 on Build GRCh38
rs771590048 Chr.12: 4761118-4761118 on Build GRCh38
rs748797579 Chr.12: 4761094-4761095 on Build GRCh38
rs1051892873 Chr.12: 4760909-4760909 on Build GRCh38
rs781614065 Chr.12: 4761026-4761026 on Build GRCh38
rs779294360 Chr.12: 4760993-4760993 on Build GRCh38
rs938169577 Chr.12: 4761222-4761222 on Build GRCh38
rs765535951 Chr.12: 4761064-4761064 on Build GRCh38
rs756291725 Chr.12: 4761103-4761103 on Build GRCh38
rs766971985 Chr.12: 4761076-4761076 on Build GRCh38
rs180810709 Chr.12: 4760931-4760931 on Build GRCh38
rs773064917 Chr.12: 4760944-4760944 on Build GRCh38
rs1024292090 Chr.12: 4761042-4761042 on Build GRCh38
rs140072173 Chr.12: 4761050-4761050 on Build GRCh38
rs551073173 Chr.12: 4761213-4761213 on Build GRCh38
rs749166176 Chr.12: 4761108-4761108 on Build GRCh38
rs558918630 Chr.12: 4760919-4760919 on Build GRCh38
rs766622102 Chr.12: 4761135-4761135 on Build GRCh38
rs1046568934 Chr.12: 4760797-4760797 on Build GRCh38
rs987230661 Chr.12: 4761079-4761079 on Build GRCh38
rs554644531 Chr.12: 4760948-4760948 on Build GRCh38
rs140915911 Chr.12: 4761132-4761132 on Build GRCh38
rs150653153 Chr.12: 4760906-4760906 on Build GRCh38
rs570837147 Chr.12: 4761242-4761242 on Build GRCh38
rs748781263 Chr.12: 4760887-4760887 on Build GRCh38
rs752145439 Chr.12: 4761136-4761136 on Build GRCh38
rs377567116 Chr.12: 4761045-4761045 on Build GRCh38
rs751013055 Chr.12: 4760970-4760970 on Build GRCh38
rs373419543 Chr.12: 4761036-4761036 on Build GRCh38
rs200902818 Chr.12: 4761075-4761075 on Build GRCh38
rs781382912 Chr.12: 4761084-4761084 on Build GRCh38
rs745816218 Chr.12: 4761190-4761190 on Build GRCh38
rs766979231 Chr.12: 4761056-4761056 on Build GRCh38
rs751255900 Chr.12: 4761170-4761170 on Build GRCh38
rs1033563546 Chr.12: 4761054-4761054 on Build GRCh38
rs142857763 Chr.12: 4761063-4761063 on Build GRCh38
rs982327543 Chr.12: 4761197-4761197 on Build GRCh38
rs891955042 Chr.12: 4761163-4761163 on Build GRCh38
rs755494737 Chr.12: 4761140-4761140 on Build GRCh38
rs372359115 Chr.12: 4760915-4760915 on Build GRCh38
rs759042813 Chr.12: 4760977-4760977 on Build GRCh38
rs919016840 Chr.12: 4760904-4760904 on Build GRCh38
rs141832600 Chr.12: 4760871-4760871 on Build GRCh38
rs776991597 Chr.12: 4760922-4760922 on Build GRCh38
rs145805938 Chr.12: 4761009-4761009 on Build GRCh38
rs774308714 Chr.12: 4761111-4761111 on Build GRCh38
rs766726075 Chr.12: 4761051-4761051 on Build GRCh38
rs755703307 Chr.12: 4760936-4760937 on Build GRCh38
rs780137035 Chr.12: 4760908-4760908 on Build GRCh38
rs73264336 Chr.12: 4761035-4761035 on Build GRCh38
rs760326905 Chr.12: 4761124-4761124 on Build GRCh38
rs574197753 Chr.12: 4760888-4760888 on Build GRCh38
rs770537818 Chr.12: 4761135-4761135 on Build GRCh38
rs777376083 Chr.12: 4760993-4760994 on Build GRCh38
rs186503466 Chr.12: 4760982-4760982 on Build GRCh38
rs1039301407 Chr.12: 4760968-4760968 on Build GRCh38
rs61758971 Chr.12: 4761141-4761141 on Build GRCh38
rs770967813 Chr.12: 4761049-4761049 on Build GRCh38
rs773653105 Chr.12: 4761023-4761023 on Build GRCh38
rs562315540 Chr.12: 4761196-4761196 on Build GRCh38
rs963237641 Chr.12: 4761066-4761066 on Build GRCh38
rs767214354 Chr.12: 4760981-4760981 on Build GRCh38
rs913914619 Chr.12: 4761177-4761177 on Build GRCh38
rs201474343 Chr.12: 4760947-4760947 on Build GRCh38
rs763424884 Chr.12: 4761134-4761134 on Build GRCh38
rs943020133 Chr.12: 4761113-4761113 on Build GRCh38
rs201577168 Chr.12: 4761171-4761171 on Build GRCh38
rs761758945 Chr.12: 4760923-4760923 on Build GRCh38
rs189363417 Chr.12: 4760825-4760825 on Build GRCh38
rs778104085 Chr.12: 4761160-4761160 on Build GRCh38
rs1033311572 Chr.12: 4760852-4760852 on Build GRCh38
rs936658038 Chr.12: 4760955-4760955 on Build GRCh38
rs766264335 Chr.12: 4761083-4761083 on Build GRCh38
rs774927231 Chr.12: 4761122-4761122 on Build GRCh38
rs745763113 Chr.12: 4761116-4761116 on Build GRCh38
rs777851092 Chr.12: 4761104-4761104 on Build GRCh38
rs756134721 Chr.12: 4761157-4761157 on Build GRCh38
rs768844170 Chr.12: 4761021-4761021 on Build GRCh38
rs763877686 Chr.12: 4761130-4761130 on Build GRCh38
rs752889603 Chr.12: 4761146-4761146 on Build GRCh38
rs774838521 Chr.12: 4760794-4760794 on Build GRCh38
rs778612793 Chr.12: 4761183-4761185 on Build GRCh38
rs961307102 Chr.12: 4760890-4760890 on Build GRCh38

View Full Product Details