Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6637977-6638483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACRBP
Entrez Gene ID
84519
Gene Name
acrosin binding protein
Gene Aliases
CT23, OY-TES-1, SP32
UniGene
Hs.123239
Gene Chromosome Location
Chr.12: 6638076-6647414 on Build GRCh38
Transcripts
NM_032489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5375825 c.1569T>C p.S523S Chr.12: 6638345-6638345 on Build GRCh38
5492908 c.1628G>A p.G543E Chr.12: 6638286-6638286 on Build GRCh38
3688411 c.1592G>T p.S531I Chr.12: 6638322-6638322 on Build GRCh38
1363790 c.1626C>T p.F542F Chr.12: 6638288-6638288 on Build GRCh38
942779 c.1585C>T p.R529* Chr.12: 6638329-6638329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575755465 Chr.12: 6638116-6638116 on Build GRCh38
rs756765669 Chr.12: 6638338-6638338 on Build GRCh38
rs543641819 Chr.12: 6638339-6638339 on Build GRCh38
rs775333041 Chr.12: 6638305-6638305 on Build GRCh38
rs1008989760 Chr.12: 6638232-6638232 on Build GRCh38
rs763808261 Chr.12: 6638418-6638418 on Build GRCh38
rs988007817 Chr.12: 6638093-6638093 on Build GRCh38
rs563442506 Chr.12: 6638363-6638363 on Build GRCh38
rs545210881 Chr.12: 6638181-6638181 on Build GRCh38
rs1009083149 Chr.12: 6637999-6637999 on Build GRCh38
rs756470203 Chr.12: 6638358-6638358 on Build GRCh38
rs138674706 Chr.12: 6638085-6638085 on Build GRCh38
rs1020828593 Chr.12: 6638308-6638308 on Build GRCh38
rs545499100 Chr.12: 6638455-6638455 on Build GRCh38
rs534254213 Chr.12: 6638010-6638010 on Build GRCh38
rs78896594 Chr.12: 6638052-6638052 on Build GRCh38
rs368696301 Chr.12: 6638287-6638287 on Build GRCh38
rs377230865 Chr.12: 6638397-6638397 on Build GRCh38
rs755759374 Chr.12: 6638255-6638255 on Build GRCh38
rs946979972 Chr.12: 6638167-6638167 on Build GRCh38
rs756855333 Chr.12: 6638275-6638275 on Build GRCh38
rs1882553 Chr.12: 6638412-6638412 on Build GRCh38
rs372540538 Chr.12: 6638109-6638109 on Build GRCh38
rs751530631 Chr.12: 6638233-6638233 on Build GRCh38
rs571984630 Chr.12: 6638177-6638177 on Build GRCh38
rs1017874129 Chr.12: 6638234-6638234 on Build GRCh38
rs938046181 Chr.12: 6638131-6638131 on Build GRCh38
rs869309803 Chr.12: 6638054-6638054 on Build GRCh38
rs748504394 Chr.12: 6638278-6638278 on Build GRCh38
rs899258065 Chr.12: 6638176-6638176 on Build GRCh38
rs781602941 Chr.12: 6638356-6638356 on Build GRCh38
rs1045553 Chr.12: 6638129-6638129 on Build GRCh38
rs1047813159 Chr.12: 6638438-6638438 on Build GRCh38
rs900778468 Chr.12: 6638136-6638136 on Build GRCh38
rs752499660 Chr.12: 6638249-6638249 on Build GRCh38
rs5796246 Chr.12: 6638052-6638052 on Build GRCh38
rs755513976 Chr.12: 6638355-6638355 on Build GRCh38
rs202127738 Chr.12: 6638312-6638312 on Build GRCh38
rs371627956 Chr.12: 6638285-6638285 on Build GRCh38
rs890608124 Chr.12: 6638204-6638204 on Build GRCh38
rs929388448 Chr.12: 6638180-6638180 on Build GRCh38
rs556293213 Chr.12: 6638086-6638086 on Build GRCh38
rs752305322 Chr.12: 6638166-6638166 on Build GRCh38
rs777989155 Chr.12: 6638361-6638361 on Build GRCh38
rs2365031 Chr.12: 6638053-6638053 on Build GRCh38
rs555669427 Chr.12: 6638369-6638369 on Build GRCh38
rs745655155 Chr.12: 6638071-6638071 on Build GRCh38
rs386759946 Chr.12: 6638052-6638053 on Build GRCh38
rs771203599 Chr.12: 6638366-6638366 on Build GRCh38
rs538314213 Chr.12: 6638117-6638117 on Build GRCh38
rs371900051 Chr.12: 6638240-6638240 on Build GRCh38
rs944878239 Chr.12: 6638457-6638457 on Build GRCh38
rs375909099 Chr.12: 6638443-6638443 on Build GRCh38
rs774380289 Chr.12: 6638368-6638368 on Build GRCh38
rs1019265841 Chr.12: 6638006-6638006 on Build GRCh38
rs760451740 Chr.12: 6638414-6638414 on Build GRCh38
rs748496890 Chr.12: 6638357-6638357 on Build GRCh38
rs749487726 Chr.12: 6638375-6638377 on Build GRCh38
rs767314803 Chr.12: 6638246-6638246 on Build GRCh38
rs929419314 Chr.12: 6638437-6638437 on Build GRCh38
rs983225540 Chr.12: 6638398-6638398 on Build GRCh38
rs138951455 Chr.12: 6638303-6638303 on Build GRCh38
rs750900669 Chr.12: 6638034-6638034 on Build GRCh38
rs982012794 Chr.12: 6638128-6638128 on Build GRCh38
rs963759221 Chr.12: 6638019-6638019 on Build GRCh38
rs67911191 Chr.12: 6638053-6638053 on Build GRCh38
rs374375773 Chr.12: 6638259-6638259 on Build GRCh38
rs750023336 Chr.12: 6638345-6638345 on Build GRCh38
rs765024556 Chr.12: 6638427-6638427 on Build GRCh38
rs761554770 Chr.12: 6638426-6638426 on Build GRCh38
rs749931586 Chr.12: 6638428-6638428 on Build GRCh38
rs755724643 Chr.12: 6638276-6638276 on Build GRCh38
rs367935466 Chr.12: 6638266-6638266 on Build GRCh38
rs142857326 Chr.12: 6638329-6638329 on Build GRCh38
rs149312090 Chr.12: 6638288-6638288 on Build GRCh38
rs771993653 Chr.12: 6638409-6638409 on Build GRCh38
rs755307123 Chr.12: 6638051-6638052 on Build GRCh38
rs926565801 Chr.12: 6638393-6638393 on Build GRCh38
rs759463912 Chr.12: 6638298-6638298 on Build GRCh38
rs565182439 Chr.12: 6638458-6638458 on Build GRCh38
rs867512372 Chr.12: 6638142-6638142 on Build GRCh38
rs759110707 Chr.12: 6638038-6638038 on Build GRCh38
rs779032071 Chr.12: 6638424-6638424 on Build GRCh38
rs757869254 Chr.12: 6638447-6638447 on Build GRCh38
rs763758076 Chr.12: 6638328-6638328 on Build GRCh38
rs770039654 Chr.12: 6638280-6638280 on Build GRCh38

View Full Product Details