Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6955292-6955795 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN6
Entrez Gene ID
5777
Gene Name
protein tyrosine phosphatase, non-receptor type 6
Gene Aliases
HCP, HCPH, HPTP1C, PTP-1C, SH-PTP1, SHP-1, SHP-1L, SHP1
UniGene
Hs.63489
Gene Chromosome Location
Chr.12: 6946576-6961316 on Build GRCh38
Transcripts
NM_002831.5 NM_080548.4 NM_080549.3 XM_006718994.1 XM_011520988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3355862 c.727G>A p.G243S Chr.12: 6955465-6955465 on Build GRCh38
3792936 c.735C>T p.G245G Chr.12: 6955467-6955467 on Build GRCh38
6073317 c.829C>T p.R277C Chr.12: 6955735-6955735 on Build GRCh38
4926903 c.764A>G p.K255R Chr.12: 6955670-6955670 on Build GRCh38
5165549 c.642G>A p.P214P Chr.12: 6955374-6955374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936226647 Chr.12: 6955330-6955330 on Build GRCh38
rs782305808 Chr.12: 6955618-6955618 on Build GRCh38
rs781935785 Chr.12: 6955313-6955313 on Build GRCh38
rs782576939 Chr.12: 6955506-6955506 on Build GRCh38
rs781816472 Chr.12: 6955476-6955476 on Build GRCh38
rs782774511 Chr.12: 6955501-6955501 on Build GRCh38
rs781935337 Chr.12: 6955415-6955415 on Build GRCh38
rs187349009 Chr.12: 6955551-6955551 on Build GRCh38
rs782631186 Chr.12: 6955522-6955522 on Build GRCh38
rs782605040 Chr.12: 6955374-6955374 on Build GRCh38
rs781886724 Chr.12: 6955596-6955596 on Build GRCh38
rs782075547 Chr.12: 6955704-6955704 on Build GRCh38
rs782130684 Chr.12: 6955447-6955447 on Build GRCh38
rs782045447 Chr.12: 6955457-6955457 on Build GRCh38
rs913445901 Chr.12: 6955560-6955560 on Build GRCh38
rs782244694 Chr.12: 6955529-6955529 on Build GRCh38
rs782673400 Chr.12: 6955392-6955392 on Build GRCh38
rs371829165 Chr.12: 6955355-6955355 on Build GRCh38
rs782524619 Chr.12: 6955390-6955390 on Build GRCh38
rs199730259 Chr.12: 6955446-6955446 on Build GRCh38
rs140592255 Chr.12: 6955386-6955386 on Build GRCh38
rs782637799 Chr.12: 6955717-6955717 on Build GRCh38
rs782105964 Chr.12: 6955315-6955315 on Build GRCh38
rs990847370 Chr.12: 6955557-6955557 on Build GRCh38
rs782356549 Chr.12: 6955445-6955445 on Build GRCh38
rs782652143 Chr.12: 6955534-6955534 on Build GRCh38
rs782302331 Chr.12: 6955406-6955406 on Build GRCh38
rs782535430 Chr.12: 6955348-6955348 on Build GRCh38
rs782035997 Chr.12: 6955455-6955455 on Build GRCh38
rs374959527 Chr.12: 6955695-6955695 on Build GRCh38
rs1027567228 Chr.12: 6955490-6955490 on Build GRCh38
rs781954634 Chr.12: 6955311-6955311 on Build GRCh38
rs112278988 Chr.12: 6955653-6955653 on Build GRCh38
rs781912378 Chr.12: 6955545-6955545 on Build GRCh38
rs782737291 Chr.12: 6955707-6955707 on Build GRCh38
rs201345298 Chr.12: 6955413-6955413 on Build GRCh38
rs782291295 Chr.12: 6955609-6955609 on Build GRCh38
rs781784030 Chr.12: 6955360-6955360 on Build GRCh38
rs1045849676 Chr.12: 6955594-6955594 on Build GRCh38
rs371920672 Chr.12: 6955488-6955488 on Build GRCh38
rs1054694626 Chr.12: 6955332-6955332 on Build GRCh38
rs782721984 Chr.12: 6955467-6955467 on Build GRCh38
rs115220993 Chr.12: 6955518-6955518 on Build GRCh38
rs1054790754 Chr.12: 6955603-6955603 on Build GRCh38
rs781869681 Chr.12: 6955324-6955324 on Build GRCh38
rs966087881 Chr.12: 6955527-6955527 on Build GRCh38
rs782212727 Chr.12: 6955429-6955429 on Build GRCh38
rs782721100 Chr.12: 6955553-6955553 on Build GRCh38
rs782801811 Chr.12: 6955533-6955533 on Build GRCh38
rs782758640 Chr.12: 6955321-6955321 on Build GRCh38
rs376383265 Chr.12: 6955514-6955514 on Build GRCh38
rs368935562 Chr.12: 6955716-6955716 on Build GRCh38
rs782440412 Chr.12: 6955365-6955365 on Build GRCh38
rs200557634 Chr.12: 6955648-6955648 on Build GRCh38
rs782100604 Chr.12: 6955597-6955597 on Build GRCh38
rs1033285581 Chr.12: 6955428-6955428 on Build GRCh38
rs983263472 Chr.12: 6955505-6955505 on Build GRCh38
rs891988424 Chr.12: 6955343-6955343 on Build GRCh38
rs782146589 Chr.12: 6955646-6955646 on Build GRCh38
rs1027985579 Chr.12: 6955649-6955649 on Build GRCh38
rs782686894 Chr.12: 6955540-6955540 on Build GRCh38
rs781936921 Chr.12: 6955623-6955623 on Build GRCh38
rs782350431 Chr.12: 6955631-6955631 on Build GRCh38
rs782710705 Chr.12: 6955573-6955573 on Build GRCh38
rs761631984 Chr.12: 6955697-6955697 on Build GRCh38
rs782819686 Chr.12: 6955688-6955688 on Build GRCh38
rs113260400 Chr.12: 6955608-6955608 on Build GRCh38
rs781982213 Chr.12: 6955639-6955639 on Build GRCh38
rs371508473 Chr.12: 6955614-6955614 on Build GRCh38
rs946215454 Chr.12: 6955581-6955581 on Build GRCh38
rs782082558 Chr.12: 6955627-6955627 on Build GRCh38
rs782099814 Chr.12: 6955506-6955506 on Build GRCh38
rs994508716 Chr.12: 6955630-6955630 on Build GRCh38

View Full Product Details