Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15322175-15322670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRO
Entrez Gene ID
5800
Gene Name
protein tyrosine phosphatase, receptor type O
Gene Aliases
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O
UniGene
Hs.160871
Gene Chromosome Location
Chr.12: 15322257-15598331 on Build GRCh38
Transcripts
NM_002848.3 NM_030667.2 XM_017019725.1

SNP Detail

SNP ID Chromosome Location
rs948902369 Chr.12: 15322526-15322526 on Build GRCh38
rs547765079 Chr.12: 15322254-15322254 on Build GRCh38
rs558321018 Chr.12: 15322324-15322324 on Build GRCh38
rs916164207 Chr.12: 15322499-15322499 on Build GRCh38
rs146286050 Chr.12: 15322201-15322201 on Build GRCh38
rs544374940 Chr.12: 15322585-15322585 on Build GRCh38
rs964501303 Chr.12: 15322253-15322253 on Build GRCh38
rs114714581 Chr.12: 15322542-15322542 on Build GRCh38
rs183981869 Chr.12: 15322274-15322274 on Build GRCh38
rs189099415 Chr.12: 15322316-15322316 on Build GRCh38
rs984656214 Chr.12: 15322206-15322206 on Build GRCh38
rs558772504 Chr.12: 15322536-15322536 on Build GRCh38
rs534308387 Chr.12: 15322465-15322465 on Build GRCh38
rs997308395 Chr.12: 15322636-15322636 on Build GRCh38
rs983068532 Chr.12: 15322463-15322463 on Build GRCh38
rs907275325 Chr.12: 15322372-15322372 on Build GRCh38
rs1045889599 Chr.12: 15322363-15322363 on Build GRCh38
rs1037822813 Chr.12: 15322487-15322487 on Build GRCh38
rs958567901 Chr.12: 15322411-15322411 on Build GRCh38
rs116182634 Chr.12: 15322287-15322287 on Build GRCh38
rs900393754 Chr.12: 15322633-15322633 on Build GRCh38
rs907877177 Chr.12: 15322485-15322485 on Build GRCh38
rs573106509 Chr.12: 15322435-15322435 on Build GRCh38
rs536680245 Chr.12: 15322370-15322370 on Build GRCh38
rs745762607 Chr.12: 15322560-15322562 on Build GRCh38
rs1045891301 Chr.12: 15322533-15322533 on Build GRCh38
rs559858535 Chr.12: 15322269-15322269 on Build GRCh38
rs948850135 Chr.12: 15322358-15322359 on Build GRCh38
rs777358774 Chr.12: 15322200-15322200 on Build GRCh38
rs73057219 Chr.12: 15322293-15322293 on Build GRCh38
rs950008145 Chr.12: 15322439-15322439 on Build GRCh38
rs537638243 Chr.12: 15322309-15322309 on Build GRCh38
rs975879130 Chr.12: 15322453-15322453 on Build GRCh38
rs529667095 Chr.12: 15322203-15322203 on Build GRCh38
rs890591264 Chr.12: 15322565-15322565 on Build GRCh38
rs1006122486 Chr.12: 15322423-15322423 on Build GRCh38
rs1047792519 Chr.12: 15322562-15322562 on Build GRCh38
rs924912902 Chr.12: 15322477-15322477 on Build GRCh38
rs1026707465 Chr.12: 15322400-15322400 on Build GRCh38
rs924892888 Chr.12: 15322297-15322297 on Build GRCh38
rs958465451 Chr.12: 15322632-15322632 on Build GRCh38
rs894678016 Chr.12: 15322343-15322343 on Build GRCh38
rs1038961018 Chr.12: 15322610-15322610 on Build GRCh38
rs977803767 Chr.12: 15322476-15322476 on Build GRCh38
rs999810961 Chr.12: 15322598-15322598 on Build GRCh38
rs940670148 Chr.12: 15322486-15322486 on Build GRCh38
rs1050635183 Chr.12: 15322556-15322556 on Build GRCh38
rs562875174 Chr.12: 15322587-15322587 on Build GRCh38
rs1032754043 Chr.12: 15322624-15322624 on Build GRCh38
rs991318004 Chr.12: 15322416-15322416 on Build GRCh38
rs990426770 Chr.12: 15322493-15322493 on Build GRCh38
rs922331628 Chr.12: 15322404-15322404 on Build GRCh38
rs1009160175 Chr.12: 15322570-15322570 on Build GRCh38
rs1042027718 Chr.12: 15322397-15322397 on Build GRCh38
rs1017360188 Chr.12: 15322209-15322209 on Build GRCh38
rs555020047 Chr.12: 15322425-15322425 on Build GRCh38

View Full Product Details