Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49272172-49272664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBA1C
Entrez Gene ID
84790
Gene Name
tubulin alpha 1c
Gene Aliases
TUBA6, bcm948
UniGene
Hs.652390
Gene Chromosome Location
Chr.12: 49227926-49273338 on Build GRCh38
Transcripts
NM_001303114.1 NM_001303115.1 NM_001303116.1 NM_001303117.1 NM_032704.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940119 c.592T>C p.S198P Chr.12: 49272469-49272469 on Build GRCh38
5636250 c.468T>G p.R156R Chr.12: 49272345-49272345 on Build GRCh38
3359770 c.522G>A p.A174A Chr.12: 49272399-49272399 on Build GRCh38
5640165 c.518C>T p.P173L Chr.12: 49272395-49272395 on Build GRCh38
6278146 c.453G>C p.S151S Chr.12: 49272330-49272330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765475086 Chr.12: 49272318-49272318 on Build GRCh38
rs970897384 Chr.12: 49272344-49272344 on Build GRCh38
rs528051136 Chr.12: 49272396-49272396 on Build GRCh38
rs761488643 Chr.12: 49272264-49272264 on Build GRCh38
rs375327803 Chr.12: 49272204-49272204 on Build GRCh38
rs140286355 Chr.12: 49272630-49272630 on Build GRCh38
rs371686692 Chr.12: 49272241-49272241 on Build GRCh38
rs748763426 Chr.12: 49272395-49272395 on Build GRCh38
rs182625891 Chr.12: 49272240-49272240 on Build GRCh38
rs370276397 Chr.12: 49272538-49272538 on Build GRCh38
rs978664272 Chr.12: 49272351-49272351 on Build GRCh38
rs771017257 Chr.12: 49272562-49272562 on Build GRCh38
rs745415373 Chr.12: 49272221-49272221 on Build GRCh38
rs577787708 Chr.12: 49272196-49272196 on Build GRCh38
rs764409983 Chr.12: 49272517-49272517 on Build GRCh38
rs772849834 Chr.12: 49272238-49272239 on Build GRCh38
rs372926418 Chr.12: 49272530-49272530 on Build GRCh38
rs190363191 Chr.12: 49272219-49272219 on Build GRCh38
rs769228545 Chr.12: 49272228-49272228 on Build GRCh38
rs769178494 Chr.12: 49272238-49272240 on Build GRCh38
rs748002587 Chr.12: 49272406-49272406 on Build GRCh38
rs752824086 Chr.12: 49272326-49272326 on Build GRCh38
rs199599214 Chr.12: 49272369-49272369 on Build GRCh38
rs187663409 Chr.12: 49272325-49272325 on Build GRCh38
rs914503173 Chr.12: 49272202-49272202 on Build GRCh38
rs758736997 Chr.12: 49272330-49272330 on Build GRCh38
rs771693036 Chr.12: 49272411-49272411 on Build GRCh38
rs778628978 Chr.12: 49272535-49272535 on Build GRCh38
rs781724941 Chr.12: 49272362-49272362 on Build GRCh38
rs773210533 Chr.12: 49272476-49272476 on Build GRCh38
rs974687853 Chr.12: 49272550-49272550 on Build GRCh38
rs759689411 Chr.12: 49272304-49272304 on Build GRCh38
rs756652533 Chr.12: 49272528-49272528 on Build GRCh38
rs1025109200 Chr.12: 49272324-49272324 on Build GRCh38
rs763080716 Chr.12: 49272516-49272516 on Build GRCh38
rs373730264 Chr.12: 49272571-49272571 on Build GRCh38
rs559395061 Chr.12: 49272393-49272393 on Build GRCh38
rs113331886 Chr.12: 49272387-49272387 on Build GRCh38
rs11542119 Chr.12: 49272401-49272401 on Build GRCh38
rs777838541 Chr.12: 49272340-49272340 on Build GRCh38
rs771519685 Chr.12: 49272220-49272220 on Build GRCh38
rs369308226 Chr.12: 49272361-49272361 on Build GRCh38
rs374916047 Chr.12: 49272244-49272244 on Build GRCh38
rs778351891 Chr.12: 49272215-49272215 on Build GRCh38
rs780513922 Chr.12: 49272529-49272529 on Build GRCh38
rs565412741 Chr.12: 49272506-49272506 on Build GRCh38
rs771754850 Chr.12: 49272539-49272539 on Build GRCh38
rs762007855 Chr.12: 49272595-49272595 on Build GRCh38
rs933206043 Chr.12: 49272612-49272612 on Build GRCh38
rs767509779 Chr.12: 49272267-49272267 on Build GRCh38
rs750284115 Chr.12: 49272283-49272283 on Build GRCh38
rs375933607 Chr.12: 49272534-49272534 on Build GRCh38
rs750781969 Chr.12: 49272526-49272526 on Build GRCh38
rs371959264 Chr.12: 49272203-49272203 on Build GRCh38
rs746781513 Chr.12: 49272549-49272549 on Build GRCh38
rs561090052 Chr.12: 49272540-49272540 on Build GRCh38
rs767747131 Chr.12: 49272618-49272618 on Build GRCh38
rs376776166 Chr.12: 49272511-49272511 on Build GRCh38
rs1057748 Chr.12: 49272399-49272399 on Build GRCh38
rs751977535 Chr.12: 49272343-49272343 on Build GRCh38
rs192154754 Chr.12: 49272356-49272356 on Build GRCh38
rs371063872 Chr.12: 49272518-49272518 on Build GRCh38
rs762735005 Chr.12: 49272345-49272346 on Build GRCh38
rs760765175 Chr.12: 49272495-49272495 on Build GRCh38
rs776672588 Chr.12: 49272563-49272563 on Build GRCh38
rs774239753 Chr.12: 49272253-49272253 on Build GRCh38
rs747834833 Chr.12: 49272230-49272231 on Build GRCh38
rs774476787 Chr.12: 49272574-49272574 on Build GRCh38

View Full Product Details