Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68852425-68852935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
-
UniGene
Hs.654387
Gene Chromosome Location
Chr.12: 68842200-68963240 on Build GRCh38
Transcripts
NM_001005502.2 NM_001874.4 NM_198320.3

SNP Detail

SNP ID Chromosome Location
rs376929375 Chr.12: 68852547-68852548 on Build GRCh38
rs370407199 Chr.12: 68852493-68852493 on Build GRCh38
rs1018744583 Chr.12: 68852552-68852552 on Build GRCh38
rs981346124 Chr.12: 68852866-68852866 on Build GRCh38
rs1020773452 Chr.12: 68852465-68852466 on Build GRCh38
rs1043177480 Chr.12: 68852465-68852465 on Build GRCh38
rs926996923 Chr.12: 68852628-68852628 on Build GRCh38
rs530495121 Chr.12: 68852548-68852548 on Build GRCh38
rs138943330 Chr.12: 68852583-68852583 on Build GRCh38
rs532919792 Chr.12: 68852509-68852509 on Build GRCh38
rs866941126 Chr.12: 68852549-68852549 on Build GRCh38
rs571583491 Chr.12: 68852718-68852718 on Build GRCh38
rs978545022 Chr.12: 68852620-68852620 on Build GRCh38
rs1009789337 Chr.12: 68852839-68852839 on Build GRCh38
rs1010247367 Chr.12: 68852564-68852564 on Build GRCh38
rs886583222 Chr.12: 68852511-68852511 on Build GRCh38
rs541320376 Chr.12: 68852466-68852466 on Build GRCh38
rs1046982198 Chr.12: 68852905-68852905 on Build GRCh38
rs547271657 Chr.12: 68852708-68852708 on Build GRCh38
rs905542687 Chr.12: 68852468-68852468 on Build GRCh38
rs1057074507 Chr.12: 68852486-68852486 on Build GRCh38
rs138012936 Chr.12: 68852719-68852719 on Build GRCh38
rs937063150 Chr.12: 68852678-68852678 on Build GRCh38
rs763187919 Chr.12: 68852686-68852686 on Build GRCh38
rs946847657 Chr.12: 68852609-68852609 on Build GRCh38
rs771350159 Chr.12: 68852715-68852715 on Build GRCh38
rs1056811287 Chr.12: 68852693-68852693 on Build GRCh38
rs895764191 Chr.12: 68852704-68852704 on Build GRCh38
rs974429545 Chr.12: 68852697-68852697 on Build GRCh38
rs966529875 Chr.12: 68852599-68852599 on Build GRCh38
rs143490827 Chr.12: 68852824-68852824 on Build GRCh38
rs118177787 Chr.12: 68852453-68852453 on Build GRCh38
rs575809571 Chr.12: 68852852-68852852 on Build GRCh38
rs55947127 Chr.12: 68852569-68852570 on Build GRCh38
rs1054398009 Chr.12: 68852835-68852835 on Build GRCh38
rs1047956171 Chr.12: 68852500-68852500 on Build GRCh38
rs868629663 Chr.12: 68852797-68852797 on Build GRCh38
rs143310129 Chr.12: 68852741-68852741 on Build GRCh38
rs555638988 Chr.12: 68852907-68852907 on Build GRCh38
rs916937218 Chr.12: 68852889-68852889 on Build GRCh38
rs893309456 Chr.12: 68852602-68852602 on Build GRCh38
rs540840265 Chr.12: 68852491-68852491 on Build GRCh38
rs12581244 Chr.12: 68852672-68852672 on Build GRCh38
rs992516812 Chr.12: 68852879-68852879 on Build GRCh38
rs920382197 Chr.12: 68852490-68852491 on Build GRCh38
rs947141688 Chr.12: 68852461-68852461 on Build GRCh38
rs138141526 Chr.12: 68852538-68852541 on Build GRCh38
rs1152927 Chr.12: 68852877-68852877 on Build GRCh38
rs774664936 Chr.12: 68852845-68852845 on Build GRCh38
rs899910994 Chr.12: 68852782-68852782 on Build GRCh38
rs936969653 Chr.12: 68852470-68852470 on Build GRCh38
rs1029603379 Chr.12: 68852805-68852805 on Build GRCh38
rs563410889 Chr.12: 68852547-68852547 on Build GRCh38
rs911549692 Chr.12: 68852473-68852473 on Build GRCh38
rs1054365109 Chr.12: 68852556-68852556 on Build GRCh38
rs974399872 Chr.12: 68852487-68852487 on Build GRCh38
rs772423594 Chr.12: 68852550-68852553 on Build GRCh38
rs372119090 Chr.12: 68852681-68852681 on Build GRCh38
rs143257504 Chr.12: 68852603-68852603 on Build GRCh38
rs369578653 Chr.12: 68852853-68852863 on Build GRCh38
rs369410287 Chr.12: 68852475-68852475 on Build GRCh38
rs386763970 Chr.12: 68852548-68852553 on Build GRCh38
rs968248414 Chr.12: 68852863-68852863 on Build GRCh38
rs929598867 Chr.12: 68852903-68852903 on Build GRCh38
rs796897432 Chr.12: 68852546-68852547 on Build GRCh38
rs893113906 Chr.12: 68852837-68852837 on Build GRCh38
rs368680151 Chr.12: 68852549-68852552 on Build GRCh38
rs957447221 Chr.12: 68852449-68852449 on Build GRCh38
rs950214824 Chr.12: 68852785-68852785 on Build GRCh38
rs964622385 Chr.12: 68852690-68852690 on Build GRCh38
rs569067532 Chr.12: 68852732-68852733 on Build GRCh38
rs983199563 Chr.12: 68852733-68852733 on Build GRCh38
rs1055074187 Chr.12: 68852472-68852472 on Build GRCh38
rs976680173 Chr.12: 68852562-68852562 on Build GRCh38
rs112411459 Chr.12: 68852853-68852853 on Build GRCh38
rs964083883 Chr.12: 68852554-68852554 on Build GRCh38
rs908040931 Chr.12: 68852733-68852733 on Build GRCh38
rs1029906626 Chr.12: 68852563-68852563 on Build GRCh38
rs549067021 Chr.12: 68852540-68852540 on Build GRCh38
rs1025469676 Chr.12: 68852541-68852541 on Build GRCh38
rs1018361670 Chr.12: 68852761-68852761 on Build GRCh38
rs1010279774 Chr.12: 68852842-68852842 on Build GRCh38
rs545322481 Chr.12: 68852556-68852557 on Build GRCh38
rs929860532 Chr.12: 68852705-68852705 on Build GRCh38
rs535026169 Chr.12: 68852669-68852669 on Build GRCh38
rs559131483 Chr.12: 68852471-68852471 on Build GRCh38
rs988294593 Chr.12: 68852596-68852596 on Build GRCh38
rs895733115 Chr.12: 68852512-68852512 on Build GRCh38
rs913014559 Chr.12: 68852454-68852454 on Build GRCh38
rs773595366 Chr.12: 68852457-68852457 on Build GRCh38
rs927098707 Chr.12: 68852876-68852876 on Build GRCh38

View Full Product Details