Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11658614-11659109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
UniGene
Hs.556006
Gene Chromosome Location
Chr.1: 11654375-11663327 on Build GRCh38
Transcripts
NM_001014765.1 NM_001304790.1 NM_001304791.1 NM_033182.5 NM_183412.2 NM_183413.2 NR_130909.1 XM_005263535.1 XM_005263536.4 XM_005263537.1 XM_006711043.2 XM_006711045.2 XM_011542435.1 XM_017002842.1 XM_017002843.1 XM_017002844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1206857 c.550G>A p.A184T Chr.1: 11658797-11658797 on Build GRCh38
6275695 c.525G>T p.Q175H Chr.1: 11658772-11658772 on Build GRCh38
1196637 c.552G>C p.A184A Chr.1: 11658799-11658799 on Build GRCh38
7416980 c.588G>T p.A196A Chr.1: 11658835-11658835 on Build GRCh38
3471908 c.492C>T p.F164F Chr.1: 11658739-11658739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764944524 Chr.1: 11658688-11658688 on Build GRCh38
rs200779554 Chr.1: 11658643-11658643 on Build GRCh38
rs201882677 Chr.1: 11658698-11658698 on Build GRCh38
rs759439362 Chr.1: 11658726-11658726 on Build GRCh38
rs907478811 Chr.1: 11658829-11658829 on Build GRCh38
rs747192108 Chr.1: 11658767-11658767 on Build GRCh38
rs565261668 Chr.1: 11658703-11658703 on Build GRCh38
rs759276267 Chr.1: 11658776-11658776 on Build GRCh38
rs754397708 Chr.1: 11658934-11658934 on Build GRCh38
rs918112096 Chr.1: 11659075-11659075 on Build GRCh38
rs553003864 Chr.1: 11658891-11658891 on Build GRCh38
rs747162819 Chr.1: 11658812-11658812 on Build GRCh38
rs371459091 Chr.1: 11658875-11658875 on Build GRCh38
rs780557168 Chr.1: 11658755-11658755 on Build GRCh38
rs951119532 Chr.1: 11658680-11658680 on Build GRCh38
rs74054208 Chr.1: 11658704-11658704 on Build GRCh38
rs889457022 Chr.1: 11658902-11658902 on Build GRCh38
rs753718849 Chr.1: 11658656-11658656 on Build GRCh38
rs572795365 Chr.1: 11658683-11658683 on Build GRCh38
rs760967785 Chr.1: 11658711-11658711 on Build GRCh38
rs377017011 Chr.1: 11658773-11658773 on Build GRCh38
rs374979753 Chr.1: 11658904-11658904 on Build GRCh38
rs750041756 Chr.1: 11658888-11658888 on Build GRCh38
rs375500495 Chr.1: 11658667-11658667 on Build GRCh38
rs529948237 Chr.1: 11658774-11658774 on Build GRCh38
rs752667911 Chr.1: 11658743-11658755 on Build GRCh38
rs754966515 Chr.1: 11658665-11658665 on Build GRCh38
rs376877549 Chr.1: 11658675-11658675 on Build GRCh38
rs776793625 Chr.1: 11658831-11658831 on Build GRCh38
rs748590247 Chr.1: 11658803-11658803 on Build GRCh38
rs749723056 Chr.1: 11658757-11658757 on Build GRCh38
rs367623463 Chr.1: 11658835-11658835 on Build GRCh38
rs756987946 Chr.1: 11658751-11658751 on Build GRCh38
rs776387122 Chr.1: 11658775-11658775 on Build GRCh38
rs372518301 Chr.1: 11658758-11658758 on Build GRCh38
rs552972903 Chr.1: 11658682-11658682 on Build GRCh38
rs780575796 Chr.1: 11658692-11658692 on Build GRCh38
rs751144824 Chr.1: 11658792-11658792 on Build GRCh38
rs952548622 Chr.1: 11658955-11658955 on Build GRCh38
rs202154565 Chr.1: 11658786-11658786 on Build GRCh38
rs763495084 Chr.1: 11658790-11658790 on Build GRCh38
rs984888283 Chr.1: 11659000-11659000 on Build GRCh38
rs985356444 Chr.1: 11658736-11658736 on Build GRCh38
rs369440151 Chr.1: 11658673-11658673 on Build GRCh38
rs202223354 Chr.1: 11658779-11658779 on Build GRCh38
rs767233539 Chr.1: 11658797-11658797 on Build GRCh38
rs148817401 Chr.1: 11658762-11658762 on Build GRCh38
rs948734081 Chr.1: 11659003-11659003 on Build GRCh38
rs12746243 Chr.1: 11658659-11658659 on Build GRCh38
rs376358727 Chr.1: 11658805-11658805 on Build GRCh38
rs774181465 Chr.1: 11658874-11658874 on Build GRCh38
rs760462829 Chr.1: 11658890-11658890 on Build GRCh38
rs111261340 Chr.1: 11658876-11658876 on Build GRCh38
rs962703413 Chr.1: 11658977-11658977 on Build GRCh38
rs1015436401 Chr.1: 11658972-11658972 on Build GRCh38
rs752651963 Chr.1: 11658733-11658733 on Build GRCh38
rs142346784 Chr.1: 11658740-11658740 on Build GRCh38
rs374387099 Chr.1: 11658796-11658796 on Build GRCh38
rs752037754 Chr.1: 11658908-11658908 on Build GRCh38
rs762726808 Chr.1: 11658819-11658839 on Build GRCh38
rs965241684 Chr.1: 11658933-11658933 on Build GRCh38
rs775410949 Chr.1: 11658842-11658842 on Build GRCh38
rs113666189 Chr.1: 11658813-11658813 on Build GRCh38
rs980535512 Chr.1: 11659019-11659019 on Build GRCh38
rs201438913 Chr.1: 11658834-11658834 on Build GRCh38
rs773242935 Chr.1: 11658706-11658706 on Build GRCh38
rs543294017 Chr.1: 11658685-11658685 on Build GRCh38
rs758103142 Chr.1: 11658780-11658780 on Build GRCh38
rs751112563 Chr.1: 11658745-11658745 on Build GRCh38
rs878970006 Chr.1: 11658744-11658744 on Build GRCh38
rs1033776951 Chr.1: 11659021-11659021 on Build GRCh38
rs928673780 Chr.1: 11659034-11659034 on Build GRCh38
rs992252309 Chr.1: 11659061-11659061 on Build GRCh38
rs200078448 Chr.1: 11658686-11658686 on Build GRCh38
rs765282099 Chr.1: 11658708-11658708 on Build GRCh38
rs369457346 Chr.1: 11658712-11658712 on Build GRCh38
rs938195425 Chr.1: 11658793-11658793 on Build GRCh38
rs766203545 Chr.1: 11658653-11658653 on Build GRCh38
rs1051409912 Chr.1: 11658886-11658886 on Build GRCh38
rs995873863 Chr.1: 11658997-11658997 on Build GRCh38
rs145966428 Chr.1: 11658804-11658804 on Build GRCh38
rs371453824 Chr.1: 11658895-11658895 on Build GRCh38
rs150816569 Chr.1: 11658802-11658802 on Build GRCh38
rs1044642175 Chr.1: 11658819-11658819 on Build GRCh38
rs535431374 Chr.1: 11659023-11659023 on Build GRCh38
rs1025397397 Chr.1: 11658942-11658942 on Build GRCh38
rs1054216806 Chr.1: 11658854-11658854 on Build GRCh38
rs534641827 Chr.1: 11658679-11658679 on Build GRCh38
rs1004115444 Chr.1: 11658937-11658937 on Build GRCh38
rs1019508344 Chr.1: 11658919-11658919 on Build GRCh38
rs112449604 Chr.1: 11658674-11658674 on Build GRCh38
rs924300706 Chr.1: 11658839-11658839 on Build GRCh38
rs771019879 Chr.1: 11658821-11658821 on Build GRCh38
rs180915564 Chr.1: 11658700-11658700 on Build GRCh38
rs143936849 Chr.1: 11658850-11658850 on Build GRCh38
rs200887520 Chr.1: 11658739-11658739 on Build GRCh38
rs771989360 Chr.1: 11658765-11658765 on Build GRCh38
rs749796397 Chr.1: 11658694-11658694 on Build GRCh38
rs750684404 Chr.1: 11658644-11658644 on Build GRCh38
rs909321208 Chr.1: 11659001-11659001 on Build GRCh38
rs1003075233 Chr.1: 11658681-11658681 on Build GRCh38
rs758809690 Chr.1: 11658900-11658900 on Build GRCh38
rs762959609 Chr.1: 11658787-11658787 on Build GRCh38
rs555008441 Chr.1: 11659044-11659044 on Build GRCh38
rs765207374 Chr.1: 11658729-11658729 on Build GRCh38
rs773401009 Chr.1: 11658766-11658766 on Build GRCh38
rs750135661 Chr.1: 11658798-11658798 on Build GRCh38

View Full Product Details