Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102731307-102731820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
UniGene
Hs.368382
Gene Chromosome Location
Chr.13: 102729367-102759072 on Build GRCh38
Transcripts
NM_001146197.1 XM_011521106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4650322 c.5195A>G p.D1732G Chr.13: 102731615-102731615 on Build GRCh38
7225005 c.5072C>T p.P1691L Chr.13: 102731738-102731738 on Build GRCh38
5728982 c.5364T>C p.T1788T Chr.13: 102731446-102731446 on Build GRCh38
1365262 c.5194G>T p.D1732Y Chr.13: 102731616-102731616 on Build GRCh38
401439 c.5108C>G p.A1703G Chr.13: 102731702-102731702 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748392683 Chr.13: 102731590-102731590 on Build GRCh38
rs953158663 Chr.13: 102731460-102731460 on Build GRCh38
rs755363736 Chr.13: 102731585-102731585 on Build GRCh38
rs1014990515 Chr.13: 102731350-102731350 on Build GRCh38
rs746285561 Chr.13: 102731403-102731403 on Build GRCh38
rs780661259 Chr.13: 102731576-102731576 on Build GRCh38
rs555334685 Chr.13: 102731416-102731416 on Build GRCh38
rs775089892 Chr.13: 102731677-102731677 on Build GRCh38
rs749863534 Chr.13: 102731377-102731377 on Build GRCh38
rs76643569 Chr.13: 102731442-102731442 on Build GRCh38
rs73587211 Chr.13: 102731509-102731509 on Build GRCh38
rs72653638 Chr.13: 102731422-102731422 on Build GRCh38
rs375966075 Chr.13: 102731704-102731704 on Build GRCh38
rs377506995 Chr.13: 102731463-102731463 on Build GRCh38
rs997973321 Chr.13: 102731415-102731415 on Build GRCh38
rs779174080 Chr.13: 102731341-102731341 on Build GRCh38
rs967684279 Chr.13: 102731493-102731493 on Build GRCh38
rs866478231 Chr.13: 102731788-102731788 on Build GRCh38
rs74351854 Chr.13: 102731737-102731737 on Build GRCh38
rs368923115 Chr.13: 102731720-102731720 on Build GRCh38
rs867648973 Chr.13: 102731718-102731718 on Build GRCh38
rs541214181 Chr.13: 102731514-102731514 on Build GRCh38
rs543331120 Chr.13: 102731393-102731393 on Build GRCh38
rs79250647 Chr.13: 102731647-102731647 on Build GRCh38
rs942934566 Chr.13: 102731359-102731359 on Build GRCh38
rs931457076 Chr.13: 102731537-102731537 on Build GRCh38
rs910142030 Chr.13: 102731354-102731354 on Build GRCh38
rs747039599 Chr.13: 102731606-102731606 on Build GRCh38
rs991947588 Chr.13: 102731525-102731525 on Build GRCh38
rs765652233 Chr.13: 102731794-102731794 on Build GRCh38
rs147982063 Chr.13: 102731343-102731343 on Build GRCh38
rs764834970 Chr.13: 102731451-102731451 on Build GRCh38
rs747566959 Chr.13: 102731592-102731595 on Build GRCh38
rs146908292 Chr.13: 102731601-102731601 on Build GRCh38
rs1000420057 Chr.13: 102731500-102731500 on Build GRCh38
rs534716273 Chr.13: 102731784-102731784 on Build GRCh38
rs779072083 Chr.13: 102731587-102731587 on Build GRCh38
rs901466912 Chr.13: 102731666-102731666 on Build GRCh38
rs554350172 Chr.13: 102731351-102731351 on Build GRCh38
rs1021530630 Chr.13: 102731488-102731488 on Build GRCh38
rs762669247 Chr.13: 102731679-102731679 on Build GRCh38
rs762428718 Chr.13: 102731491-102731491 on Build GRCh38
rs565198850 Chr.13: 102731501-102731501 on Build GRCh38
rs189753227 Chr.13: 102731734-102731734 on Build GRCh38
rs779400148 Chr.13: 102731345-102731345 on Build GRCh38
rs372262515 Chr.13: 102731698-102731698 on Build GRCh38
rs548058110 Chr.13: 102731642-102731642 on Build GRCh38
rs776430354 Chr.13: 102731433-102731433 on Build GRCh38
rs999137397 Chr.13: 102731787-102731787 on Build GRCh38
rs1021457234 Chr.13: 102731719-102731719 on Build GRCh38
rs770901472 Chr.13: 102731624-102731624 on Build GRCh38
rs545297083 Chr.13: 102731653-102731653 on Build GRCh38
rs754393763 Chr.13: 102731778-102731778 on Build GRCh38
rs989996953 Chr.13: 102731477-102731477 on Build GRCh38
rs769617641 Chr.13: 102731671-102731671 on Build GRCh38
rs758495311 Chr.13: 102731592-102731592 on Build GRCh38
rs775228604 Chr.13: 102731474-102731474 on Build GRCh38
rs186534782 Chr.13: 102731566-102731566 on Build GRCh38
rs754305885 Chr.13: 102731583-102731583 on Build GRCh38
rs565344019 Chr.13: 102731609-102731609 on Build GRCh38
rs930129695 Chr.13: 102731348-102731348 on Build GRCh38
rs746445815 Chr.13: 102731532-102731532 on Build GRCh38
rs572722039 Chr.13: 102731374-102731374 on Build GRCh38
rs760622860 Chr.13: 102731402-102731402 on Build GRCh38
rs530389215 Chr.13: 102731670-102731670 on Build GRCh38
rs767033172 Chr.13: 102731738-102731738 on Build GRCh38

View Full Product Details