Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 34567561-34568075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX6
Entrez Gene ID
58533
Gene Name
sorting nexin 6
Gene Aliases
MSTP010, TFAF2
UniGene
Hs.356647
Gene Chromosome Location
Chr.14: 34561409-34630160 on Build GRCh38
Transcripts
NM_021249.3 NM_152233.2 XM_006720224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053925 c.1010T>C p.V337A Chr.14: 34567925-34567925 on Build GRCh38
7260686 c.1060_1061insA p.I354fs*3 Chr.14: 34567874-34567875 on Build GRCh38
5629320 c.994T>A p.Y332N Chr.14: 34567977-34567977 on Build GRCh38
4460158 c.1155C>G p.F385L Chr.14: 34567734-34567734 on Build GRCh38
5629318 c.995A>T p.Y332F Chr.14: 34567976-34567976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776786467 Chr.14: 34567873-34567873 on Build GRCh38
rs748069262 Chr.14: 34567808-34567809 on Build GRCh38
rs780629260 Chr.14: 34567821-34567821 on Build GRCh38
rs762194265 Chr.14: 34567793-34567793 on Build GRCh38
rs187257640 Chr.14: 34568000-34568000 on Build GRCh38
rs139160191 Chr.14: 34567984-34567984 on Build GRCh38
rs746373030 Chr.14: 34567681-34567681 on Build GRCh38
rs374064886 Chr.14: 34568034-34568034 on Build GRCh38
rs899579684 Chr.14: 34567599-34567599 on Build GRCh38
rs201442232 Chr.14: 34567934-34567934 on Build GRCh38
rs766624633 Chr.14: 34568044-34568044 on Build GRCh38
rs1053640744 Chr.14: 34567649-34567649 on Build GRCh38
rs1028791309 Chr.14: 34567995-34567995 on Build GRCh38
rs951056361 Chr.14: 34567619-34567619 on Build GRCh38
rs759694816 Chr.14: 34568042-34568042 on Build GRCh38
rs766231179 Chr.14: 34567647-34567647 on Build GRCh38
rs764130738 Chr.14: 34567775-34567775 on Build GRCh38
rs768089111 Chr.14: 34567724-34567724 on Build GRCh38
rs759451423 Chr.14: 34567867-34567867 on Build GRCh38
rs763349093 Chr.14: 34567899-34567899 on Build GRCh38
rs746481560 Chr.14: 34567715-34567715 on Build GRCh38
rs764336286 Chr.14: 34567652-34567652 on Build GRCh38
rs748201916 Chr.14: 34568029-34568029 on Build GRCh38
rs899715470 Chr.14: 34567914-34567914 on Build GRCh38
rs758911433 Chr.14: 34567693-34567693 on Build GRCh38
rs776322454 Chr.14: 34567726-34567726 on Build GRCh38
rs182878648 Chr.14: 34567850-34567850 on Build GRCh38
rs769378117 Chr.14: 34567837-34567837 on Build GRCh38
rs1048141943 Chr.14: 34567941-34567941 on Build GRCh38
rs780443143 Chr.14: 34567708-34567708 on Build GRCh38
rs11156848 Chr.14: 34567802-34567802 on Build GRCh38
rs773116752 Chr.14: 34567642-34567642 on Build GRCh38
rs763149034 Chr.14: 34568038-34568038 on Build GRCh38
rs752097622 Chr.14: 34567908-34567908 on Build GRCh38
rs773551472 Chr.14: 34568036-34568036 on Build GRCh38
rs146496878 Chr.14: 34567903-34567903 on Build GRCh38
rs879376341 Chr.14: 34567610-34567610 on Build GRCh38
rs748990249 Chr.14: 34567989-34567989 on Build GRCh38
rs374475021 Chr.14: 34567790-34567790 on Build GRCh38
rs190789373 Chr.14: 34567844-34567844 on Build GRCh38
rs769885941 Chr.14: 34567844-34567845 on Build GRCh38
rs752153327 Chr.14: 34567956-34567956 on Build GRCh38
rs771850249 Chr.14: 34568017-34568017 on Build GRCh38
rs778943998 Chr.14: 34567964-34567964 on Build GRCh38
rs777426040 Chr.14: 34567842-34567842 on Build GRCh38
rs760779667 Chr.14: 34567769-34567769 on Build GRCh38
rs371653687 Chr.14: 34568053-34568053 on Build GRCh38
rs775595628 Chr.14: 34568028-34568028 on Build GRCh38
rs144963273 Chr.14: 34567594-34567594 on Build GRCh38
rs780574313 Chr.14: 34567806-34567806 on Build GRCh38
rs747657671 Chr.14: 34567823-34567823 on Build GRCh38
rs199807355 Chr.14: 34568048-34568048 on Build GRCh38
rs754048194 Chr.14: 34567660-34567660 on Build GRCh38
rs758715322 Chr.14: 34567805-34567805 on Build GRCh38
rs765511727 Chr.14: 34567883-34567883 on Build GRCh38
rs983407682 Chr.14: 34567700-34567700 on Build GRCh38
rs770851145 Chr.14: 34567759-34567759 on Build GRCh38
rs745680570 Chr.14: 34568016-34568016 on Build GRCh38
rs768777521 Chr.14: 34567870-34567870 on Build GRCh38
rs762002042 Chr.14: 34567878-34567878 on Build GRCh38
rs769569157 Chr.14: 34567737-34567737 on Build GRCh38
rs957571588 Chr.14: 34568035-34568035 on Build GRCh38
rs985444819 Chr.14: 34567603-34567603 on Build GRCh38
rs747851305 Chr.14: 34567733-34567733 on Build GRCh38
rs936080802 Chr.14: 34567613-34567613 on Build GRCh38
rs752052091 Chr.14: 34567809-34567809 on Build GRCh38
rs762750251 Chr.14: 34567743-34567743 on Build GRCh38
rs765564940 Chr.14: 34567801-34567801 on Build GRCh38
rs765062347 Chr.14: 34567641-34567641 on Build GRCh38
rs200347901 Chr.14: 34567915-34567915 on Build GRCh38
rs979798362 Chr.14: 34568024-34568024 on Build GRCh38
rs886747737 Chr.14: 34567961-34567961 on Build GRCh38
rs778640726 Chr.14: 34568013-34568013 on Build GRCh38
rs754495132 Chr.14: 34567816-34567816 on Build GRCh38
rs757676452 Chr.14: 34567675-34567675 on Build GRCh38
rs10149570 Chr.14: 34567843-34567843 on Build GRCh38
rs142172227 Chr.14: 34567765-34567765 on Build GRCh38
rs369365933 Chr.14: 34567710-34567710 on Build GRCh38
rs147604232 Chr.14: 34567740-34567740 on Build GRCh38
rs567128446 Chr.14: 34567662-34567662 on Build GRCh38
rs756167870 Chr.14: 34567651-34567651 on Build GRCh38
rs752763929 Chr.14: 34567650-34567650 on Build GRCh38
rs762941017 Chr.14: 34567646-34567646 on Build GRCh38
rs960462452 Chr.14: 34567750-34567750 on Build GRCh38
rs879875112 Chr.14: 34567965-34567965 on Build GRCh38
rs78342473 Chr.14: 34567807-34567807 on Build GRCh38
rs767971374 Chr.14: 34568050-34568050 on Build GRCh38

View Full Product Details