Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36298770-36299265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBIP
Entrez Gene ID
51562
Gene Name
MAP3K12 binding inhibitory protein 1
Gene Aliases
-
UniGene
Hs.368647
Gene Chromosome Location
Chr.14: 36298558-36320676 on Build GRCh38
Transcripts
NM_001144891.1 NM_001308110.1 NM_016586.2 XM_005267754.3 XM_005267756.4 XM_017021367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1168785 c.984C>T p.L328L Chr.14: 36299134-36299134 on Build GRCh38
3495892 c.1031C>T p.P344L Chr.14: 36299087-36299087 on Build GRCh38
4945295 c.987C>G p.L329L Chr.14: 36299131-36299131 on Build GRCh38
3814763 c.1025A>C p.H342P Chr.14: 36299093-36299093 on Build GRCh38
7111596 c.998G>A p.R333K Chr.14: 36299120-36299120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17849936 Chr.14: 36299089-36299089 on Build GRCh38
rs751614247 Chr.14: 36299216-36299216 on Build GRCh38
rs1036324878 Chr.14: 36299148-36299148 on Build GRCh38
rs1023388909 Chr.14: 36298952-36298952 on Build GRCh38
rs978481651 Chr.14: 36299042-36299042 on Build GRCh38
rs868073530 Chr.14: 36299166-36299166 on Build GRCh38
rs369598362 Chr.14: 36298840-36298840 on Build GRCh38
rs774710795 Chr.14: 36299056-36299056 on Build GRCh38
rs763621996 Chr.14: 36299145-36299145 on Build GRCh38
rs956560323 Chr.14: 36299011-36299011 on Build GRCh38
rs905944340 Chr.14: 36298822-36298822 on Build GRCh38
rs370304446 Chr.14: 36299075-36299075 on Build GRCh38
rs1049602260 Chr.14: 36299083-36299083 on Build GRCh38
rs374762601 Chr.14: 36299106-36299106 on Build GRCh38
rs376202361 Chr.14: 36299048-36299048 on Build GRCh38
rs747046528 Chr.14: 36299105-36299105 on Build GRCh38
rs1027703271 Chr.14: 36298898-36298898 on Build GRCh38
rs752454840 Chr.14: 36299076-36299076 on Build GRCh38
rs765868169 Chr.14: 36299039-36299039 on Build GRCh38
rs755047481 Chr.14: 36299220-36299220 on Build GRCh38
rs147919885 Chr.14: 36299113-36299113 on Build GRCh38
rs777509253 Chr.14: 36299087-36299087 on Build GRCh38
rs149882078 Chr.14: 36298902-36298902 on Build GRCh38
rs757193691 Chr.14: 36299202-36299202 on Build GRCh38
rs200795735 Chr.14: 36299066-36299066 on Build GRCh38
rs1013456717 Chr.14: 36298934-36298934 on Build GRCh38
rs969205434 Chr.14: 36298997-36298997 on Build GRCh38
rs781101416 Chr.14: 36299240-36299240 on Build GRCh38
rs912217005 Chr.14: 36299027-36299027 on Build GRCh38
rs763938430 Chr.14: 36299191-36299191 on Build GRCh38
rs767840178 Chr.14: 36298999-36298999 on Build GRCh38
rs773698376 Chr.14: 36299133-36299133 on Build GRCh38
rs953704090 Chr.14: 36299008-36299008 on Build GRCh38
rs1019173138 Chr.14: 36298911-36298911 on Build GRCh38
rs896440055 Chr.14: 36298918-36298918 on Build GRCh38
rs773816893 Chr.14: 36299057-36299057 on Build GRCh38
rs748555600 Chr.14: 36299118-36299118 on Build GRCh38
rs35822592 Chr.14: 36298850-36298851 on Build GRCh38
rs1004916622 Chr.14: 36298903-36298903 on Build GRCh38
rs865815138 Chr.14: 36299103-36299104 on Build GRCh38
rs773746033 Chr.14: 36299153-36299153 on Build GRCh38
rs571382371 Chr.14: 36299183-36299183 on Build GRCh38
rs1000683214 Chr.14: 36299205-36299205 on Build GRCh38
rs760295782 Chr.14: 36299190-36299190 on Build GRCh38
rs987864947 Chr.14: 36299015-36299018 on Build GRCh38
rs555226736 Chr.14: 36298825-36298825 on Build GRCh38
rs537837993 Chr.14: 36299096-36299096 on Build GRCh38
rs116641058 Chr.14: 36299055-36299055 on Build GRCh38
rs994841073 Chr.14: 36298859-36298859 on Build GRCh38
rs1049109801 Chr.14: 36299179-36299179 on Build GRCh38
rs886579675 Chr.14: 36298901-36298901 on Build GRCh38
rs979126497 Chr.14: 36299004-36299004 on Build GRCh38
rs759035265 Chr.14: 36299169-36299169 on Build GRCh38
rs759736512 Chr.14: 36299104-36299105 on Build GRCh38
rs763524252 Chr.14: 36299062-36299062 on Build GRCh38
rs753023308 Chr.14: 36299140-36299143 on Build GRCh38
rs753863204 Chr.14: 36299185-36299185 on Build GRCh38
rs976444811 Chr.14: 36298956-36298956 on Build GRCh38
rs767626898 Chr.14: 36299120-36299120 on Build GRCh38
rs780069019 Chr.14: 36299097-36299097 on Build GRCh38
rs79341640 Chr.14: 36299071-36299071 on Build GRCh38
rs966721642 Chr.14: 36298950-36298950 on Build GRCh38
rs757317256 Chr.14: 36299204-36299204 on Build GRCh38
rs4900833 Chr.14: 36299070-36299070 on Build GRCh38
rs17175248 Chr.14: 36299201-36299201 on Build GRCh38
rs771400986 Chr.14: 36299149-36299149 on Build GRCh38
rs770324179 Chr.14: 36299120-36299120 on Build GRCh38
rs755859738 Chr.14: 36299084-36299084 on Build GRCh38

View Full Product Details