Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 36314551-36315070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBIP
Entrez Gene ID
51562
Gene Name
MAP3K12 binding inhibitory protein 1
Gene Aliases
-
UniGene
Hs.368647
Gene Chromosome Location
Chr.14: 36298558-36320676 on Build GRCh38
Transcripts
NM_001144891.1 NM_001308110.1 NM_016586.2 XM_005267754.3 XM_005267756.4 XM_017021367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3706263 c.508A>T p.K170* Chr.14: 36314575-36314575 on Build GRCh38
162403 c.517G>C p.E173Q Chr.14: 36314566-36314566 on Build GRCh38
5832764 c.513delA p.A172fs*40 Chr.14: 36314570-36314570 on Build GRCh38
3495893 c.360C>T p.S120S Chr.14: 36314805-36314805 on Build GRCh38
6709107 c.424_425delCA p.Q142fs*4 Chr.14: 36314740-36314741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756037339 Chr.14: 36314645-36314645 on Build GRCh38
rs370022879 Chr.14: 36314823-36314823 on Build GRCh38
rs754695745 Chr.14: 36314798-36314800 on Build GRCh38
rs926271042 Chr.14: 36314921-36314923 on Build GRCh38
rs114544697 Chr.14: 36314640-36314640 on Build GRCh38
rs371260304 Chr.14: 36314579-36314579 on Build GRCh38
rs373272751 Chr.14: 36314832-36314832 on Build GRCh38
rs781069288 Chr.14: 36314877-36314879 on Build GRCh38
rs745900941 Chr.14: 36314653-36314653 on Build GRCh38
rs767365791 Chr.14: 36314728-36314728 on Build GRCh38
rs900958407 Chr.14: 36314634-36314634 on Build GRCh38
rs752418798 Chr.14: 36314753-36314753 on Build GRCh38
rs781517721 Chr.14: 36314950-36314950 on Build GRCh38
rs150300355 Chr.14: 36314667-36314667 on Build GRCh38
rs748556016 Chr.14: 36314955-36314955 on Build GRCh38
rs773729671 Chr.14: 36314854-36314854 on Build GRCh38
rs971894605 Chr.14: 36314793-36314793 on Build GRCh38
rs759852538 Chr.14: 36314806-36314806 on Build GRCh38
rs1053868596 Chr.14: 36314957-36314957 on Build GRCh38
rs145521896 Chr.14: 36314799-36314799 on Build GRCh38
rs1034494992 Chr.14: 36314790-36314790 on Build GRCh38
rs1010690973 Chr.14: 36314727-36314727 on Build GRCh38
rs760420589 Chr.14: 36314890-36314890 on Build GRCh38
rs776345733 Chr.14: 36314614-36314616 on Build GRCh38
rs370912794 Chr.14: 36314618-36314618 on Build GRCh38
rs755773197 Chr.14: 36314974-36314975 on Build GRCh38
rs755265717 Chr.14: 36314945-36314945 on Build GRCh38
rs748466016 Chr.14: 36314835-36314835 on Build GRCh38
rs759190033 Chr.14: 36314859-36314859 on Build GRCh38
rs971855516 Chr.14: 36314762-36314762 on Build GRCh38
rs762921881 Chr.14: 36314653-36314655 on Build GRCh38
rs777635730 Chr.14: 36314771-36314771 on Build GRCh38
rs202097612 Chr.14: 36314639-36314639 on Build GRCh38
rs752939609 Chr.14: 36314936-36314936 on Build GRCh38
rs780286223 Chr.14: 36314666-36314666 on Build GRCh38
rs752238204 Chr.14: 36314955-36314971 on Build GRCh38
rs777320509 Chr.14: 36314589-36314589 on Build GRCh38
rs115444173 Chr.14: 36314671-36314671 on Build GRCh38
rs891784782 Chr.14: 36314705-36314705 on Build GRCh38
rs746152906 Chr.14: 36314575-36314575 on Build GRCh38
rs765773562 Chr.14: 36314717-36314717 on Build GRCh38
rs766417788 Chr.14: 36314662-36314663 on Build GRCh38
rs373403289 Chr.14: 36314726-36314726 on Build GRCh38
rs962277995 Chr.14: 36314777-36314777 on Build GRCh38
rs770532209 Chr.14: 36314850-36314850 on Build GRCh38
rs200417762 Chr.14: 36314769-36314769 on Build GRCh38
rs770447208 Chr.14: 36314840-36314840 on Build GRCh38
rs753866419 Chr.14: 36314920-36314920 on Build GRCh38
rs773730348 Chr.14: 36314845-36314845 on Build GRCh38
rs779979052 Chr.14: 36314812-36314812 on Build GRCh38
rs767745871 Chr.14: 36314878-36314878 on Build GRCh38
rs908160331 Chr.14: 36315029-36315029 on Build GRCh38
rs367921849 Chr.14: 36314942-36314942 on Build GRCh38
rs757132009 Chr.14: 36314933-36314933 on Build GRCh38
rs756593868 Chr.14: 36314971-36314971 on Build GRCh38
rs768999386 Chr.14: 36314822-36314822 on Build GRCh38
rs763848477 Chr.14: 36314899-36314899 on Build GRCh38
rs980226559 Chr.14: 36314775-36314775 on Build GRCh38
rs763345642 Chr.14: 36314847-36314847 on Build GRCh38
rs375997568 Chr.14: 36314580-36314580 on Build GRCh38
rs113771477 Chr.14: 36314889-36314889 on Build GRCh38
rs374764232 Chr.14: 36314672-36314672 on Build GRCh38
rs751271956 Chr.14: 36314720-36314720 on Build GRCh38
rs549054515 Chr.14: 36314789-36314789 on Build GRCh38
rs1053381563 Chr.14: 36314659-36314659 on Build GRCh38
rs772203591 Chr.14: 36314654-36314654 on Build GRCh38
rs747317967 Chr.14: 36314817-36314817 on Build GRCh38
rs749949801 Chr.14: 36314598-36314598 on Build GRCh38
rs192945923 Chr.14: 36314599-36314599 on Build GRCh38
rs376443009 Chr.14: 36314864-36314864 on Build GRCh38
rs764902116 Chr.14: 36314595-36314595 on Build GRCh38
rs751575011 Chr.14: 36314683-36314685 on Build GRCh38
rs368521465 Chr.14: 36314582-36314582 on Build GRCh38
rs751341509 Chr.14: 36314637-36314637 on Build GRCh38
rs756184023 Chr.14: 36315014-36315014 on Build GRCh38
rs777984459 Chr.14: 36314982-36314982 on Build GRCh38
rs1016143718 Chr.14: 36314764-36314764 on Build GRCh38
rs752688629 Chr.14: 36314642-36314642 on Build GRCh38
rs764543362 Chr.14: 36314581-36314581 on Build GRCh38
rs767095806 Chr.14: 36314869-36314869 on Build GRCh38
rs199625798 Chr.14: 36314578-36314578 on Build GRCh38
rs762392490 Chr.14: 36314594-36314594 on Build GRCh38
rs778963559 Chr.14: 36314650-36314650 on Build GRCh38
rs375762801 Chr.14: 36314673-36314673 on Build GRCh38
rs753984480 Chr.14: 36314644-36314645 on Build GRCh38

View Full Product Details