Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49892881-49893401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF6
Entrez Gene ID
382
Gene Name
ADP ribosylation factor 6
Gene Aliases
-
UniGene
Hs.525330
Gene Chromosome Location
Chr.14: 49893018-49897054 on Build GRCh38
Transcripts
NM_001663.3

SNP Detail

SNP ID Chromosome Location
rs947846417 Chr.14: 49893381-49893381 on Build GRCh38
rs371561780 Chr.14: 49893317-49893317 on Build GRCh38
rs1038671389 Chr.14: 49893133-49893133 on Build GRCh38
rs930298427 Chr.14: 49893379-49893379 on Build GRCh38
rs968677594 Chr.14: 49893238-49893241 on Build GRCh38
rs367624660 Chr.14: 49893026-49893026 on Build GRCh38
rs540912617 Chr.14: 49893137-49893137 on Build GRCh38
rs929414635 Chr.14: 49892979-49892979 on Build GRCh38
rs917210685 Chr.14: 49892936-49892936 on Build GRCh38
rs971431069 Chr.14: 49893176-49893176 on Build GRCh38
rs977893482 Chr.14: 49893288-49893288 on Build GRCh38
rs946365269 Chr.14: 49893014-49893014 on Build GRCh38
rs973580763 Chr.14: 49892913-49892913 on Build GRCh38
rs572623999 Chr.14: 49893346-49893346 on Build GRCh38
rs911654252 Chr.14: 49893298-49893298 on Build GRCh38
rs1039115635 Chr.14: 49893340-49893340 on Build GRCh38
rs980108794 Chr.14: 49893047-49893047 on Build GRCh38
rs962694146 Chr.14: 49893061-49893061 on Build GRCh38
rs181163514 Chr.14: 49893237-49893237 on Build GRCh38
rs569187172 Chr.14: 49893042-49893042 on Build GRCh38
rs945182015 Chr.14: 49893103-49893103 on Build GRCh38
rs756490454 Chr.14: 49893064-49893064 on Build GRCh38
rs944976346 Chr.14: 49892900-49892900 on Build GRCh38
rs1008583523 Chr.14: 49893132-49893132 on Build GRCh38
rs547169128 Chr.14: 49892998-49892998 on Build GRCh38
rs995448179 Chr.14: 49893156-49893156 on Build GRCh38
rs950053468 Chr.14: 49892944-49892944 on Build GRCh38
rs375262270 Chr.14: 49892968-49892968 on Build GRCh38
rs920697451 Chr.14: 49892914-49892914 on Build GRCh38
rs564509428 Chr.14: 49892903-49892903 on Build GRCh38
rs1054029220 Chr.14: 49893077-49893077 on Build GRCh38
rs957597806 Chr.14: 49893236-49893236 on Build GRCh38
rs189681437 Chr.14: 49892954-49892954 on Build GRCh38
rs929380534 Chr.14: 49892916-49892916 on Build GRCh38
rs944867706 Chr.14: 49893090-49893090 on Build GRCh38
rs1042156904 Chr.14: 49892977-49892977 on Build GRCh38
rs1004994871 Chr.14: 49893165-49893165 on Build GRCh38
rs890158642 Chr.14: 49892981-49892981 on Build GRCh38
rs1028321043 Chr.14: 49893141-49893141 on Build GRCh38
rs374397428 Chr.14: 49892962-49892962 on Build GRCh38
rs964964144 Chr.14: 49893011-49893011 on Build GRCh38
rs1018978171 Chr.14: 49893215-49893215 on Build GRCh38
rs974972072 Chr.14: 49893094-49893094 on Build GRCh38
rs1024367613 Chr.14: 49893172-49893172 on Build GRCh38
rs530276908 Chr.14: 49892996-49892996 on Build GRCh38
rs917647476 Chr.14: 49893378-49893378 on Build GRCh38
rs923615271 Chr.14: 49893126-49893126 on Build GRCh38
rs900174611 Chr.14: 49892905-49892905 on Build GRCh38
rs999347803 Chr.14: 49892948-49892948 on Build GRCh38
rs753072986 Chr.14: 49893045-49893045 on Build GRCh38
rs1053596710 Chr.14: 49892960-49892960 on Build GRCh38
rs767244109 Chr.14: 49893112-49893112 on Build GRCh38
rs1004878622 Chr.14: 49893193-49893193 on Build GRCh38
rs950806964 Chr.14: 49893017-49893017 on Build GRCh38
rs186296964 Chr.14: 49893256-49893256 on Build GRCh38
rs896329852 Chr.14: 49893030-49893030 on Build GRCh38
rs868547716 Chr.14: 49893358-49893358 on Build GRCh38
rs922164126 Chr.14: 49893100-49893100 on Build GRCh38
rs890188327 Chr.14: 49893131-49893131 on Build GRCh38
rs975297570 Chr.14: 49893234-49893234 on Build GRCh38
rs1042188373 Chr.14: 49893120-49893120 on Build GRCh38
rs983426236 Chr.14: 49892925-49892925 on Build GRCh38
rs112027915 Chr.14: 49893039-49893039 on Build GRCh38
rs566980328 Chr.14: 49893022-49893022 on Build GRCh38
rs866289812 Chr.14: 49893369-49893369 on Build GRCh38
rs551024365 Chr.14: 49892957-49892957 on Build GRCh38
rs945213560 Chr.14: 49893266-49893266 on Build GRCh38
rs973791187 Chr.14: 49893062-49893062 on Build GRCh38
rs117146976 Chr.14: 49893135-49893135 on Build GRCh38
rs539101567 Chr.14: 49893024-49893024 on Build GRCh38
rs965278340 Chr.14: 49892901-49892901 on Build GRCh38
rs934984005 Chr.14: 49893321-49893321 on Build GRCh38
rs559015031 Chr.14: 49893027-49893027 on Build GRCh38
rs1008553761 Chr.14: 49892992-49892992 on Build GRCh38
rs927276044 Chr.14: 49893217-49893217 on Build GRCh38
rs1056230439 Chr.14: 49893134-49893134 on Build GRCh38
rs950920254 Chr.14: 49893162-49893162 on Build GRCh38
rs979688135 Chr.14: 49893268-49893268 on Build GRCh38
rs926547880 Chr.14: 49892950-49892950 on Build GRCh38
rs552712480 Chr.14: 49893308-49893308 on Build GRCh38
rs1012128820 Chr.14: 49893037-49893037 on Build GRCh38
rs995429282 Chr.14: 49893012-49893012 on Build GRCh38
rs530504148 Chr.14: 49892949-49892949 on Build GRCh38
rs1056178469 Chr.14: 49893353-49893353 on Build GRCh38
rs912073206 Chr.14: 49892984-49892984 on Build GRCh38
rs898380945 Chr.14: 49893142-49893142 on Build GRCh38
rs557017124 Chr.14: 49893232-49893232 on Build GRCh38
rs778191642 Chr.14: 49893105-49893105 on Build GRCh38
rs1018294398 Chr.14: 49892999-49892999 on Build GRCh38
rs915117725 Chr.14: 49893083-49893083 on Build GRCh38
rs937865061 Chr.14: 49892958-49892958 on Build GRCh38
rs980517531 Chr.14: 49893188-49893188 on Build GRCh38
rs1027790835 Chr.14: 49893013-49893013 on Build GRCh38
rs944407552 Chr.14: 49893304-49893304 on Build GRCh38
rs1034822730 Chr.14: 49893029-49893029 on Build GRCh38
rs1010615958 Chr.14: 49892910-49892910 on Build GRCh38
rs1009280381 Chr.14: 49892969-49892969 on Build GRCh38
rs937908578 Chr.14: 49893072-49893072 on Build GRCh38
rs983506274 Chr.14: 49893082-49893082 on Build GRCh38
rs542464282 Chr.14: 49893248-49893248 on Build GRCh38

View Full Product Details