Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49893165-49893662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF6
Entrez Gene ID
382
Gene Name
ADP ribosylation factor 6
Gene Aliases
-
UniGene
Hs.525330
Gene Chromosome Location
Chr.14: 49893018-49897054 on Build GRCh38
Transcripts
NM_001663.3

SNP Detail

SNP ID Chromosome Location
rs947846417 Chr.14: 49893381-49893381 on Build GRCh38
rs371561780 Chr.14: 49893317-49893317 on Build GRCh38
rs1004878622 Chr.14: 49893193-49893193 on Build GRCh38
rs757757407 Chr.14: 49893453-49893453 on Build GRCh38
rs930298427 Chr.14: 49893379-49893379 on Build GRCh38
rs968677594 Chr.14: 49893238-49893241 on Build GRCh38
rs537902204 Chr.14: 49893526-49893526 on Build GRCh38
rs186296964 Chr.14: 49893256-49893256 on Build GRCh38
rs554905833 Chr.14: 49893605-49893605 on Build GRCh38
rs868547716 Chr.14: 49893358-49893358 on Build GRCh38
rs995342123 Chr.14: 49893394-49893394 on Build GRCh38
rs975297570 Chr.14: 49893234-49893234 on Build GRCh38
rs567044312 Chr.14: 49893472-49893472 on Build GRCh38
rs996624862 Chr.14: 49893589-49893589 on Build GRCh38
rs943975008 Chr.14: 49893553-49893553 on Build GRCh38
rs886674598 Chr.14: 49893391-49893391 on Build GRCh38
rs887006651 Chr.14: 49893539-49893539 on Build GRCh38
rs977893482 Chr.14: 49893288-49893288 on Build GRCh38
rs544191647 Chr.14: 49893412-49893412 on Build GRCh38
rs866289812 Chr.14: 49893369-49893369 on Build GRCh38
rs572623999 Chr.14: 49893346-49893346 on Build GRCh38
rs911654252 Chr.14: 49893298-49893298 on Build GRCh38
rs1039115635 Chr.14: 49893340-49893340 on Build GRCh38
rs945213560 Chr.14: 49893266-49893266 on Build GRCh38
rs59477608 Chr.14: 49893537-49893537 on Build GRCh38
rs990536648 Chr.14: 49893459-49893459 on Build GRCh38
rs574175855 Chr.14: 49893422-49893422 on Build GRCh38
rs181163514 Chr.14: 49893237-49893237 on Build GRCh38
rs1046962735 Chr.14: 49893402-49893402 on Build GRCh38
rs934984005 Chr.14: 49893321-49893321 on Build GRCh38
rs927276044 Chr.14: 49893217-49893217 on Build GRCh38
rs1024393714 Chr.14: 49893408-49893408 on Build GRCh38
rs567460586 Chr.14: 49893630-49893630 on Build GRCh38
rs536111990 Chr.14: 49893633-49893633 on Build GRCh38
rs530379898 Chr.14: 49893428-49893428 on Build GRCh38
rs957421449 Chr.14: 49893448-49893448 on Build GRCh38
rs147968439 Chr.14: 49893387-49893387 on Build GRCh38
rs1018969532 Chr.14: 49893418-49893418 on Build GRCh38
rs991807856 Chr.14: 49893479-49893479 on Build GRCh38
rs979688135 Chr.14: 49893268-49893268 on Build GRCh38
rs1029541181 Chr.14: 49893447-49893448 on Build GRCh38
rs1029076390 Chr.14: 49893603-49893603 on Build GRCh38
rs552712480 Chr.14: 49893308-49893308 on Build GRCh38
rs547230601 Chr.14: 49893471-49893471 on Build GRCh38
rs1056178469 Chr.14: 49893353-49893353 on Build GRCh38
rs557017124 Chr.14: 49893232-49893232 on Build GRCh38
rs901852424 Chr.14: 49893576-49893576 on Build GRCh38
rs1040749326 Chr.14: 49893557-49893557 on Build GRCh38
rs532731427 Chr.14: 49893499-49893499 on Build GRCh38
rs957597806 Chr.14: 49893236-49893236 on Build GRCh38
rs544729252 Chr.14: 49893385-49893385 on Build GRCh38
rs377296430 Chr.14: 49893468-49893468 on Build GRCh38
rs1046909084 Chr.14: 49893532-49893532 on Build GRCh38
rs552741821 Chr.14: 49893509-49893509 on Build GRCh38
rs35832046 Chr.14: 49893444-49893444 on Build GRCh38
rs530462938 Chr.14: 49893411-49893411 on Build GRCh38
rs780937416 Chr.14: 49893608-49893608 on Build GRCh38
rs980517531 Chr.14: 49893188-49893188 on Build GRCh38
rs112017635 Chr.14: 49893511-49893511 on Build GRCh38
rs924184581 Chr.14: 49893449-49893449 on Build GRCh38
rs1004826188 Chr.14: 49893415-49893415 on Build GRCh38
rs1018978171 Chr.14: 49893215-49893215 on Build GRCh38
rs763529690 Chr.14: 49893469-49893469 on Build GRCh38
rs1034838727 Chr.14: 49893447-49893447 on Build GRCh38
rs944407552 Chr.14: 49893304-49893304 on Build GRCh38
rs919707802 Chr.14: 49893517-49893517 on Build GRCh38
rs917647476 Chr.14: 49893378-49893378 on Build GRCh38
rs1005026477 Chr.14: 49893404-49893404 on Build GRCh38
rs931139842 Chr.14: 49893520-49893520 on Build GRCh38
rs878926625 Chr.14: 49893550-49893550 on Build GRCh38
rs996290368 Chr.14: 49893439-49893439 on Build GRCh38
rs542464282 Chr.14: 49893248-49893248 on Build GRCh38
rs903974837 Chr.14: 49893623-49893623 on Build GRCh38

View Full Product Details