Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77270751-77271250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGB
Entrez Gene ID
58157
Gene Name
neuroglobin
Gene Aliases
-
UniGene
Hs.274363
Gene Chromosome Location
Chr.14: 77265491-77271312 on Build GRCh38
Transcripts
NM_021257.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3671978 c.49A>G p.S17G Chr.14: 77270889-77270889 on Build GRCh38
7273362 c.56G>A p.S19N Chr.14: 77270882-77270882 on Build GRCh38
4052576 c.29G>A p.R10Q Chr.14: 77270909-77270909 on Build GRCh38
3667674 c.21G>T p.E7D Chr.14: 77270917-77270917 on Build GRCh38
5660816 c.31C>T p.Q11* Chr.14: 77270907-77270907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956717718 Chr.14: 77271229-77271229 on Build GRCh38
rs539113525 Chr.14: 77270859-77270859 on Build GRCh38
rs1000841511 Chr.14: 77271210-77271210 on Build GRCh38
rs780819123 Chr.14: 77270880-77270880 on Build GRCh38
rs897354665 Chr.14: 77270776-77270776 on Build GRCh38
rs768850017 Chr.14: 77270903-77270903 on Build GRCh38
rs757839467 Chr.14: 77270857-77270857 on Build GRCh38
rs919686210 Chr.14: 77271079-77271079 on Build GRCh38
rs997593401 Chr.14: 77271209-77271209 on Build GRCh38
rs918843304 Chr.14: 77271063-77271063 on Build GRCh38
rs1023684294 Chr.14: 77271198-77271198 on Build GRCh38
rs767755645 Chr.14: 77270823-77270823 on Build GRCh38
rs777835042 Chr.14: 77270864-77270864 on Build GRCh38
rs575783505 Chr.14: 77270946-77270946 on Build GRCh38
rs1046734354 Chr.14: 77271097-77271097 on Build GRCh38
rs10140032 Chr.14: 77270963-77270963 on Build GRCh38
rs912381417 Chr.14: 77271046-77271046 on Build GRCh38
rs765889701 Chr.14: 77271161-77271161 on Build GRCh38
rs980060641 Chr.14: 77271034-77271034 on Build GRCh38
rs762200098 Chr.14: 77270907-77270907 on Build GRCh38
rs1017311809 Chr.14: 77270851-77270851 on Build GRCh38
rs1005078693 Chr.14: 77271123-77271123 on Build GRCh38
rs35425974 Chr.14: 77271218-77271218 on Build GRCh38
rs375404842 Chr.14: 77270926-77270926 on Build GRCh38
rs574758439 Chr.14: 77270973-77270973 on Build GRCh38
rs1031918023 Chr.14: 77270919-77270919 on Build GRCh38
rs1030996381 Chr.14: 77271216-77271216 on Build GRCh38
rs770160840 Chr.14: 77270889-77270889 on Build GRCh38
rs1039884274 Chr.14: 77270793-77270793 on Build GRCh38
rs1049269155 Chr.14: 77270836-77270836 on Build GRCh38
rs757962176 Chr.14: 77270951-77270951 on Build GRCh38
rs762301290 Chr.14: 77270818-77270818 on Build GRCh38
rs898158820 Chr.14: 77270815-77270815 on Build GRCh38
rs764823992 Chr.14: 77270835-77270835 on Build GRCh38
rs553366744 Chr.14: 77270829-77270829 on Build GRCh38
rs891727388 Chr.14: 77271217-77271217 on Build GRCh38
rs111765077 Chr.14: 77271038-77271038 on Build GRCh38
rs565097630 Chr.14: 77271083-77271083 on Build GRCh38
rs886662907 Chr.14: 77271114-77271114 on Build GRCh38
rs1011392853 Chr.14: 77271228-77271228 on Build GRCh38
rs886695630 Chr.14: 77270875-77270875 on Build GRCh38
rs887545123 Chr.14: 77270831-77270831 on Build GRCh38
rs1038711399 Chr.14: 77271193-77271193 on Build GRCh38
rs763372679 Chr.14: 77270942-77270943 on Build GRCh38
rs760974784 Chr.14: 77270941-77270941 on Build GRCh38
rs923832763 Chr.14: 77271030-77271030 on Build GRCh38
rs754660630 Chr.14: 77270830-77270830 on Build GRCh38
rs35619737 Chr.14: 77270971-77270971 on Build GRCh38
rs1029373306 Chr.14: 77271178-77271178 on Build GRCh38
rs1039447481 Chr.14: 77271070-77271070 on Build GRCh38
rs955857909 Chr.14: 77270945-77270945 on Build GRCh38
rs145643020 Chr.14: 77271142-77271142 on Build GRCh38
rs968029955 Chr.14: 77271005-77271005 on Build GRCh38
rs942454073 Chr.14: 77270782-77270782 on Build GRCh38
rs775796017 Chr.14: 77270892-77270892 on Build GRCh38
rs545875706 Chr.14: 77271141-77271141 on Build GRCh38
rs757125125 Chr.14: 77270878-77270878 on Build GRCh38
rs961188598 Chr.14: 77270935-77270935 on Build GRCh38
rs938935865 Chr.14: 77271044-77271045 on Build GRCh38
rs1012861181 Chr.14: 77271184-77271184 on Build GRCh38
rs138171652 Chr.14: 77271202-77271202 on Build GRCh38
rs746907994 Chr.14: 77270868-77270868 on Build GRCh38
rs57515409 Chr.14: 77271057-77271057 on Build GRCh38
rs538009726 Chr.14: 77270949-77270949 on Build GRCh38
rs887507387 Chr.14: 77271179-77271180 on Build GRCh38
rs566906024 Chr.14: 77270838-77270838 on Build GRCh38
rs771364406 Chr.14: 77270983-77270983 on Build GRCh38
rs956623570 Chr.14: 77271039-77271039 on Build GRCh38
rs987153573 Chr.14: 77271040-77271040 on Build GRCh38
rs896647533 Chr.14: 77271153-77271153 on Build GRCh38
rs979434308 Chr.14: 77271021-77271021 on Build GRCh38
rs753572047 Chr.14: 77270825-77270825 on Build GRCh38
rs751555750 Chr.14: 77270983-77270983 on Build GRCh38
rs28988618 Chr.14: 77270997-77270997 on Build GRCh38
rs745635450 Chr.14: 77270882-77270882 on Build GRCh38
rs752220425 Chr.14: 77270848-77270848 on Build GRCh38
rs749380443 Chr.14: 77270894-77270894 on Build GRCh38
rs375726021 Chr.14: 77270929-77270929 on Build GRCh38
rs764875277 Chr.14: 77270943-77270943 on Build GRCh38

View Full Product Details