Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95184066-95184572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLMN
Entrez Gene ID
79789
Gene Name
calmin
Gene Aliases
-
UniGene
Hs.301478
Gene Chromosome Location
Chr.14: 95181939-95319908 on Build GRCh38
Transcripts
NM_024734.3 XM_011537158.1 XM_017021646.1

SNP Detail

SNP ID Chromosome Location
rs147881051 Chr.14: 95184129-95184129 on Build GRCh38
rs914773069 Chr.14: 95184449-95184449 on Build GRCh38
rs1055156561 Chr.14: 95184370-95184370 on Build GRCh38
rs139907492 Chr.14: 95184263-95184263 on Build GRCh38
rs761571823 Chr.14: 95184344-95184344 on Build GRCh38
rs1025082736 Chr.14: 95184412-95184412 on Build GRCh38
rs894955184 Chr.14: 95184092-95184092 on Build GRCh38
rs1042815559 Chr.14: 95184327-95184327 on Build GRCh38
rs368662048 Chr.14: 95184418-95184418 on Build GRCh38
rs967646275 Chr.14: 95184514-95184514 on Build GRCh38
rs745606242 Chr.14: 95184497-95184497 on Build GRCh38
rs1013736462 Chr.14: 95184387-95184387 on Build GRCh38
rs199567429 Chr.14: 95184329-95184330 on Build GRCh38
rs757363700 Chr.14: 95184385-95184385 on Build GRCh38
rs371681209 Chr.14: 95184427-95184427 on Build GRCh38
rs994424264 Chr.14: 95184466-95184466 on Build GRCh38
rs530985991 Chr.14: 95184542-95184542 on Build GRCh38
rs527925594 Chr.14: 95184209-95184209 on Build GRCh38
rs535953200 Chr.14: 95184405-95184405 on Build GRCh38
rs1012102293 Chr.14: 95184113-95184113 on Build GRCh38
rs560741390 Chr.14: 95184190-95184190 on Build GRCh38
rs571815505 Chr.14: 95184485-95184485 on Build GRCh38
rs553928634 Chr.14: 95184428-95184428 on Build GRCh38
rs1018384032 Chr.14: 95184367-95184367 on Build GRCh38
rs913175009 Chr.14: 95184248-95184248 on Build GRCh38
rs985093474 Chr.14: 95184549-95184549 on Build GRCh38
rs994526212 Chr.14: 95184166-95184166 on Build GRCh38
rs759024349 Chr.14: 95184173-95184173 on Build GRCh38
rs897441329 Chr.14: 95184417-95184417 on Build GRCh38
rs952607089 Chr.14: 95184133-95184133 on Build GRCh38
rs145338567 Chr.14: 95184509-95184509 on Build GRCh38
rs551746666 Chr.14: 95184368-95184368 on Build GRCh38
rs552341631 Chr.14: 95184235-95184235 on Build GRCh38
rs895206990 Chr.14: 95184386-95184386 on Build GRCh38
rs1027618280 Chr.14: 95184474-95184474 on Build GRCh38
rs577682688 Chr.14: 95184140-95184140 on Build GRCh38
rs976567997 Chr.14: 95184238-95184238 on Build GRCh38
rs903861118 Chr.14: 95184340-95184340 on Build GRCh38
rs777994760 Chr.14: 95184189-95184189 on Build GRCh38
rs1025641950 Chr.14: 95184229-95184229 on Build GRCh38
rs568328173 Chr.14: 95184381-95184381 on Build GRCh38
rs531860360 Chr.14: 95184286-95184286 on Build GRCh38
rs898421929 Chr.14: 95184155-95184155 on Build GRCh38
rs945981856 Chr.14: 95184310-95184310 on Build GRCh38
rs549784524 Chr.14: 95184376-95184376 on Build GRCh38

View Full Product Details