Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 53516824-53517372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR72
Entrez Gene ID
256764
Gene Name
WD repeat domain 72
Gene Aliases
AI2A3
UniGene
Hs.122125
Gene Chromosome Location
Chr.15: 53513741-53762878 on Build GRCh38
Transcripts
NM_001277176.1 NM_182758.3 NR_102334.1 NR_102335.1 NR_102336.1 XM_011521433.2 XM_011521435.2 XM_011521436.2 XM_011521437.2 XM_017022061.1

SNP Detail

SNP ID Chromosome Location
rs151023186 Chr.15: 53516997-53516997 on Build GRCh38
rs749876180 Chr.15: 53517172-53517172 on Build GRCh38
rs959639766 Chr.15: 53517280-53517280 on Build GRCh38
rs1054154008 Chr.15: 53516879-53516879 on Build GRCh38
rs978038735 Chr.15: 53517016-53517016 on Build GRCh38
rs945201533 Chr.15: 53516965-53516965 on Build GRCh38
rs545948620 Chr.15: 53517201-53517201 on Build GRCh38
rs995945930 Chr.15: 53517235-53517235 on Build GRCh38
rs1026275850 Chr.15: 53516869-53516869 on Build GRCh38
rs770330268 Chr.15: 53517034-53517034 on Build GRCh38
rs967916181 Chr.15: 53517262-53517262 on Build GRCh38
rs113567186 Chr.15: 53517238-53517238 on Build GRCh38
rs923796626 Chr.15: 53517286-53517286 on Build GRCh38
rs185103111 Chr.15: 53516850-53516850 on Build GRCh38
rs889480719 Chr.15: 53517098-53517099 on Build GRCh38
rs974476935 Chr.15: 53516943-53516943 on Build GRCh38
rs923820804 Chr.15: 53517030-53517030 on Build GRCh38
rs78618140 Chr.15: 53517052-53517052 on Build GRCh38
rs997960216 Chr.15: 53517014-53517014 on Build GRCh38
rs16966157 Chr.15: 53517127-53517127 on Build GRCh38
rs140879069 Chr.15: 53517111-53517111 on Build GRCh38
rs796948790 Chr.15: 53517243-53517247 on Build GRCh38
rs963563033 Chr.15: 53517156-53517158 on Build GRCh38
rs946519024 Chr.15: 53516972-53516972 on Build GRCh38
rs74015396 Chr.15: 53517200-53517200 on Build GRCh38
rs992026776 Chr.15: 53517332-53517332 on Build GRCh38
rs144192167 Chr.15: 53516933-53516933 on Build GRCh38
rs569731592 Chr.15: 53516920-53516920 on Build GRCh38
rs192271514 Chr.15: 53517343-53517343 on Build GRCh38
rs895134949 Chr.15: 53517055-53517055 on Build GRCh38
rs950419952 Chr.15: 53516870-53516870 on Build GRCh38
rs769215874 Chr.15: 53517202-53517202 on Build GRCh38
rs573198171 Chr.15: 53517338-53517338 on Build GRCh38
rs960933915 Chr.15: 53517329-53517329 on Build GRCh38
rs900944289 Chr.15: 53517001-53517001 on Build GRCh38
rs772788325 Chr.15: 53517204-53517204 on Build GRCh38
rs978365349 Chr.15: 53517271-53517271 on Build GRCh38
rs535023044 Chr.15: 53517050-53517050 on Build GRCh38
rs574919890 Chr.15: 53517091-53517091 on Build GRCh38
rs552516569 Chr.15: 53516976-53516976 on Build GRCh38
rs377194991 Chr.15: 53516864-53516864 on Build GRCh38
rs898664010 Chr.15: 53517169-53517169 on Build GRCh38
rs964345350 Chr.15: 53517224-53517224 on Build GRCh38
rs915091048 Chr.15: 53516960-53516960 on Build GRCh38
rs746055002 Chr.15: 53516994-53516994 on Build GRCh38
rs987575853 Chr.15: 53516901-53516901 on Build GRCh38
rs994707834 Chr.15: 53517198-53517198 on Build GRCh38

View Full Product Details