Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 57706977-57707484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCOM1
Entrez Gene ID
145781
Gene Name
GRINL1A complex locus 1
Gene Aliases
GRINL1A, Gcom2, MYZAP, MYZAP-POLR2M, gcom
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57591904-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018102.2 NM_015532.4

Gene Symbol
POLR2M
Entrez Gene ID
81488
Gene Name
RNA polymerase II subunit M
Gene Aliases
GCOM1, GRINL1A, Gdown, Gdown1
UniGene
Hs.437256
Gene Chromosome Location
Chr.15: 57706521-57717557 on Build GRCh38
Transcripts
NM_001018090.6 NM_001018091.6 NM_001285900.3 NR_104367.2 NR_104368.2 NR_104369.2 NR_104370.2 NR_104371.3 NM_001018102.2 NM_015532.4

SNP Detail

SNP ID Chromosome Location
rs763686413 Chr.15: 57707037-57707037 on Build GRCh38
rs552932195 Chr.15: 57707206-57707206 on Build GRCh38
rs561416693 Chr.15: 57707182-57707182 on Build GRCh38
rs779632122 Chr.15: 57707164-57707164 on Build GRCh38
rs886581881 Chr.15: 57707420-57707420 on Build GRCh38
rs754807105 Chr.15: 57706998-57706998 on Build GRCh38
rs777940094 Chr.15: 57707138-57707138 on Build GRCh38
rs568432629 Chr.15: 57707319-57707319 on Build GRCh38
rs886613141 Chr.15: 57707177-57707177 on Build GRCh38
rs747966351 Chr.15: 57707036-57707036 on Build GRCh38
rs935174968 Chr.15: 57707291-57707291 on Build GRCh38
rs771300536 Chr.15: 57706996-57706996 on Build GRCh38
rs770417011 Chr.15: 57707197-57707197 on Build GRCh38
rs1044431454 Chr.15: 57707128-57707128 on Build GRCh38
rs935211579 Chr.15: 57707101-57707101 on Build GRCh38
rs770213291 Chr.15: 57707252-57707252 on Build GRCh38
rs905864724 Chr.15: 57707332-57707332 on Build GRCh38
rs539342022 Chr.15: 57707453-57707453 on Build GRCh38
rs1049185350 Chr.15: 57707396-57707396 on Build GRCh38
rs768112424 Chr.15: 57707072-57707072 on Build GRCh38
rs758097041 Chr.15: 57707039-57707039 on Build GRCh38
rs113169869 Chr.15: 57707181-57707181 on Build GRCh38
rs761357454 Chr.15: 57707085-57707085 on Build GRCh38
rs546122446 Chr.15: 57707006-57707006 on Build GRCh38
rs114392733 Chr.15: 57706999-57706999 on Build GRCh38
rs967770229 Chr.15: 57707264-57707264 on Build GRCh38
rs948030489 Chr.15: 57707314-57707314 on Build GRCh38
rs992066412 Chr.15: 57707303-57707303 on Build GRCh38
rs113716246 Chr.15: 57707104-57707104 on Build GRCh38
rs192742731 Chr.15: 57707204-57707204 on Build GRCh38
rs764474987 Chr.15: 57707444-57707444 on Build GRCh38
rs530632374 Chr.15: 57707191-57707191 on Build GRCh38
rs775287071 Chr.15: 57707028-57707028 on Build GRCh38
rs921661816 Chr.15: 57707056-57707056 on Build GRCh38
rs748875938 Chr.15: 57707189-57707190 on Build GRCh38
rs28412380 Chr.15: 57707457-57707457 on Build GRCh38
rs753251819 Chr.15: 57707063-57707063 on Build GRCh38
rs182506077 Chr.15: 57707304-57707304 on Build GRCh38
rs189163583 Chr.15: 57707440-57707440 on Build GRCh38
rs149494746 Chr.15: 57707430-57707430 on Build GRCh38
rs532691401 Chr.15: 57707205-57707205 on Build GRCh38
rs11858659 Chr.15: 57707106-57707106 on Build GRCh38
rs11854253 Chr.15: 57707277-57707277 on Build GRCh38
rs372765766 Chr.15: 57707029-57707029 on Build GRCh38
rs771019008 Chr.15: 57707149-57707149 on Build GRCh38
rs541437398 Chr.15: 57707141-57707141 on Build GRCh38
rs756241083 Chr.15: 57707111-57707111 on Build GRCh38
rs1027784635 Chr.15: 57707166-57707166 on Build GRCh38
rs750343881 Chr.15: 57707092-57707092 on Build GRCh38
rs746311671 Chr.15: 57707175-57707175 on Build GRCh38
rs772562988 Chr.15: 57707012-57707012 on Build GRCh38
rs752925924 Chr.15: 57707107-57707107 on Build GRCh38
rs979155750 Chr.15: 57707276-57707276 on Build GRCh38
rs902086900 Chr.15: 57707460-57707460 on Build GRCh38
rs777374770 Chr.15: 57707082-57707082 on Build GRCh38
rs974493501 Chr.15: 57707024-57707024 on Build GRCh38
rs137987577 Chr.15: 57707390-57707390 on Build GRCh38
rs930680201 Chr.15: 57707345-57707345 on Build GRCh38
rs747820695 Chr.15: 57707027-57707027 on Build GRCh38
rs994960176 Chr.15: 57707148-57707148 on Build GRCh38
rs762585642 Chr.15: 57707312-57707312 on Build GRCh38
rs565182386 Chr.15: 57707087-57707087 on Build GRCh38
rs150284835 Chr.15: 57707203-57707203 on Build GRCh38
rs566064752 Chr.15: 57707246-57707246 on Build GRCh38

View Full Product Details