Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 58138131-58138639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
UniGene
Hs.104624
Gene Chromosome Location
Chr.15: 58138169-58185911 on Build GRCh38
Transcripts
NM_001320635.1 NM_001320636.1 NM_020980.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7090907 c.54G>T p.L18F Chr.15: 58138619-58138619 on Build GRCh38
1373759 c.68C>T p.A23V Chr.15: 58138633-58138633 on Build GRCh38
3502745 c.25G>A p.G9R Chr.15: 58138590-58138590 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558341486 Chr.15: 58138311-58138311 on Build GRCh38
rs771920504 Chr.15: 58138541-58138541 on Build GRCh38
rs765229101 Chr.15: 58138315-58138315 on Build GRCh38
rs956255739 Chr.15: 58138420-58138420 on Build GRCh38
rs889753506 Chr.15: 58138282-58138282 on Build GRCh38
rs1040190447 Chr.15: 58138213-58138213 on Build GRCh38
rs372592046 Chr.15: 58138548-58138548 on Build GRCh38
rs190670780 Chr.15: 58138239-58138239 on Build GRCh38
rs765070864 Chr.15: 58138561-58138561 on Build GRCh38
rs549808500 Chr.15: 58138556-58138556 on Build GRCh38
rs76035967 Chr.15: 58138334-58138334 on Build GRCh38
rs1010838065 Chr.15: 58138456-58138456 on Build GRCh38
rs768237483 Chr.15: 58138553-58138553 on Build GRCh38
rs3959686 Chr.15: 58138259-58138259 on Build GRCh38
rs914202306 Chr.15: 58138496-58138496 on Build GRCh38
rs927031836 Chr.15: 58138560-58138560 on Build GRCh38
rs183209249 Chr.15: 58138274-58138274 on Build GRCh38
rs746208655 Chr.15: 58138532-58138532 on Build GRCh38
rs971559599 Chr.15: 58138523-58138523 on Build GRCh38
rs187870637 Chr.15: 58138447-58138447 on Build GRCh38
rs1000196460 Chr.15: 58138303-58138303 on Build GRCh38
rs756348851 Chr.15: 58138615-58138615 on Build GRCh38
rs767614111 Chr.15: 58138584-58138590 on Build GRCh38
rs757230089 Chr.15: 58138474-58138474 on Build GRCh38
rs578112751 Chr.15: 58138328-58138328 on Build GRCh38
rs752707818 Chr.15: 58138433-58138433 on Build GRCh38
rs150714814 Chr.15: 58138590-58138590 on Build GRCh38
rs752868547 Chr.15: 58138597-58138597 on Build GRCh38
rs201810971 Chr.15: 58138258-58138258 on Build GRCh38
rs375764372 Chr.15: 58138609-58138609 on Build GRCh38
rs34945403 Chr.15: 58138564-58138564 on Build GRCh38
rs542588358 Chr.15: 58138581-58138581 on Build GRCh38
rs775221036 Chr.15: 58138545-58138545 on Build GRCh38
rs538553060 Chr.15: 58138305-58138305 on Build GRCh38
rs369027535 Chr.15: 58138228-58138228 on Build GRCh38
rs565029613 Chr.15: 58138362-58138362 on Build GRCh38
rs774710428 Chr.15: 58138187-58138187 on Build GRCh38
rs547176007 Chr.15: 58138193-58138193 on Build GRCh38
rs776246256 Chr.15: 58138554-58138554 on Build GRCh38
rs776541852 Chr.15: 58138210-58138210 on Build GRCh38
rs755629672 Chr.15: 58138343-58138343 on Build GRCh38
rs141236597 Chr.15: 58138494-58138494 on Build GRCh38
rs761856219 Chr.15: 58138559-58138559 on Build GRCh38

View Full Product Details