Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 411768-412264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DECR2
Entrez Gene ID
26063
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
Gene Aliases
PDCR, SDR17C1
UniGene
Hs.628831
Gene Chromosome Location
Chr.16: 401858-412487 on Build GRCh38
Transcripts
NM_020664.3

SNP Detail

SNP ID Chromosome Location
rs539439704 Chr.16: 412194-412194 on Build GRCh38
rs575202895 Chr.16: 412090-412115 on Build GRCh38
rs545832714 Chr.16: 411853-411853 on Build GRCh38
rs932160349 Chr.16: 412042-412042 on Build GRCh38
rs1028727767 Chr.16: 411851-411851 on Build GRCh38
rs942020287 Chr.16: 411974-411974 on Build GRCh38
rs748040306 Chr.16: 412127-412127 on Build GRCh38
rs973287441 Chr.16: 411899-411899 on Build GRCh38
rs1014558671 Chr.16: 411875-411875 on Build GRCh38
rs777772363 Chr.16: 412230-412230 on Build GRCh38
rs967061104 Chr.16: 411902-411902 on Build GRCh38
rs961714304 Chr.16: 411893-411893 on Build GRCh38
rs1036126287 Chr.16: 412100-412100 on Build GRCh38
rs571043342 Chr.16: 412044-412044 on Build GRCh38
rs932184137 Chr.16: 412231-412231 on Build GRCh38
rs953320039 Chr.16: 411937-411937 on Build GRCh38
rs927393659 Chr.16: 412002-412002 on Build GRCh38
rs765983049 Chr.16: 411898-411898 on Build GRCh38
rs769668190 Chr.16: 412165-412165 on Build GRCh38
rs890631322 Chr.16: 412081-412081 on Build GRCh38
rs76340810 Chr.16: 412017-412017 on Build GRCh38
rs1002041385 Chr.16: 412138-412138 on Build GRCh38
rs775462770 Chr.16: 412223-412223 on Build GRCh38
rs1046535774 Chr.16: 411852-411852 on Build GRCh38
rs138649018 Chr.16: 411843-411843 on Build GRCh38
rs897470133 Chr.16: 412101-412101 on Build GRCh38
rs561945155 Chr.16: 411863-411863 on Build GRCh38
rs564219829 Chr.16: 411832-411832 on Build GRCh38
rs1040489223 Chr.16: 412069-412094 on Build GRCh38
rs575380193 Chr.16: 412176-412176 on Build GRCh38
rs976337328 Chr.16: 411914-411914 on Build GRCh38
rs967337612 Chr.16: 412199-412199 on Build GRCh38
rs546851304 Chr.16: 412108-412108 on Build GRCh38
rs894025676 Chr.16: 412087-412087 on Build GRCh38
rs557038032 Chr.16: 412168-412168 on Build GRCh38
rs946265936 Chr.16: 411837-411837 on Build GRCh38
rs897881227 Chr.16: 411827-411827 on Build GRCh38
rs1002489611 Chr.16: 411862-411862 on Build GRCh38
rs1008617574 Chr.16: 412096-412096 on Build GRCh38
rs186144808 Chr.16: 411915-411915 on Build GRCh38
rs1011188036 Chr.16: 411886-411886 on Build GRCh38
rs1043294040 Chr.16: 412028-412028 on Build GRCh38
rs545645791 Chr.16: 411831-411831 on Build GRCh38
rs905385700 Chr.16: 412067-412067 on Build GRCh38
rs1036484548 Chr.16: 411795-411795 on Build GRCh38
rs904662890 Chr.16: 411842-411842 on Build GRCh38
rs148421637 Chr.16: 412115-412115 on Build GRCh38
rs1027560620 Chr.16: 412120-412120 on Build GRCh38
rs938671559 Chr.16: 411796-411796 on Build GRCh38
rs981042696 Chr.16: 411993-411993 on Build GRCh38
rs1035228728 Chr.16: 412139-412139 on Build GRCh38
rs562424842 Chr.16: 412012-412012 on Build GRCh38
rs568332503 Chr.16: 412113-412113 on Build GRCh38
rs957072827 Chr.16: 411881-411881 on Build GRCh38
rs1028001940 Chr.16: 411938-411938 on Build GRCh38
rs1043712271 Chr.16: 411841-411841 on Build GRCh38
rs550707440 Chr.16: 411944-411944 on Build GRCh38
rs1011197585 Chr.16: 412092-412092 on Build GRCh38
rs909116519 Chr.16: 411971-411971 on Build GRCh38
rs992846187 Chr.16: 412173-412173 on Build GRCh38
rs564027626 Chr.16: 412075-412075 on Build GRCh38
rs1030475407 Chr.16: 412131-412131 on Build GRCh38
rs551399167 Chr.16: 412032-412032 on Build GRCh38
rs946066119 Chr.16: 412027-412027 on Build GRCh38
rs992877521 Chr.16: 412016-412016 on Build GRCh38

View Full Product Details