Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4575076-4575569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf96
Entrez Gene ID
342346
Gene Name
chromosome 16 open reading frame 96
Gene Aliases
-
UniGene
Hs.684212
Gene Chromosome Location
Chr.16: 4538709-4600455 on Build GRCh38
Transcripts
NM_001145011.1 XM_005255298.3 XM_005255301.3 XM_011522475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7240129 c.989C>G p.P330R Chr.16: 4575469-4575469 on Build GRCh38
7246223 c.1006C>G p.P336A Chr.16: 4575486-4575486 on Build GRCh38
7416906 c.1071T>A p.S357R Chr.16: 4575551-4575551 on Build GRCh38
4961047 c.852G>A p.E284E Chr.16: 4575332-4575332 on Build GRCh38
6016026 c.1051G>A p.G351R Chr.16: 4575531-4575531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767954720 Chr.16: 4575128-4575128 on Build GRCh38
rs774902091 Chr.16: 4575475-4575475 on Build GRCh38
rs1046739166 Chr.16: 4575541-4575541 on Build GRCh38
rs140680907 Chr.16: 4575350-4575350 on Build GRCh38
rs772162041 Chr.16: 4575403-4575403 on Build GRCh38
rs375733441 Chr.16: 4575291-4575291 on Build GRCh38
rs769192653 Chr.16: 4575190-4575190 on Build GRCh38
rs568425453 Chr.16: 4575438-4575438 on Build GRCh38
rs189102504 Chr.16: 4575410-4575410 on Build GRCh38
rs532115559 Chr.16: 4575418-4575418 on Build GRCh38
rs955733042 Chr.16: 4575183-4575183 on Build GRCh38
rs368217239 Chr.16: 4575105-4575105 on Build GRCh38
rs964289006 Chr.16: 4575212-4575212 on Build GRCh38
rs548201732 Chr.16: 4575487-4575487 on Build GRCh38
rs867357170 Chr.16: 4575432-4575432 on Build GRCh38
rs554799912 Chr.16: 4575162-4575162 on Build GRCh38
rs1011252209 Chr.16: 4575368-4575368 on Build GRCh38
rs948520506 Chr.16: 4575299-4575299 on Build GRCh38
rs745971095 Chr.16: 4575456-4575456 on Build GRCh38
rs755620336 Chr.16: 4575173-4575173 on Build GRCh38
rs1007916707 Chr.16: 4575395-4575395 on Build GRCh38
rs907078563 Chr.16: 4575349-4575349 on Build GRCh38
rs552683971 Chr.16: 4575101-4575101 on Build GRCh38
rs186761749 Chr.16: 4575131-4575131 on Build GRCh38
rs754812114 Chr.16: 4575502-4575503 on Build GRCh38
rs762294276 Chr.16: 4575423-4575423 on Build GRCh38
rs1021894811 Chr.16: 4575411-4575411 on Build GRCh38
rs576640190 Chr.16: 4575247-4575247 on Build GRCh38
rs1022561873 Chr.16: 4575217-4575217 on Build GRCh38
rs746025683 Chr.16: 4575388-4575388 on Build GRCh38
rs919961166 Chr.16: 4575525-4575525 on Build GRCh38
rs766876725 Chr.16: 4575284-4575284 on Build GRCh38
rs183130326 Chr.16: 4575363-4575363 on Build GRCh38
rs1052824125 Chr.16: 4575358-4575358 on Build GRCh38
rs753733157 Chr.16: 4575546-4575546 on Build GRCh38
rs777259095 Chr.16: 4575419-4575419 on Build GRCh38
rs77379438 Chr.16: 4575469-4575469 on Build GRCh38
rs376258492 Chr.16: 4575224-4575224 on Build GRCh38
rs9673241 Chr.16: 4575125-4575125 on Build GRCh38
rs1019782907 Chr.16: 4575401-4575401 on Build GRCh38
rs921473823 Chr.16: 4575267-4575267 on Build GRCh38
rs1256768 Chr.16: 4575273-4575273 on Build GRCh38
rs9922488 Chr.16: 4575148-4575148 on Build GRCh38
rs777206248 Chr.16: 4575211-4575211 on Build GRCh38
rs572401766 Chr.16: 4575281-4575281 on Build GRCh38
rs76048912 Chr.16: 4575163-4575163 on Build GRCh38
rs753518030 Chr.16: 4575251-4575251 on Build GRCh38
rs975612390 Chr.16: 4575234-4575234 on Build GRCh38
rs931211566 Chr.16: 4575527-4575527 on Build GRCh38
rs1022932822 Chr.16: 4575376-4575376 on Build GRCh38
rs1023103012 Chr.16: 4575106-4575106 on Build GRCh38
rs779135465 Chr.16: 4575367-4575367 on Build GRCh38
rs772007572 Chr.16: 4575142-4575142 on Build GRCh38
rs898369187 Chr.16: 4575334-4575334 on Build GRCh38
rs968329437 Chr.16: 4575409-4575409 on Build GRCh38
rs764040396 Chr.16: 4575545-4575545 on Build GRCh38
rs545505381 Chr.16: 4575268-4575268 on Build GRCh38
rs1019727564 Chr.16: 4575195-4575195 on Build GRCh38
rs917753507 Chr.16: 4575290-4575290 on Build GRCh38
rs919868689 Chr.16: 4575503-4575503 on Build GRCh38
rs866936792 Chr.16: 4575462-4575463 on Build GRCh38
rs779622953 Chr.16: 4575246-4575246 on Build GRCh38
rs995545436 Chr.16: 4575129-4575129 on Build GRCh38
rs940022778 Chr.16: 4575277-4575277 on Build GRCh38
rs191374978 Chr.16: 4575165-4575165 on Build GRCh38
rs775572258 Chr.16: 4575412-4575412 on Build GRCh38
rs931013430 Chr.16: 4575336-4575336 on Build GRCh38
rs551849751 Chr.16: 4575429-4575429 on Build GRCh38
rs771181497 Chr.16: 4575130-4575130 on Build GRCh38
rs915963046 Chr.16: 4575468-4575468 on Build GRCh38
rs752234400 Chr.16: 4575285-4575285 on Build GRCh38
rs534567411 Chr.16: 4575472-4575472 on Build GRCh38
rs147242097 Chr.16: 4575317-4575317 on Build GRCh38
rs556718262 Chr.16: 4575240-4575240 on Build GRCh38
rs751320483 Chr.16: 4575363-4575364 on Build GRCh38
rs568319102 Chr.16: 4575519-4575519 on Build GRCh38
rs772208321 Chr.16: 4575405-4575405 on Build GRCh38
rs780032758 Chr.16: 4575106-4575106 on Build GRCh38
rs886836446 Chr.16: 4575159-4575159 on Build GRCh38

View Full Product Details