Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4800408-4800901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROGDI
Entrez Gene ID
79641
Gene Name
rogdi homolog
Gene Aliases
KTZS
UniGene
Hs.459795
Gene Chromosome Location
Chr.16: 4796962-4802950 on Build GRCh38
Transcripts
NM_024589.2 NR_046480.1 XM_006720947.3 XM_006720948.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7315521 c.301G>A p.A101T Chr.16: 4800533-4800533 on Build GRCh38
1563099 c.265C>T p.L89L Chr.16: 4800569-4800569 on Build GRCh38
1378072 c.300C>T p.F100F Chr.16: 4800534-4800534 on Build GRCh38
3957609 c.267G>A p.L89L Chr.16: 4800567-4800567 on Build GRCh38
6586388 c.304T>C p.F102L Chr.16: 4800530-4800530 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757476127 Chr.16: 4800611-4800611 on Build GRCh38
rs865861514 Chr.16: 4800556-4800556 on Build GRCh38
rs747930420 Chr.16: 4800448-4800449 on Build GRCh38
rs951463653 Chr.16: 4800775-4800775 on Build GRCh38
rs910144273 Chr.16: 4800845-4800845 on Build GRCh38
rs371996695 Chr.16: 4800827-4800827 on Build GRCh38
rs759570412 Chr.16: 4800617-4800618 on Build GRCh38
rs752580831 Chr.16: 4800524-4800524 on Build GRCh38
rs986321644 Chr.16: 4800609-4800609 on Build GRCh38
rs945932425 Chr.16: 4800428-4800428 on Build GRCh38
rs75393034 Chr.16: 4800813-4800813 on Build GRCh38
rs7200235 Chr.16: 4800640-4800640 on Build GRCh38
rs552833661 Chr.16: 4800526-4800526 on Build GRCh38
rs543743437 Chr.16: 4800664-4800664 on Build GRCh38
rs941485363 Chr.16: 4800479-4800479 on Build GRCh38
rs999207503 Chr.16: 4800605-4800605 on Build GRCh38
rs866695631 Chr.16: 4800439-4800439 on Build GRCh38
rs985988626 Chr.16: 4800789-4800789 on Build GRCh38
rs387907145 Chr.16: 4800548-4800548 on Build GRCh38
rs1012316009 Chr.16: 4800862-4800862 on Build GRCh38
rs750537590 Chr.16: 4800617-4800617 on Build GRCh38
rs559805960 Chr.16: 4800817-4800817 on Build GRCh38
rs774459203 Chr.16: 4800610-4800611 on Build GRCh38
rs888652558 Chr.16: 4800708-4800708 on Build GRCh38
rs753613367 Chr.16: 4800610-4800610 on Build GRCh38
rs1005712514 Chr.16: 4800718-4800718 on Build GRCh38
rs368831986 Chr.16: 4800521-4800521 on Build GRCh38
rs1043490336 Chr.16: 4800602-4800602 on Build GRCh38
rs375818418 Chr.16: 4800710-4800710 on Build GRCh38
rs992797311 Chr.16: 4800528-4800528 on Build GRCh38
rs775593920 Chr.16: 4800594-4800594 on Build GRCh38
rs963267452 Chr.16: 4800864-4800864 on Build GRCh38
rs771161868 Chr.16: 4800451-4800453 on Build GRCh38
rs994400338 Chr.16: 4800467-4800467 on Build GRCh38
rs765836538 Chr.16: 4800495-4800495 on Build GRCh38
rs774073956 Chr.16: 4800577-4800577 on Build GRCh38
rs777654665 Chr.16: 4800859-4800859 on Build GRCh38
rs962943021 Chr.16: 4800666-4800666 on Build GRCh38
rs1024272703 Chr.16: 4800733-4800733 on Build GRCh38
rs961006814 Chr.16: 4800435-4800435 on Build GRCh38
rs767613340 Chr.16: 4800589-4800589 on Build GRCh38
rs147294587 Chr.16: 4800672-4800672 on Build GRCh38
rs930243477 Chr.16: 4800681-4800681 on Build GRCh38
rs752909289 Chr.16: 4800688-4800688 on Build GRCh38
rs772506959 Chr.16: 4800457-4800457 on Build GRCh38
rs576947082 Chr.16: 4800489-4800489 on Build GRCh38
rs1014320198 Chr.16: 4800867-4800867 on Build GRCh38
rs184684856 Chr.16: 4800438-4800438 on Build GRCh38
rs912546744 Chr.16: 4800538-4800538 on Build GRCh38
rs900337891 Chr.16: 4800587-4800587 on Build GRCh38
rs759605875 Chr.16: 4800514-4800514 on Build GRCh38
rs941570499 Chr.16: 4800851-4800851 on Build GRCh38
rs769472180 Chr.16: 4800480-4800480 on Build GRCh38
rs866450093 Chr.16: 4800807-4800807 on Build GRCh38
rs1011154405 Chr.16: 4800597-4800597 on Build GRCh38
rs775919468 Chr.16: 4800626-4800627 on Build GRCh38
rs201101477 Chr.16: 4800604-4800604 on Build GRCh38
rs1047798559 Chr.16: 4800825-4800825 on Build GRCh38
rs977330702 Chr.16: 4800749-4800749 on Build GRCh38
rs532451022 Chr.16: 4800702-4800702 on Build GRCh38
rs996818693 Chr.16: 4800771-4800771 on Build GRCh38
rs942658592 Chr.16: 4800798-4800798 on Build GRCh38
rs557011803 Chr.16: 4800465-4800465 on Build GRCh38
rs1023916593 Chr.16: 4800863-4800863 on Build GRCh38
rs143620755 Chr.16: 4800560-4800560 on Build GRCh38
rs545861164 Chr.16: 4800517-4800517 on Build GRCh38
rs902326485 Chr.16: 4800828-4800828 on Build GRCh38
rs946581550 Chr.16: 4800822-4800822 on Build GRCh38
rs189706864 Chr.16: 4800728-4800728 on Build GRCh38
rs944453578 Chr.16: 4800799-4800799 on Build GRCh38
rs745932601 Chr.16: 4800549-4800549 on Build GRCh38
rs777014363 Chr.16: 4800559-4800559 on Build GRCh38
rs778875889 Chr.16: 4800541-4800541 on Build GRCh38
rs746381981 Chr.16: 4800454-4800454 on Build GRCh38
rs541268543 Chr.16: 4800615-4800615 on Build GRCh38
rs758447128 Chr.16: 4800619-4800619 on Build GRCh38
rs1034899126 Chr.16: 4800722-4800722 on Build GRCh38
rs1028345933 Chr.16: 4800781-4800781 on Build GRCh38
rs780059307 Chr.16: 4800620-4800620 on Build GRCh38
rs944655165 Chr.16: 4800555-4800555 on Build GRCh38
rs967337947 Chr.16: 4800737-4800737 on Build GRCh38
rs374098348 Chr.16: 4800585-4800585 on Build GRCh38
rs572561598 Chr.16: 4800583-4800583 on Build GRCh38
rs754421468 Chr.16: 4800437-4800437 on Build GRCh38
rs1025833011 Chr.16: 4800487-4800487 on Build GRCh38
rs918696536 Chr.16: 4800762-4800762 on Build GRCh38
rs780503540 Chr.16: 4800557-4800557 on Build GRCh38
rs368283919 Chr.16: 4800488-4800488 on Build GRCh38
rs1041352943 Chr.16: 4800453-4800453 on Build GRCh38
rs760445194 Chr.16: 4800531-4800531 on Build GRCh38
rs376597130 Chr.16: 4800558-4800558 on Build GRCh38
rs1056101647 Chr.16: 4800858-4800858 on Build GRCh38
rs561219384 Chr.16: 4800621-4800621 on Build GRCh38
rs775888608 Chr.16: 4800460-4800460 on Build GRCh38
rs958993433 Chr.16: 4800527-4800527 on Build GRCh38
rs757436102 Chr.16: 4800540-4800540 on Build GRCh38
rs548104184 Chr.16: 4800824-4800824 on Build GRCh38
rs568326961 Chr.16: 4800871-4800871 on Build GRCh38
rs373890537 Chr.16: 4800629-4800630 on Build GRCh38
rs559599196 Chr.16: 4800783-4800783 on Build GRCh38
rs888611327 Chr.16: 4800507-4800507 on Build GRCh38
rs1014267896 Chr.16: 4800731-4800731 on Build GRCh38
rs537326410 Chr.16: 4800877-4800877 on Build GRCh38
rs986855782 Chr.16: 4800779-4800779 on Build GRCh38
rs910354870 Chr.16: 4800790-4800790 on Build GRCh38
rs758702587 Chr.16: 4800689-4800689 on Build GRCh38
rs755444169 Chr.16: 4800629-4800629 on Build GRCh38
rs772631534 Chr.16: 4800554-4800554 on Build GRCh38
rs772142185 Chr.16: 4800625-4800626 on Build GRCh38
rs367928686 Chr.16: 4800766-4800766 on Build GRCh38
rs770605828 Chr.16: 4800567-4800567 on Build GRCh38
rs371657994 Chr.16: 4800534-4800534 on Build GRCh38
rs747545149 Chr.16: 4800622-4800622 on Build GRCh38
rs1047326132 Chr.16: 4800693-4800693 on Build GRCh38
rs372097881 Chr.16: 4800500-4800500 on Build GRCh38
rs371783831 Chr.16: 4800533-4800533 on Build GRCh38
rs999238605 Chr.16: 4800850-4800850 on Build GRCh38

View Full Product Details