Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31110309-31110811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCKDK
Entrez Gene ID
10295
Gene Name
branched chain ketoacid dehydrogenase kinase
Gene Aliases
BCKDKD, BDK
UniGene
Hs.513520
Gene Chromosome Location
Chr.16: 31108294-31117651 on Build GRCh38
Transcripts
NM_001122957.2 NM_001271926.1 NM_005881.3 XM_017022859.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556653 c.699G>A p.K233K Chr.16: 31110744-31110744 on Build GRCh38
1478828 c.654_656delCGG p.G219delG Chr.16: 31110699-31110701 on Build GRCh38
6610581 c.552G>C p.K184N Chr.16: 31110409-31110409 on Build GRCh38
1478827 c.559C>T p.R187C Chr.16: 31110416-31110416 on Build GRCh38
1377588 c.670C>T p.R224C Chr.16: 31110715-31110715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs14235 Chr.16: 31110472-31110472 on Build GRCh38
rs761322791 Chr.16: 31110420-31110420 on Build GRCh38
rs761806585 Chr.16: 31110644-31110655 on Build GRCh38
rs573728679 Chr.16: 31110622-31110622 on Build GRCh38
rs201843180 Chr.16: 31110709-31110709 on Build GRCh38
rs758510551 Chr.16: 31110418-31110418 on Build GRCh38
rs200689545 Chr.16: 31110737-31110737 on Build GRCh38
rs368764723 Chr.16: 31110638-31110638 on Build GRCh38
rs752891319 Chr.16: 31110397-31110397 on Build GRCh38
rs188092823 Chr.16: 31110413-31110413 on Build GRCh38
rs561728909 Chr.16: 31110621-31110621 on Build GRCh38
rs558120140 Chr.16: 31110484-31110484 on Build GRCh38
rs753438536 Chr.16: 31110347-31110347 on Build GRCh38
rs764416605 Chr.16: 31110648-31110648 on Build GRCh38
rs745889690 Chr.16: 31110658-31110658 on Build GRCh38
rs745800138 Chr.16: 31110459-31110459 on Build GRCh38
rs553956491 Chr.16: 31110444-31110444 on Build GRCh38
rs147210405 Chr.16: 31110716-31110716 on Build GRCh38
rs370032941 Chr.16: 31110682-31110682 on Build GRCh38
rs768365083 Chr.16: 31110483-31110483 on Build GRCh38
rs761417171 Chr.16: 31110489-31110489 on Build GRCh38
rs777877602 Chr.16: 31110392-31110392 on Build GRCh38
rs776044468 Chr.16: 31110720-31110720 on Build GRCh38
rs768472775 Chr.16: 31110681-31110681 on Build GRCh38
rs753178162 Chr.16: 31110641-31110641 on Build GRCh38
rs898604240 Chr.16: 31110437-31110437 on Build GRCh38
rs727503823 Chr.16: 31110771-31110771 on Build GRCh38
rs781670286 Chr.16: 31110434-31110434 on Build GRCh38
rs116079081 Chr.16: 31110656-31110656 on Build GRCh38
rs374568685 Chr.16: 31110435-31110435 on Build GRCh38
rs755577638 Chr.16: 31110433-31110433 on Build GRCh38
rs370950849 Chr.16: 31110391-31110391 on Build GRCh38
rs375380019 Chr.16: 31110671-31110671 on Build GRCh38
rs920073282 Chr.16: 31110341-31110341 on Build GRCh38
rs139120593 Chr.16: 31110461-31110461 on Build GRCh38
rs376498393 Chr.16: 31110331-31110331 on Build GRCh38
rs1032069191 Chr.16: 31110567-31110567 on Build GRCh38
rs1054129437 Chr.16: 31110728-31110728 on Build GRCh38
rs980680430 Chr.16: 31110604-31110604 on Build GRCh38
rs771021107 Chr.16: 31110458-31110458 on Build GRCh38
rs752120800 Chr.16: 31110368-31110368 on Build GRCh38
rs565911171 Chr.16: 31110655-31110655 on Build GRCh38
rs746468438 Chr.16: 31110370-31110371 on Build GRCh38
rs757631135 Chr.16: 31110757-31110757 on Build GRCh38
rs762568662 Chr.16: 31110521-31110521 on Build GRCh38
rs750015325 Chr.16: 31110490-31110490 on Build GRCh38
rs182442686 Chr.16: 31110351-31110351 on Build GRCh38
rs963711173 Chr.16: 31110549-31110549 on Build GRCh38
rs543375050 Chr.16: 31110570-31110570 on Build GRCh38
rs768073750 Chr.16: 31110539-31110539 on Build GRCh38
rs754213472 Chr.16: 31110756-31110756 on Build GRCh38
rs775418426 Chr.16: 31110471-31110471 on Build GRCh38
rs199722503 Chr.16: 31110333-31110333 on Build GRCh38
rs775304293 Chr.16: 31110412-31110412 on Build GRCh38
rs979103906 Chr.16: 31110557-31110557 on Build GRCh38
rs776418633 Chr.16: 31110536-31110539 on Build GRCh38
rs1009283606 Chr.16: 31110744-31110744 on Build GRCh38
rs745733651 Chr.16: 31110500-31110500 on Build GRCh38
rs905825367 Chr.16: 31110416-31110416 on Build GRCh38
rs772935236 Chr.16: 31110780-31110781 on Build GRCh38
rs201839495 Chr.16: 31110402-31110402 on Build GRCh38
rs150773521 Chr.16: 31110445-31110445 on Build GRCh38
rs764514534 Chr.16: 31110748-31110748 on Build GRCh38
rs1018942913 Chr.16: 31110508-31110508 on Build GRCh38
rs961697319 Chr.16: 31110526-31110526 on Build GRCh38
rs949246621 Chr.16: 31110580-31110580 on Build GRCh38
rs554631884 Chr.16: 31110714-31110714 on Build GRCh38
rs974123511 Chr.16: 31110568-31110568 on Build GRCh38
rs758475834 Chr.16: 31110664-31110664 on Build GRCh38
rs368040635 Chr.16: 31110699-31110699 on Build GRCh38
rs1032004173 Chr.16: 31110726-31110726 on Build GRCh38
rs770918327 Chr.16: 31110408-31110408 on Build GRCh38
rs1008477370 Chr.16: 31110503-31110503 on Build GRCh38
rs752102857 Chr.16: 31110426-31110426 on Build GRCh38
rs747827915 Chr.16: 31110698-31110698 on Build GRCh38
rs926546915 Chr.16: 31110607-31110607 on Build GRCh38
rs768262297 Chr.16: 31110382-31110383 on Build GRCh38
rs144100726 Chr.16: 31110373-31110373 on Build GRCh38
rs754059958 Chr.16: 31110652-31110652 on Build GRCh38
rs756453432 Chr.16: 31110647-31110647 on Build GRCh38
rs765164352 Chr.16: 31110664-31110664 on Build GRCh38
rs774214869 Chr.16: 31110409-31110409 on Build GRCh38
rs762515151 Chr.16: 31110715-31110715 on Build GRCh38
rs756741732 Chr.16: 31110348-31110348 on Build GRCh38
rs764857104 Chr.16: 31110423-31110423 on Build GRCh38
rs748345434 Chr.16: 31110388-31110388 on Build GRCh38
rs756441659 Chr.16: 31110436-31110436 on Build GRCh38
rs554174460 Chr.16: 31110569-31110569 on Build GRCh38

View Full Product Details