Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67166242-67166732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSF4
Entrez Gene ID
3299
Gene Name
heat shock transcription factor 4
Gene Aliases
CTM, CTRCT5
UniGene
Hs.512156
Gene Chromosome Location
Chr.16: 67163385-67169945 on Build GRCh38
Transcripts
NM_001040667.2 NM_001538.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4591857 c.512T>G p.V171G Chr.16: 67166346-67166346 on Build GRCh38
4852343 c.600G>T p.P200P Chr.16: 67166596-67166596 on Build GRCh38
2994222 c.489G>C p.Q163H Chr.16: 67166323-67166323 on Build GRCh38
460503 c.574C>T p.L192F Chr.16: 67166570-67166570 on Build GRCh38
6682460 c.594G>A p.A198A Chr.16: 67166590-67166590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765098750 Chr.16: 67166414-67166420 on Build GRCh38
rs758084052 Chr.16: 67166664-67166664 on Build GRCh38
rs933255894 Chr.16: 67166472-67166473 on Build GRCh38
rs950680548 Chr.16: 67166267-67166267 on Build GRCh38
rs777140776 Chr.16: 67166424-67166424 on Build GRCh38
rs770632910 Chr.16: 67166673-67166673 on Build GRCh38
rs760450511 Chr.16: 67166407-67166407 on Build GRCh38
rs761316430 Chr.16: 67166343-67166343 on Build GRCh38
rs552547561 Chr.16: 67166497-67166497 on Build GRCh38
rs767567922 Chr.16: 67166602-67166602 on Build GRCh38
rs375017943 Chr.16: 67166332-67166332 on Build GRCh38
rs199734886 Chr.16: 67166368-67166368 on Build GRCh38
rs765507007 Chr.16: 67166431-67166431 on Build GRCh38
rs757189381 Chr.16: 67166410-67166410 on Build GRCh38
rs766888101 Chr.16: 67166349-67166349 on Build GRCh38
rs973312180 Chr.16: 67166263-67166263 on Build GRCh38
rs1052100454 Chr.16: 67166480-67166480 on Build GRCh38
rs947204738 Chr.16: 67166419-67166419 on Build GRCh38
rs377009893 Chr.16: 67166357-67166357 on Build GRCh38
rs778816975 Chr.16: 67166621-67166621 on Build GRCh38
rs377630856 Chr.16: 67166370-67166370 on Build GRCh38
rs371071221 Chr.16: 67166381-67166381 on Build GRCh38
rs759887807 Chr.16: 67166430-67166430 on Build GRCh38
rs983817524 Chr.16: 67166528-67166528 on Build GRCh38
rs781320816 Chr.16: 67166513-67166513 on Build GRCh38
rs754459542 Chr.16: 67166270-67166270 on Build GRCh38
rs751639850 Chr.16: 67166595-67166595 on Build GRCh38
rs138295025 Chr.16: 67166510-67166510 on Build GRCh38
rs920616056 Chr.16: 67166266-67166266 on Build GRCh38
rs1012283750 Chr.16: 67166521-67166521 on Build GRCh38
rs35824613 Chr.16: 67166662-67166663 on Build GRCh38
rs372829523 Chr.16: 67166471-67166471 on Build GRCh38
rs569813935 Chr.16: 67166423-67166423 on Build GRCh38
rs755476901 Chr.16: 67166278-67166278 on Build GRCh38
rs758556801 Chr.16: 67166304-67166304 on Build GRCh38
rs1052878983 Chr.16: 67166453-67166453 on Build GRCh38
rs765709564 Chr.16: 67166365-67166365 on Build GRCh38
rs761820702 Chr.16: 67166598-67166598 on Build GRCh38
rs895523534 Chr.16: 67166455-67166455 on Build GRCh38
rs746930815 Chr.16: 67166665-67166665 on Build GRCh38
rs376624407 Chr.16: 67166504-67166504 on Build GRCh38
rs778567385 Chr.16: 67166529-67166529 on Build GRCh38
rs372135974 Chr.16: 67166550-67166550 on Build GRCh38
rs1016736139 Chr.16: 67166330-67166330 on Build GRCh38
rs779844642 Chr.16: 67166378-67166378 on Build GRCh38
rs754860432 Chr.16: 67166516-67166516 on Build GRCh38
rs189553377 Chr.16: 67166279-67166279 on Build GRCh38
rs776484170 Chr.16: 67166574-67166574 on Build GRCh38
rs773762218 Chr.16: 67166342-67166342 on Build GRCh38
rs201403716 Chr.16: 67166346-67166346 on Build GRCh38
rs758416940 Chr.16: 67166649-67166649 on Build GRCh38
rs180744555 Chr.16: 67166399-67166399 on Build GRCh38
rs764542115 Chr.16: 67166591-67166591 on Build GRCh38
rs757450852 Chr.16: 67166373-67166373 on Build GRCh38
rs994782458 Chr.16: 67166506-67166506 on Build GRCh38
rs750723656 Chr.16: 67166606-67166606 on Build GRCh38
rs372473149 Chr.16: 67166344-67166344 on Build GRCh38
rs914235834 Chr.16: 67166687-67166687 on Build GRCh38
rs757499561 Chr.16: 67166511-67166511 on Build GRCh38
rs769646890 Chr.16: 67166326-67166326 on Build GRCh38
rs979674078 Chr.16: 67166664-67166664 on Build GRCh38
rs773056248 Chr.16: 67166345-67166345 on Build GRCh38
rs988842171 Chr.16: 67166653-67166653 on Build GRCh38
rs755472426 Chr.16: 67166364-67166364 on Build GRCh38
rs753317849 Chr.16: 67166438-67166438 on Build GRCh38
rs777796744 Chr.16: 67166651-67166651 on Build GRCh38
rs753339378 Chr.16: 67166332-67166332 on Build GRCh38
rs772816388 Chr.16: 67166400-67166400 on Build GRCh38
rs749039219 Chr.16: 67166379-67166379 on Build GRCh38
rs774282666 Chr.16: 67166382-67166382 on Build GRCh38
rs773108951 Chr.16: 67166679-67166679 on Build GRCh38
rs969995114 Chr.16: 67166261-67166261 on Build GRCh38
rs749270991 Chr.16: 67166337-67166337 on Build GRCh38
rs950713309 Chr.16: 67166519-67166519 on Build GRCh38
rs369594985 Chr.16: 67166323-67166323 on Build GRCh38
rs374589300 Chr.16: 67166589-67166589 on Build GRCh38
rs763541172 Chr.16: 67166590-67166590 on Build GRCh38
rs770722728 Chr.16: 67166564-67166564 on Build GRCh38
rs748155702 Chr.16: 67166536-67166536 on Build GRCh38
rs373573708 Chr.16: 67166508-67166508 on Build GRCh38
rs766091707 Chr.16: 67166412-67166412 on Build GRCh38
rs375086636 Chr.16: 67166286-67166286 on Build GRCh38
rs369154621 Chr.16: 67166341-67166341 on Build GRCh38
rs757301978 Chr.16: 67166509-67166510 on Build GRCh38
rs746454603 Chr.16: 67166563-67166563 on Build GRCh38
rs374553565 Chr.16: 67166369-67166369 on Build GRCh38
rs772110943 Chr.16: 67166541-67166541 on Build GRCh38
rs753585363 Chr.16: 67166515-67166515 on Build GRCh38
rs752383385 Chr.16: 67166637-67166637 on Build GRCh38
rs756259393 Chr.16: 67166607-67166607 on Build GRCh38
rs748021821 Chr.16: 67166387-67166387 on Build GRCh38
rs968487777 Chr.16: 67166703-67166703 on Build GRCh38
rs1023258223 Chr.16: 67166470-67166470 on Build GRCh38
rs777997735 Chr.16: 67166306-67166306 on Build GRCh38
rs759109686 Chr.16: 67166587-67166587 on Build GRCh38
rs1039577199 Chr.16: 67166339-67166339 on Build GRCh38
rs538698506 Chr.16: 67166457-67166457 on Build GRCh38

View Full Product Details