Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 77978265-77978809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAT1L
Entrez Gene ID
57687
Gene Name
vesicle amine transport 1-like
Gene Aliases
-
UniGene
Hs.461405
Gene Chromosome Location
Chr.16: 77788564-77980107 on Build GRCh38
Transcripts
NM_020927.2

SNP Detail

SNP ID Chromosome Location
rs912089334 Chr.16: 77978456-77978456 on Build GRCh38
rs569692417 Chr.16: 77978775-77978775 on Build GRCh38
rs143838699 Chr.16: 77978468-77978468 on Build GRCh38
rs867917196 Chr.16: 77978684-77978684 on Build GRCh38
rs767134731 Chr.16: 77978397-77978397 on Build GRCh38
rs372083542 Chr.16: 77978755-77978755 on Build GRCh38
rs409260 Chr.16: 77978332-77978332 on Build GRCh38
rs539474974 Chr.16: 77978431-77978431 on Build GRCh38
rs559402709 Chr.16: 77978494-77978496 on Build GRCh38
rs996479579 Chr.16: 77978506-77978506 on Build GRCh38
rs527913080 Chr.16: 77978762-77978762 on Build GRCh38
rs147238744 Chr.16: 77978562-77978562 on Build GRCh38
rs994787930 Chr.16: 77978653-77978653 on Build GRCh38
rs367834862 Chr.16: 77978785-77978785 on Build GRCh38
rs945699062 Chr.16: 77978500-77978500 on Build GRCh38
rs542016127 Chr.16: 77978712-77978712 on Build GRCh38
rs539439574 Chr.16: 77978780-77978780 on Build GRCh38
rs773582631 Chr.16: 77978297-77978297 on Build GRCh38
rs141568673 Chr.16: 77978667-77978667 on Build GRCh38
rs117559201 Chr.16: 77978385-77978385 on Build GRCh38
rs986689587 Chr.16: 77978396-77978396 on Build GRCh38
rs982115471 Chr.16: 77978625-77978625 on Build GRCh38
rs1001772389 Chr.16: 77978351-77978351 on Build GRCh38
rs966245915 Chr.16: 77978694-77978694 on Build GRCh38
rs897784764 Chr.16: 77978635-77978635 on Build GRCh38
rs928269576 Chr.16: 77978361-77978361 on Build GRCh38
rs760173293 Chr.16: 77978672-77978672 on Build GRCh38
rs186600624 Chr.16: 77978773-77978773 on Build GRCh38
rs1128695 Chr.16: 77978757-77978757 on Build GRCh38
rs961430786 Chr.16: 77978623-77978623 on Build GRCh38
rs138355165 Chr.16: 77978334-77978334 on Build GRCh38
rs953520200 Chr.16: 77978390-77978390 on Build GRCh38
rs528993497 Chr.16: 77978728-77978728 on Build GRCh38
rs1040064272 Chr.16: 77978701-77978701 on Build GRCh38
rs962025437 Chr.16: 77978384-77978384 on Build GRCh38
rs959676937 Chr.16: 77978664-77978664 on Build GRCh38
rs915381672 Chr.16: 77978382-77978382 on Build GRCh38
rs1035039655 Chr.16: 77978779-77978779 on Build GRCh38
rs181776039 Chr.16: 77978693-77978693 on Build GRCh38
rs1027519653 Chr.16: 77978510-77978510 on Build GRCh38
rs923720040 Chr.16: 77978692-77978692 on Build GRCh38
rs150889966 Chr.16: 77978682-77978682 on Build GRCh38
rs931932368 Chr.16: 77978746-77978746 on Build GRCh38
rs887616814 Chr.16: 77978760-77978760 on Build GRCh38
rs3751771 Chr.16: 77978774-77978774 on Build GRCh38
rs1014552487 Chr.16: 77978614-77978614 on Build GRCh38
rs952851967 Chr.16: 77978312-77978312 on Build GRCh38
rs759210728 Chr.16: 77978387-77978387 on Build GRCh38
rs984159049 Chr.16: 77978316-77978316 on Build GRCh38
rs988406958 Chr.16: 77978472-77978472 on Build GRCh38
rs111871756 Chr.16: 77978354-77978354 on Build GRCh38
rs912923149 Chr.16: 77978289-77978289 on Build GRCh38
rs532937454 Chr.16: 77978691-77978691 on Build GRCh38
rs925659149 Chr.16: 77978743-77978743 on Build GRCh38
rs405764 Chr.16: 77978319-77978319 on Build GRCh38
rs895663437 Chr.16: 77978589-77978589 on Build GRCh38
rs547667276 Chr.16: 77978440-77978440 on Build GRCh38
rs556527557 Chr.16: 77978786-77978786 on Build GRCh38
rs576590605 Chr.16: 77978630-77978630 on Build GRCh38
rs190682510 Chr.16: 77978376-77978376 on Build GRCh38
rs556549512 Chr.16: 77978372-77978372 on Build GRCh38
rs576517964 Chr.16: 77978501-77978501 on Build GRCh38
rs1015596369 Chr.16: 77978620-77978620 on Build GRCh38
rs978411435 Chr.16: 77978730-77978730 on Build GRCh38
rs568330779 Chr.16: 77978758-77978758 on Build GRCh38
rs746332490 Chr.16: 77978507-77978507 on Build GRCh38
rs866376567 Chr.16: 77978608-77978608 on Build GRCh38
rs3751770 Chr.16: 77978538-77978538 on Build GRCh38
rs991260704 Chr.16: 77978756-77978756 on Build GRCh38
rs1056010714 Chr.16: 77978584-77978584 on Build GRCh38
rs938258698 Chr.16: 77978375-77978375 on Build GRCh38
rs139367345 Chr.16: 77978629-77978629 on Build GRCh38
rs902184107 Chr.16: 77978402-77978402 on Build GRCh38
rs946827039 Chr.16: 77978673-77978673 on Build GRCh38
rs1027638380 Chr.16: 77978665-77978665 on Build GRCh38

View Full Product Details