Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 85710085-85710573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf74
Entrez Gene ID
404550
Gene Name
chromosome 16 open reading frame 74
Gene Aliases
-
UniGene
Hs.461655
Gene Chromosome Location
Chr.16: 85707517-85751083 on Build GRCh38
Transcripts
NM_206967.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4852854 c.223G>A p.E75K Chr.16: 85710266-85710266 on Build GRCh38
4141937 c.306G>A p.P102P Chr.16: 85710183-85710183 on Build GRCh38
6407363 c.283A>G p.T95A Chr.16: 85710206-85710206 on Build GRCh38
3755160 c.189A>T p.Q63H Chr.16: 85710300-85710300 on Build GRCh38
1198198 c.325+2T>C p.? Chr.16: 85710162-85710162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747980872 Chr.16: 85710350-85710353 on Build GRCh38
rs118106422 Chr.16: 85710472-85710472 on Build GRCh38
rs919105724 Chr.16: 85710395-85710395 on Build GRCh38
rs750583912 Chr.16: 85710167-85710167 on Build GRCh38
rs369228649 Chr.16: 85710357-85710357 on Build GRCh38
rs555442564 Chr.16: 85710249-85710249 on Build GRCh38
rs533328181 Chr.16: 85710491-85710491 on Build GRCh38
rs914883199 Chr.16: 85710109-85710109 on Build GRCh38
rs748067415 Chr.16: 85710229-85710229 on Build GRCh38
rs920546704 Chr.16: 85710434-85710434 on Build GRCh38
rs781002103 Chr.16: 85710307-85710307 on Build GRCh38
rs779026239 Chr.16: 85710179-85710179 on Build GRCh38
rs914072415 Chr.16: 85710375-85710375 on Build GRCh38
rs548675860 Chr.16: 85710111-85710111 on Build GRCh38
rs781653770 Chr.16: 85710166-85710166 on Build GRCh38
rs377716191 Chr.16: 85710227-85710227 on Build GRCh38
rs772762860 Chr.16: 85710130-85710130 on Build GRCh38
rs770628496 Chr.16: 85710197-85710197 on Build GRCh38
rs748838192 Chr.16: 85710325-85710325 on Build GRCh38
rs990462560 Chr.16: 85710522-85710522 on Build GRCh38
rs780331075 Chr.16: 85710221-85710221 on Build GRCh38
rs530890823 Chr.16: 85710462-85710462 on Build GRCh38
rs201669939 Chr.16: 85710174-85710174 on Build GRCh38
rs140423977 Chr.16: 85710523-85710523 on Build GRCh38
rs766570607 Chr.16: 85710273-85710275 on Build GRCh38
rs931737043 Chr.16: 85710110-85710110 on Build GRCh38
rs547325559 Chr.16: 85710515-85710515 on Build GRCh38
rs993170853 Chr.16: 85710484-85710484 on Build GRCh38
rs1011585762 Chr.16: 85710471-85710471 on Build GRCh38
rs762297690 Chr.16: 85710261-85710261 on Build GRCh38
rs372383869 Chr.16: 85710259-85710259 on Build GRCh38
rs139037414 Chr.16: 85710468-85710468 on Build GRCh38
rs1004250159 Chr.16: 85710448-85710448 on Build GRCh38
rs751878722 Chr.16: 85710157-85710157 on Build GRCh38
rs542456065 Chr.16: 85710437-85710437 on Build GRCh38
rs753450624 Chr.16: 85710421-85710421 on Build GRCh38
rs775052111 Chr.16: 85710260-85710260 on Build GRCh38
rs529047452 Chr.16: 85710105-85710105 on Build GRCh38
rs997048179 Chr.16: 85710343-85710343 on Build GRCh38
rs919055434 Chr.16: 85710262-85710262 on Build GRCh38
rs1032246010 Chr.16: 85710198-85710198 on Build GRCh38
rs768774637 Chr.16: 85710122-85710122 on Build GRCh38
rs767239587 Chr.16: 85710129-85710129 on Build GRCh38
rs759548700 Chr.16: 85710470-85710470 on Build GRCh38
rs937695585 Chr.16: 85710412-85710412 on Build GRCh38
rs767753044 Chr.16: 85710233-85710252 on Build GRCh38
rs927613887 Chr.16: 85710406-85710406 on Build GRCh38
rs755292101 Chr.16: 85710285-85710285 on Build GRCh38
rs373401837 Chr.16: 85710144-85710144 on Build GRCh38
rs1026524047 Chr.16: 85710486-85710486 on Build GRCh38
rs771741917 Chr.16: 85710183-85710183 on Build GRCh38
rs773133735 Chr.16: 85710426-85710426 on Build GRCh38
rs751825593 Chr.16: 85710273-85710281 on Build GRCh38
rs374582245 Chr.16: 85710172-85710172 on Build GRCh38
rs966057797 Chr.16: 85710481-85710481 on Build GRCh38
rs769688679 Chr.16: 85710272-85710273 on Build GRCh38
rs748355555 Chr.16: 85710366-85710366 on Build GRCh38
rs911680439 Chr.16: 85710232-85710232 on Build GRCh38
rs927728564 Chr.16: 85710513-85710513 on Build GRCh38
rs115139300 Chr.16: 85710213-85710213 on Build GRCh38
rs111385851 Chr.16: 85710271-85710271 on Build GRCh38
rs774232213 Chr.16: 85710123-85710123 on Build GRCh38
rs775682890 Chr.16: 85710117-85710117 on Build GRCh38
rs930542153 Chr.16: 85710150-85710150 on Build GRCh38
rs374680936 Chr.16: 85710214-85710214 on Build GRCh38
rs965389118 Chr.16: 85710339-85710339 on Build GRCh38
rs1041143666 Chr.16: 85710449-85710449 on Build GRCh38
rs761137086 Chr.16: 85710338-85710338 on Build GRCh38
rs1028562371 Chr.16: 85710349-85710349 on Build GRCh38
rs996825303 Chr.16: 85710461-85710461 on Build GRCh38
rs754975130 Chr.16: 85710243-85710243 on Build GRCh38
rs1045690096 Chr.16: 85710279-85710279 on Build GRCh38
rs571195534 Chr.16: 85710163-85710163 on Build GRCh38
rs774927447 Chr.16: 85710322-85710322 on Build GRCh38
rs981862193 Chr.16: 85710396-85710396 on Build GRCh38
rs200347042 Chr.16: 85710297-85710297 on Build GRCh38
rs906204885 Chr.16: 85710254-85710254 on Build GRCh38
rs768084861 Chr.16: 85710269-85710269 on Build GRCh38
rs747903346 Chr.16: 85710182-85710182 on Build GRCh38
rs376689280 Chr.16: 85710168-85710168 on Build GRCh38
rs755848520 Chr.16: 85710384-85710384 on Build GRCh38
rs750689222 Chr.16: 85710270-85710270 on Build GRCh38
rs368866028 Chr.16: 85710442-85710442 on Build GRCh38
rs760230112 Chr.16: 85710184-85710184 on Build GRCh38
rs374677968 Chr.16: 85710335-85710335 on Build GRCh38
rs779987426 Chr.16: 85710171-85710171 on Build GRCh38
rs147002684 Chr.16: 85710361-85710361 on Build GRCh38
rs541222718 Chr.16: 85710136-85710136 on Build GRCh38
rs141454251 Chr.16: 85710300-85710300 on Build GRCh38
rs577189860 Chr.16: 85710316-85710316 on Build GRCh38
rs1047523351 Chr.16: 85710438-85710438 on Build GRCh38
rs762344436 Chr.16: 85710210-85710210 on Build GRCh38
rs756428810 Chr.16: 85710169-85710169 on Build GRCh38
rs189450880 Chr.16: 85710240-85710240 on Build GRCh38
rs776284296 Chr.16: 85710257-85710257 on Build GRCh38
rs1018608551 Chr.16: 85710371-85710371 on Build GRCh38
rs767773782 Chr.16: 85710212-85710212 on Build GRCh38
rs201787608 Chr.16: 85710199-85710199 on Build GRCh38
rs779183719 Chr.16: 85710228-85710228 on Build GRCh38
rs375249399 Chr.16: 85710282-85710282 on Build GRCh38
rs906421913 Chr.16: 85710106-85710106 on Build GRCh38
rs745907453 Chr.16: 85710116-85710116 on Build GRCh38
rs368142094 Chr.16: 85710258-85710258 on Build GRCh38
rs759075732 Chr.16: 85710200-85710200 on Build GRCh38
rs2305357 Chr.16: 85710126-85710126 on Build GRCh38
rs529091049 Chr.16: 85710409-85710409 on Build GRCh38
rs369824478 Chr.16: 85710296-85710296 on Build GRCh38
rs550982675 Chr.16: 85710155-85710155 on Build GRCh38
rs374494410 Chr.16: 85710234-85710234 on Build GRCh38
rs745511811 Chr.16: 85710312-85710312 on Build GRCh38
rs969969252 Chr.16: 85710543-85710543 on Build GRCh38
rs749180890 Chr.16: 85710120-85710120 on Build GRCh38
rs181372894 Chr.16: 85710451-85710451 on Build GRCh38
rs761134549 Chr.16: 85710274-85710274 on Build GRCh38
rs752836530 Chr.16: 85710287-85710287 on Build GRCh38
rs112376688 Chr.16: 85710207-85710207 on Build GRCh38
rs1049062549 Chr.16: 85710151-85710151 on Build GRCh38
rs890070347 Chr.16: 85710469-85710469 on Build GRCh38
rs765170842 Chr.16: 85710206-85710207 on Build GRCh38
rs769498389 Chr.16: 85710315-85710315 on Build GRCh38
rs766820240 Chr.16: 85710353-85710353 on Build GRCh38
rs199610353 Chr.16: 85710334-85710334 on Build GRCh38
rs1045724625 Chr.16: 85710430-85710430 on Build GRCh38
rs981850894 Chr.16: 85710498-85710498 on Build GRCh38
rs144813253 Chr.16: 85710387-85710387 on Build GRCh38
rs752036541 Chr.16: 85710205-85710205 on Build GRCh38
rs758449740 Chr.16: 85710276-85710278 on Build GRCh38
rs967322472 Chr.16: 85710479-85710479 on Build GRCh38
rs764533571 Chr.16: 85710202-85710202 on Build GRCh38
rs887175499 Chr.16: 85710305-85710305 on Build GRCh38
rs765802106 Chr.16: 85710143-85710143 on Build GRCh38
rs758202202 Chr.16: 85710288-85710288 on Build GRCh38
rs368435391 Chr.16: 85710156-85710156 on Build GRCh38

View Full Product Details