Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44931971-44932474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF18B
Entrez Gene ID
146909
Gene Name
kinesin family member 18B
Gene Aliases
-
UniGene
Hs.135094
Gene Chromosome Location
Chr.17: 44924711-44947746 on Build GRCh38
Transcripts
NM_001264573.1 NM_001265577.1 XM_011524385.2 XM_011524386.2 XM_011524387.2 XM_011524388.2 XM_011524389.2 XM_011524390.2 XM_011524391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6883564 c.1403C>T p.S468L Chr.17: 44932105-44932105 on Build GRCh38
3518186 c.1373C>T p.S458F Chr.17: 44932108-44932108 on Build GRCh38
5715661 c.1270C>T p.P424S Chr.17: 44932211-44932211 on Build GRCh38
4908261 c.1305G>A p.Q435Q Chr.17: 44932203-44932203 on Build GRCh38
3518191 c.1280C>T p.P427L Chr.17: 44932228-44932228 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751828520 Chr.17: 44932135-44932135 on Build GRCh38
rs751521043 Chr.17: 44932019-44932019 on Build GRCh38
rs949022033 Chr.17: 44932197-44932197 on Build GRCh38
rs1023835812 Chr.17: 44932449-44932449 on Build GRCh38
rs773129249 Chr.17: 44932223-44932223 on Build GRCh38
rs1005275370 Chr.17: 44932446-44932446 on Build GRCh38
rs768375974 Chr.17: 44932167-44932167 on Build GRCh38
rs776066453 Chr.17: 44932233-44932233 on Build GRCh38
rs774243763 Chr.17: 44932215-44932215 on Build GRCh38
rs200150609 Chr.17: 44932250-44932250 on Build GRCh38
rs543450495 Chr.17: 44932432-44932432 on Build GRCh38
rs952552379 Chr.17: 44932101-44932101 on Build GRCh38
rs746672667 Chr.17: 44932042-44932050 on Build GRCh38
rs73307487 Chr.17: 44932030-44932030 on Build GRCh38
rs780312006 Chr.17: 44932258-44932258 on Build GRCh38
rs374781031 Chr.17: 44932035-44932035 on Build GRCh38
rs550488305 Chr.17: 44932130-44932130 on Build GRCh38
rs757818902 Chr.17: 44932139-44932139 on Build GRCh38
rs545491869 Chr.17: 44932328-44932328 on Build GRCh38
rs745924164 Chr.17: 44932063-44932063 on Build GRCh38
rs777930906 Chr.17: 44932036-44932036 on Build GRCh38
rs747562767 Chr.17: 44932046-44932046 on Build GRCh38
rs993615769 Chr.17: 44932426-44932426 on Build GRCh38
rs1025810908 Chr.17: 44932157-44932157 on Build GRCh38
rs563064595 Chr.17: 44932248-44932248 on Build GRCh38
rs910524033 Chr.17: 44932385-44932387 on Build GRCh38
rs759214882 Chr.17: 44932234-44932234 on Build GRCh38
rs768164382 Chr.17: 44932071-44932072 on Build GRCh38
rs766391737 Chr.17: 44932377-44932377 on Build GRCh38
rs750984882 Chr.17: 44932249-44932249 on Build GRCh38
rs149127441 Chr.17: 44932218-44932218 on Build GRCh38
rs754564392 Chr.17: 44932126-44932126 on Build GRCh38
rs765884599 Chr.17: 44932231-44932231 on Build GRCh38
rs768722842 Chr.17: 44932198-44932198 on Build GRCh38
rs898119937 Chr.17: 44932423-44932423 on Build GRCh38
rs369789498 Chr.17: 44932025-44932025 on Build GRCh38
rs554670566 Chr.17: 44932413-44932413 on Build GRCh38
rs749544071 Chr.17: 44932186-44932186 on Build GRCh38
rs753492545 Chr.17: 44932123-44932123 on Build GRCh38
rs530568919 Chr.17: 44932100-44932100 on Build GRCh38
rs761599223 Chr.17: 44932024-44932024 on Build GRCh38
rs1040584670 Chr.17: 44932391-44932391 on Build GRCh38
rs908218464 Chr.17: 44932177-44932177 on Build GRCh38
rs753116000 Chr.17: 44932034-44932034 on Build GRCh38
rs752022013 Chr.17: 44932242-44932242 on Build GRCh38
rs775853344 Chr.17: 44932073-44932073 on Build GRCh38
rs764763428 Chr.17: 44932237-44932237 on Build GRCh38
rs529815111 Chr.17: 44932066-44932066 on Build GRCh38
rs569649838 Chr.17: 44932087-44932087 on Build GRCh38
rs143296698 Chr.17: 44932329-44932329 on Build GRCh38
rs766085170 Chr.17: 44932113-44932113 on Build GRCh38
rs548342229 Chr.17: 44932077-44932077 on Build GRCh38
rs1041773419 Chr.17: 44932312-44932312 on Build GRCh38
rs762832621 Chr.17: 44932011-44932011 on Build GRCh38
rs761841829 Chr.17: 44932092-44932092 on Build GRCh38
rs775981653 Chr.17: 44932227-44932229 on Build GRCh38
rs944737525 Chr.17: 44932390-44932390 on Build GRCh38
rs956524877 Chr.17: 44932301-44932301 on Build GRCh38
rs369406463 Chr.17: 44932110-44932110 on Build GRCh38
rs780225641 Chr.17: 44932178-44932178 on Build GRCh38
rs151266142 Chr.17: 44932332-44932332 on Build GRCh38
rs771290031 Chr.17: 44932047-44932047 on Build GRCh38
rs736866 Chr.17: 44932429-44932429 on Build GRCh38
rs772827212 Chr.17: 44932104-44932104 on Build GRCh38
rs760539140 Chr.17: 44932226-44932226 on Build GRCh38
rs764196996 Chr.17: 44932014-44932014 on Build GRCh38
rs373708940 Chr.17: 44932216-44932216 on Build GRCh38
rs756270961 Chr.17: 44932173-44932173 on Build GRCh38
rs554366493 Chr.17: 44932267-44932267 on Build GRCh38
rs755278443 Chr.17: 44932291-44932291 on Build GRCh38
rs781360700 Chr.17: 44932061-44932061 on Build GRCh38
rs267604913 Chr.17: 44932185-44932185 on Build GRCh38

View Full Product Details