Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50094645-50095138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDK2
Entrez Gene ID
5164
Gene Name
pyruvate dehydrogenase kinase 2
Gene Aliases
PDHK2, PDKII
UniGene
Hs.256667
Gene Chromosome Location
Chr.17: 50094737-50111369 on Build GRCh38
Transcripts
NM_001199898.1

SNP Detail

SNP ID Chromosome Location
rs114777669 Chr.17: 50094735-50094735 on Build GRCh38
rs1054063197 Chr.17: 50094855-50094855 on Build GRCh38
rs1046439972 Chr.17: 50094813-50094813 on Build GRCh38
rs1027559590 Chr.17: 50094988-50094988 on Build GRCh38
rs538061007 Chr.17: 50095085-50095085 on Build GRCh38
rs555508380 Chr.17: 50094997-50094997 on Build GRCh38
rs968816086 Chr.17: 50095017-50095017 on Build GRCh38
rs115904699 Chr.17: 50094805-50094805 on Build GRCh38
rs929249649 Chr.17: 50095088-50095088 on Build GRCh38
rs757665884 Chr.17: 50095105-50095105 on Build GRCh38
rs144399275 Chr.17: 50095097-50095097 on Build GRCh38
rs1023033629 Chr.17: 50094944-50094944 on Build GRCh38
rs868370864 Chr.17: 50094930-50094930 on Build GRCh38
rs991324007 Chr.17: 50095002-50095002 on Build GRCh38
rs1009072005 Chr.17: 50094908-50094908 on Build GRCh38
rs952163135 Chr.17: 50094668-50094668 on Build GRCh38
rs890131689 Chr.17: 50095105-50095106 on Build GRCh38
rs71353617 Chr.17: 50094893-50094897 on Build GRCh38
rs943064731 Chr.17: 50095052-50095052 on Build GRCh38
rs1040039453 Chr.17: 50095056-50095056 on Build GRCh38
rs973194549 Chr.17: 50094986-50094986 on Build GRCh38
rs775441901 Chr.17: 50094742-50094742 on Build GRCh38
rs1040724744 Chr.17: 50094846-50094846 on Build GRCh38
rs935526201 Chr.17: 50095065-50095065 on Build GRCh38
rs370978951 Chr.17: 50094895-50094899 on Build GRCh38
rs915535825 Chr.17: 50095073-50095073 on Build GRCh38
rs16948691 Chr.17: 50094913-50094913 on Build GRCh38
rs16942045 Chr.17: 50094892-50094896 on Build GRCh38
rs910271346 Chr.17: 50095044-50095044 on Build GRCh38
rs774749489 Chr.17: 50094891-50094892 on Build GRCh38
rs979167501 Chr.17: 50095043-50095043 on Build GRCh38
rs577897706 Chr.17: 50095099-50095099 on Build GRCh38
rs568642199 Chr.17: 50094718-50094718 on Build GRCh38
rs111564639 Chr.17: 50094731-50094731 on Build GRCh38
rs546202148 Chr.17: 50094681-50094681 on Build GRCh38
rs191687639 Chr.17: 50094876-50094876 on Build GRCh38
rs981710098 Chr.17: 50094716-50094716 on Build GRCh38
rs77936529 Chr.17: 50094758-50094758 on Build GRCh38
rs376927285 Chr.17: 50094896-50094896 on Build GRCh38
rs970060513 Chr.17: 50094953-50094953 on Build GRCh38
rs935487839 Chr.17: 50094854-50094854 on Build GRCh38
rs16948695 Chr.17: 50095003-50095003 on Build GRCh38
rs183797192 Chr.17: 50094685-50094685 on Build GRCh38
rs984825161 Chr.17: 50094684-50094684 on Build GRCh38
rs943012705 Chr.17: 50094814-50094814 on Build GRCh38
rs924029668 Chr.17: 50095063-50095063 on Build GRCh38
rs970008068 Chr.17: 50094703-50094703 on Build GRCh38
rs931339255 Chr.17: 50094750-50094750 on Build GRCh38
rs981426958 Chr.17: 50094954-50094954 on Build GRCh38
rs1022979980 Chr.17: 50094689-50094689 on Build GRCh38
rs1036375487 Chr.17: 50094955-50094955 on Build GRCh38
rs1053949773 Chr.17: 50095068-50095068 on Build GRCh38
rs946460484 Chr.17: 50094788-50094788 on Build GRCh38
rs1044528929 Chr.17: 50095109-50095109 on Build GRCh38
rs932224633 Chr.17: 50095087-50095087 on Build GRCh38
rs546606678 Chr.17: 50094882-50094882 on Build GRCh38
rs1006312656 Chr.17: 50094921-50094921 on Build GRCh38
rs550955299 Chr.17: 50094807-50094807 on Build GRCh38
rs74819030 Chr.17: 50094895-50094895 on Build GRCh38
rs956314254 Chr.17: 50094983-50094983 on Build GRCh38
rs187252027 Chr.17: 50094809-50094809 on Build GRCh38

View Full Product Details